Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 217683135-217683408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA17
Entrez Gene ID
128153
Gene Name
spermatogenesis associated 17
Gene Aliases
IQCH, MSRG-11, MSRG11
UniGene
Hs.171130
Gene Chromosome Location
Chr.1: 217631324-217868151 on Build GRCh38
Transcripts
NM_138796.3 XM_005273052.1 XM_006711165.3 XM_011509194.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3746796 c.318G>A p.R106R Chr.1: 217683284-217683284 on Build GRCh38
1338899 c.304T>C p.W102R Chr.1: 217683270-217683270 on Build GRCh38
5558450 c.351G>C p.L117F Chr.1: 217683317-217683317 on Build GRCh38
7290618 c.322C>T p.R108W Chr.1: 217683288-217683288 on Build GRCh38
4028295 c.303A>T p.R101R Chr.1: 217683269-217683269 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748168217 Chr.1: 217683301-217683304 on Build GRCh38
rs764481239 Chr.1: 217683361-217683361 on Build GRCh38
rs753965734 Chr.1: 217683236-217683236 on Build GRCh38
rs764882354 Chr.1: 217683252-217683252 on Build GRCh38
rs756781689 Chr.1: 217683270-217683270 on Build GRCh38
rs762816320 Chr.1: 217683327-217683327 on Build GRCh38
rs906275094 Chr.1: 217683180-217683180 on Build GRCh38
rs148969039 Chr.1: 217683336-217683336 on Build GRCh38
rs1000722676 Chr.1: 217683164-217683164 on Build GRCh38
rs200104955 Chr.1: 217683268-217683268 on Build GRCh38
rs758472487 Chr.1: 217683265-217683265 on Build GRCh38
rs112296282 Chr.1: 217683234-217683234 on Build GRCh38
rs747605699 Chr.1: 217683165-217683165 on Build GRCh38
rs200997435 Chr.1: 217683214-217683214 on Build GRCh38
rs770848508 Chr.1: 217683293-217683293 on Build GRCh38
rs146794589 Chr.1: 217683288-217683288 on Build GRCh38
rs766609228 Chr.1: 217683235-217683235 on Build GRCh38
rs1032654660 Chr.1: 217683169-217683169 on Build GRCh38
rs747672691 Chr.1: 217683346-217683346 on Build GRCh38
rs12035909 Chr.1: 217683280-217683280 on Build GRCh38
rs755790434 Chr.1: 217683350-217683350 on Build GRCh38
rs200251214 Chr.1: 217683241-217683241 on Build GRCh38
rs765488293 Chr.1: 217683323-217683323 on Build GRCh38
rs781704723 Chr.1: 217683279-217683279 on Build GRCh38
rs759734832 Chr.1: 217683315-217683315 on Build GRCh38
rs773348571 Chr.1: 217683219-217683219 on Build GRCh38
rs777819202 Chr.1: 217683267-217683267 on Build GRCh38
rs765861852 Chr.1: 217683229-217683229 on Build GRCh38
rs755432590 Chr.1: 217683213-217683213 on Build GRCh38
rs956677987 Chr.1: 217683194-217683194 on Build GRCh38
rs754805520 Chr.1: 217683238-217683238 on Build GRCh38
rs780932192 Chr.1: 217683273-217683273 on Build GRCh38
rs749936041 Chr.1: 217683348-217683348 on Build GRCh38
rs988203605 Chr.1: 217683201-217683201 on Build GRCh38
rs35503371 Chr.1: 217683289-217683289 on Build GRCh38
rs772148924 Chr.1: 217683172-217683172 on Build GRCh38
rs375338290 Chr.1: 217683274-217683274 on Build GRCh38
rs554485013 Chr.1: 217683277-217683277 on Build GRCh38
rs368343860 Chr.1: 217683306-217683306 on Build GRCh38
rs12068167 Chr.1: 217683284-217683284 on Build GRCh38
rs764147672 Chr.1: 217683330-217683330 on Build GRCh38
rs79785026 Chr.1: 217683354-217683354 on Build GRCh38
rs752520409 Chr.1: 217683258-217683258 on Build GRCh38
rs372279654 Chr.1: 217683343-217683343 on Build GRCh38
rs752471214 Chr.1: 217683326-217683326 on Build GRCh38

View Full Product Details