Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161120530-161120795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NIT1
Entrez Gene ID
4817
Gene Name
nitrilase 1
Gene Aliases
-
UniGene
Hs.146406
Gene Chromosome Location
Chr.1: 161118072-161125446 on Build GRCh38
Transcripts
NM_001185092.1 NM_001185093.1 NM_001185094.1 NM_005600.2 XM_005245214.2 XM_005245215.4 XM_005245216.3 XM_006711341.3 XM_017001371.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3984408 c.928C>A p.P310T Chr.1: 161120754-161120754 on Build GRCh38
1180247 c.835G>T p.G279* Chr.1: 161120616-161120616 on Build GRCh38
4707014 c.764G>A p.G255D Chr.1: 161120590-161120590 on Build GRCh38
4707016 c.860G>A p.R287Q Chr.1: 161120686-161120686 on Build GRCh38
4819076 c.814G>C p.E272Q Chr.1: 161120640-161120640 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747000266 Chr.1: 161120589-161120589 on Build GRCh38
rs976623483 Chr.1: 161120700-161120700 on Build GRCh38
rs962328125 Chr.1: 161120656-161120656 on Build GRCh38
rs772746414 Chr.1: 161120600-161120600 on Build GRCh38
rs765287502 Chr.1: 161120681-161120681 on Build GRCh38
rs78463473 Chr.1: 161120622-161120622 on Build GRCh38
rs746060088 Chr.1: 161120575-161120575 on Build GRCh38
rs754296542 Chr.1: 161120617-161120617 on Build GRCh38
rs1005089076 Chr.1: 161120768-161120768 on Build GRCh38
rs778982723 Chr.1: 161120626-161120626 on Build GRCh38
rs35745502 Chr.1: 161120705-161120706 on Build GRCh38
rs779029144 Chr.1: 161120574-161120574 on Build GRCh38
rs776319738 Chr.1: 161120594-161120594 on Build GRCh38
rs768709303 Chr.1: 161120592-161120592 on Build GRCh38
rs764801882 Chr.1: 161120598-161120598 on Build GRCh38
rs768762045 Chr.1: 161120642-161120642 on Build GRCh38
rs780298602 Chr.1: 161120695-161120695 on Build GRCh38
rs762579888 Chr.1: 161120654-161120654 on Build GRCh38
rs772765955 Chr.1: 161120652-161120652 on Build GRCh38
rs11555192 Chr.1: 161120705-161120705 on Build GRCh38
rs201109189 Chr.1: 161120632-161120632 on Build GRCh38
rs1802399 Chr.1: 161120732-161120732 on Build GRCh38
rs774026600 Chr.1: 161120711-161120711 on Build GRCh38
rs368649283 Chr.1: 161120647-161120647 on Build GRCh38
rs143827217 Chr.1: 161120704-161120704 on Build GRCh38
rs200514054 Chr.1: 161120685-161120685 on Build GRCh38
rs762609568 Chr.1: 161120603-161120603 on Build GRCh38
rs761052452 Chr.1: 161120664-161120664 on Build GRCh38
rs749380684 Chr.1: 161120551-161120552 on Build GRCh38
rs184919184 Chr.1: 161120555-161120555 on Build GRCh38
rs753105093 Chr.1: 161120604-161120604 on Build GRCh38
rs761383084 Chr.1: 161120595-161120595 on Build GRCh38
rs775294207 Chr.1: 161120588-161120588 on Build GRCh38
rs761471860 Chr.1: 161120722-161120722 on Build GRCh38
rs34359959 Chr.1: 161120677-161120677 on Build GRCh38
rs757526889 Chr.1: 161120570-161120570 on Build GRCh38
rs762290156 Chr.1: 161120725-161120725 on Build GRCh38
rs376379316 Chr.1: 161120749-161120749 on Build GRCh38
rs749144819 Chr.1: 161120757-161120757 on Build GRCh38
rs893811820 Chr.1: 161120584-161120584 on Build GRCh38
rs532171421 Chr.1: 161120701-161120701 on Build GRCh38
rs765742286 Chr.1: 161120726-161120726 on Build GRCh38
rs746087991 Chr.1: 161120631-161120631 on Build GRCh38
rs755237351 Chr.1: 161120748-161120748 on Build GRCh38
rs749338832 Chr.1: 161120563-161120563 on Build GRCh38
rs766329208 Chr.1: 161120738-161120738 on Build GRCh38
rs775886525 Chr.1: 161120657-161120657 on Build GRCh38
rs754345246 Chr.1: 161120669-161120669 on Build GRCh38
rs748813720 Chr.1: 161120708-161120708 on Build GRCh38
rs758588525 Chr.1: 161120686-161120686 on Build GRCh38
rs753571571 Chr.1: 161120702-161120702 on Build GRCh38
rs200668987 Chr.1: 161120721-161120721 on Build GRCh38
rs778333607 Chr.1: 161120770-161120770 on Build GRCh38
rs776731252 Chr.1: 161120645-161120645 on Build GRCh38
rs1045450892 Chr.1: 161120742-161120742 on Build GRCh38
rs764564526 Chr.1: 161120613-161120613 on Build GRCh38
rs377471257 Chr.1: 161120692-161120692 on Build GRCh38
rs751845527 Chr.1: 161120743-161120743 on Build GRCh38
rs779778998 Chr.1: 161120637-161120637 on Build GRCh38
rs746769362 Chr.1: 161120638-161120638 on Build GRCh38
rs769203266 Chr.1: 161120649-161120649 on Build GRCh38
rs375048732 Chr.1: 161120698-161120698 on Build GRCh38
rs1053791840 Chr.1: 161120580-161120580 on Build GRCh38
rs559900921 Chr.1: 161120689-161120689 on Build GRCh38
rs531389777 Chr.1: 161120773-161120773 on Build GRCh38
rs886681997 Chr.1: 161120764-161120764 on Build GRCh38
rs770300800 Chr.1: 161120709-161120709 on Build GRCh38
rs776963968 Chr.1: 161120724-161120724 on Build GRCh38
rs199912934 Chr.1: 161120665-161120665 on Build GRCh38
rs777790277 Chr.1: 161120756-161120756 on Build GRCh38
rs367895808 Chr.1: 161120611-161120611 on Build GRCh38
rs763114911 Chr.1: 161120737-161120737 on Build GRCh38
rs751782007 Chr.1: 161120697-161120697 on Build GRCh38
rs190215650 Chr.1: 161120578-161120578 on Build GRCh38

View Full Product Details