Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49807695-49807956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBA7
Entrez Gene ID
7318
Gene Name
ubiquitin like modifier activating enzyme 7
Gene Aliases
D8, UBA1B, UBE1L, UBE2
UniGene
Hs.16695
Gene Chromosome Location
Chr.3: 49805205-49813958 on Build GRCh38
Transcripts
NM_003335.2 XM_005265430.2 XM_006713321.2 XM_011534070.1 XM_011534071.1 XM_011534072.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6814871 c.2644C>T p.R882C Chr.3: 49807807-49807807 on Build GRCh38
5951143 c.2616G>A p.V872V Chr.3: 49807835-49807835 on Build GRCh38
6504087 c.2599G>A p.E867K Chr.3: 49807852-49807852 on Build GRCh38
6504088 c.2575G>A p.A859T Chr.3: 49807876-49807876 on Build GRCh38
4118610 c.2636G>T p.S879I Chr.3: 49807815-49807815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890375355 Chr.3: 49807882-49807882 on Build GRCh38
rs769615859 Chr.3: 49807768-49807768 on Build GRCh38
rs746254806 Chr.3: 49807742-49807742 on Build GRCh38
rs772115998 Chr.3: 49807759-49807759 on Build GRCh38
rs35209290 Chr.3: 49807734-49807735 on Build GRCh38
rs750709731 Chr.3: 49807812-49807812 on Build GRCh38
rs754360075 Chr.3: 49807802-49807802 on Build GRCh38
rs114468755 Chr.3: 49807737-49807737 on Build GRCh38
rs893284300 Chr.3: 49807786-49807786 on Build GRCh38
rs751262898 Chr.3: 49807783-49807783 on Build GRCh38
rs759246920 Chr.3: 49807864-49807864 on Build GRCh38
rs761721832 Chr.3: 49807767-49807767 on Build GRCh38
rs61760190 Chr.3: 49807819-49807819 on Build GRCh38
rs776084376 Chr.3: 49807861-49807871 on Build GRCh38
rs375121069 Chr.3: 49807739-49807739 on Build GRCh38
rs758738535 Chr.3: 49807761-49807761 on Build GRCh38
rs748266061 Chr.3: 49807827-49807827 on Build GRCh38
rs760259940 Chr.3: 49807904-49807904 on Build GRCh38
rs762839252 Chr.3: 49807770-49807770 on Build GRCh38
rs773224728 Chr.3: 49807833-49807833 on Build GRCh38
rs1039128199 Chr.3: 49807911-49807911 on Build GRCh38
rs778235456 Chr.3: 49807720-49807720 on Build GRCh38
rs755192095 Chr.3: 49807922-49807922 on Build GRCh38
rs368889184 Chr.3: 49807921-49807921 on Build GRCh38
rs755085168 Chr.3: 49807825-49807825 on Build GRCh38
rs758804273 Chr.3: 49807920-49807920 on Build GRCh38
rs1008278625 Chr.3: 49807829-49807829 on Build GRCh38
rs770757975 Chr.3: 49807852-49807852 on Build GRCh38
rs757639008 Chr.3: 49807726-49807726 on Build GRCh38
rs561051731 Chr.3: 49807906-49807906 on Build GRCh38
rs767173352 Chr.3: 49807889-49807889 on Build GRCh38
rs549814453 Chr.3: 49807932-49807932 on Build GRCh38
rs145881892 Chr.3: 49807859-49807859 on Build GRCh38
rs200484624 Chr.3: 49807847-49807847 on Build GRCh38
rs372069280 Chr.3: 49807721-49807721 on Build GRCh38
rs780450954 Chr.3: 49807760-49807760 on Build GRCh38
rs776678207 Chr.3: 49807764-49807764 on Build GRCh38
rs780346278 Chr.3: 49807803-49807803 on Build GRCh38
rs767085492 Chr.3: 49807790-49807790 on Build GRCh38
rs758808886 Chr.3: 49807818-49807818 on Build GRCh38
rs952742360 Chr.3: 49807910-49807910 on Build GRCh38
rs750984592 Chr.3: 49807913-49807913 on Build GRCh38
rs747274065 Chr.3: 49807822-49807822 on Build GRCh38
rs994038047 Chr.3: 49807853-49807853 on Build GRCh38
rs752078696 Chr.3: 49807890-49807890 on Build GRCh38
rs577926401 Chr.3: 49807779-49807779 on Build GRCh38
rs150961796 Chr.3: 49807716-49807716 on Build GRCh38
rs769579838 Chr.3: 49807832-49807832 on Build GRCh38
rs545334951 Chr.3: 49807806-49807806 on Build GRCh38
rs990973642 Chr.3: 49807715-49807715 on Build GRCh38
rs768467918 Chr.3: 49807797-49807797 on Build GRCh38
rs768905524 Chr.3: 49807763-49807763 on Build GRCh38
rs762634620 Chr.3: 49807850-49807850 on Build GRCh38
rs199939644 Chr.3: 49807824-49807824 on Build GRCh38
rs947617899 Chr.3: 49807792-49807792 on Build GRCh38
rs772818030 Chr.3: 49807769-49807769 on Build GRCh38
rs759020462 Chr.3: 49807785-49807785 on Build GRCh38

View Full Product Details