Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 43075397-43075670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTLL1
Entrez Gene ID
25809
Gene Name
tubulin tyrosine ligase like 1
Gene Aliases
C22orf7, HS323M22B
UniGene
Hs.660298
Gene Chromosome Location
Chr.22: 43039516-43089428 on Build GRCh38
Transcripts
NM_012263.4 NR_027779.1 XM_006724225.3 XM_011530108.2 XM_011530110.2 XM_017028751.1 XM_017028752.1 XM_017028753.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6840275 c.71G>T p.G24V Chr.22: 43075516-43075516 on Build GRCh38
4508893 c.78C>T p.V26V Chr.22: 43075509-43075509 on Build GRCh38
7395693 c.85A>G p.T29A Chr.22: 43075502-43075502 on Build GRCh38
258271 c.33C>T p.I11I Chr.22: 43075554-43075554 on Build GRCh38
5002296 c.4G>T p.A2S Chr.22: 43075583-43075583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745361101 Chr.22: 43075499-43075499 on Build GRCh38
rs751810276 Chr.22: 43075566-43075567 on Build GRCh38
rs769033810 Chr.22: 43075631-43075631 on Build GRCh38
rs770146740 Chr.22: 43075437-43075437 on Build GRCh38
rs759008118 Chr.22: 43075545-43075546 on Build GRCh38
rs746432577 Chr.22: 43075464-43075464 on Build GRCh38
rs559982825 Chr.22: 43075510-43075510 on Build GRCh38
rs776142109 Chr.22: 43075504-43075504 on Build GRCh38
rs201076959 Chr.22: 43075523-43075523 on Build GRCh38
rs752640384 Chr.22: 43075573-43075573 on Build GRCh38
rs757565193 Chr.22: 43075575-43075575 on Build GRCh38
rs57929300 Chr.22: 43075431-43075431 on Build GRCh38
rs772551763 Chr.22: 43075465-43075465 on Build GRCh38
rs747563643 Chr.22: 43075617-43075617 on Build GRCh38
rs543640499 Chr.22: 43075435-43075435 on Build GRCh38
rs758434812 Chr.22: 43075585-43075585 on Build GRCh38
rs79088468 Chr.22: 43075577-43075577 on Build GRCh38
rs373694695 Chr.22: 43075599-43075599 on Build GRCh38
rs148475049 Chr.22: 43075554-43075554 on Build GRCh38
rs377044202 Chr.22: 43075430-43075430 on Build GRCh38
rs974301481 Chr.22: 43075474-43075474 on Build GRCh38
rs199974695 Chr.22: 43075570-43075570 on Build GRCh38
rs767582941 Chr.22: 43075572-43075572 on Build GRCh38
rs747409374 Chr.22: 43075566-43075566 on Build GRCh38
rs772570197 Chr.22: 43075563-43075563 on Build GRCh38
rs373850414 Chr.22: 43075425-43075425 on Build GRCh38
rs755922436 Chr.22: 43075542-43075542 on Build GRCh38
rs1040311820 Chr.22: 43075612-43075612 on Build GRCh38
rs775929588 Chr.22: 43075564-43075564 on Build GRCh38
rs752552171 Chr.22: 43075531-43075531 on Build GRCh38
rs745940943 Chr.22: 43075562-43075562 on Build GRCh38
rs147435458 Chr.22: 43075502-43075502 on Build GRCh38
rs370302008 Chr.22: 43075555-43075555 on Build GRCh38
rs555266269 Chr.22: 43075450-43075450 on Build GRCh38
rs781226647 Chr.22: 43075577-43075578 on Build GRCh38
rs759060978 Chr.22: 43075571-43075571 on Build GRCh38
rs751560056 Chr.22: 43075426-43075426 on Build GRCh38
rs771002467 Chr.22: 43075454-43075454 on Build GRCh38
rs762486116 Chr.22: 43075494-43075494 on Build GRCh38
rs867310988 Chr.22: 43075575-43075577 on Build GRCh38
rs745972481 Chr.22: 43075578-43075578 on Build GRCh38
rs753260986 Chr.22: 43075554-43075555 on Build GRCh38
rs931863480 Chr.22: 43075634-43075634 on Build GRCh38
rs764522452 Chr.22: 43075560-43075563 on Build GRCh38
rs755034605 Chr.22: 43075514-43075514 on Build GRCh38
rs751020424 Chr.22: 43075508-43075508 on Build GRCh38
rs779103504 Chr.22: 43075561-43075561 on Build GRCh38
rs764895397 Chr.22: 43075483-43075483 on Build GRCh38
rs775896926 Chr.22: 43075467-43075467 on Build GRCh38
rs774259292 Chr.22: 43075455-43075455 on Build GRCh38
rs763994153 Chr.22: 43075574-43075574 on Build GRCh38
rs772884426 Chr.22: 43075493-43075493 on Build GRCh38
rs763444241 Chr.22: 43075556-43075557 on Build GRCh38
rs907867082 Chr.22: 43075505-43075505 on Build GRCh38
rs778081836 Chr.22: 43075447-43075447 on Build GRCh38
rs780077456 Chr.22: 43075593-43075593 on Build GRCh38
rs764703080 Chr.22: 43075549-43075549 on Build GRCh38
rs769052922 Chr.22: 43075567-43075567 on Build GRCh38
rs757519464 Chr.22: 43075570-43075571 on Build GRCh38
rs767497165 Chr.22: 43075530-43075530 on Build GRCh38

View Full Product Details