Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 43108992-43109263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RET
Entrez Gene ID
5979
Gene Name
ret proto-oncogene
Gene Aliases
CDHF12, CDHR16, HSCR1, MEN2A, MEN2B, MTC1, PTC, RET-ELE1, RET51
UniGene
Hs.350321
Gene Chromosome Location
Chr.10: 43077069-43130351 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4721078 c.1157C>T p.A386V Chr.10: 43109124-43109124 on Build GRCh38
6967139 c.1111C>A p.Q371K Chr.10: 43109078-43109078 on Build GRCh38
5836878 c.1226C>T p.S409F Chr.10: 43109193-43109193 on Build GRCh38
6981224 c.1100A>G p.N367S Chr.10: 43109067-43109067 on Build GRCh38
3397122 c.1082A>T p.N361I Chr.10: 43109049-43109049 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768547052 Chr.10: 43109215-43109215 on Build GRCh38
rs895556824 Chr.10: 43109132-43109132 on Build GRCh38
rs746970700 Chr.10: 43109201-43109201 on Build GRCh38
rs115272158 Chr.10: 43109124-43109124 on Build GRCh38
rs763967472 Chr.10: 43109128-43109128 on Build GRCh38
rs142338976 Chr.10: 43109091-43109091 on Build GRCh38
rs906695652 Chr.10: 43109121-43109121 on Build GRCh38
rs776223166 Chr.10: 43109129-43109129 on Build GRCh38
rs748588678 Chr.10: 43109182-43109182 on Build GRCh38
rs750569981 Chr.10: 43109083-43109083 on Build GRCh38
rs757868491 Chr.10: 43109140-43109140 on Build GRCh38
rs762472027 Chr.10: 43109046-43109046 on Build GRCh38
rs756532455 Chr.10: 43109183-43109183 on Build GRCh38
rs876660102 Chr.10: 43109033-43109033 on Build GRCh38
rs774139424 Chr.10: 43109115-43109115 on Build GRCh38
rs139813765 Chr.10: 43109109-43109109 on Build GRCh38
rs769548635 Chr.10: 43109225-43109225 on Build GRCh38
rs770637118 Chr.10: 43109113-43109113 on Build GRCh38
rs373540097 Chr.10: 43109125-43109125 on Build GRCh38
rs781362020 Chr.10: 43109108-43109108 on Build GRCh38
rs759548155 Chr.10: 43109059-43109059 on Build GRCh38
rs763559646 Chr.10: 43109102-43109103 on Build GRCh38
rs763670106 Chr.10: 43109061-43109061 on Build GRCh38
rs373327241 Chr.10: 43109015-43109015 on Build GRCh38
rs78098482 Chr.10: 43109146-43109146 on Build GRCh38
rs755400887 Chr.10: 43109106-43109106 on Build GRCh38
rs536298339 Chr.10: 43109117-43109117 on Build GRCh38
rs965224147 Chr.10: 43109186-43109186 on Build GRCh38
rs745479264 Chr.10: 43109191-43109191 on Build GRCh38
rs794728690 Chr.10: 43109040-43109040 on Build GRCh38
rs376465385 Chr.10: 43109149-43109149 on Build GRCh38
rs199529397 Chr.10: 43109070-43109070 on Build GRCh38
rs768878280 Chr.10: 43109025-43109025 on Build GRCh38
rs148371113 Chr.10: 43109164-43109164 on Build GRCh38
rs201030628 Chr.10: 43109217-43109217 on Build GRCh38
rs375815926 Chr.10: 43109017-43109017 on Build GRCh38
rs765305476 Chr.10: 43109077-43109077 on Build GRCh38
rs747466717 Chr.10: 43109021-43109021 on Build GRCh38
rs778754580 Chr.10: 43109189-43109189 on Build GRCh38
rs546346375 Chr.10: 43109013-43109013 on Build GRCh38
rs371731991 Chr.10: 43109220-43109220 on Build GRCh38
rs751939820 Chr.10: 43109130-43109130 on Build GRCh38
rs183729115 Chr.10: 43109156-43109156 on Build GRCh38
rs754116867 Chr.10: 43109069-43109069 on Build GRCh38
rs864622405 Chr.10: 43109190-43109190 on Build GRCh38
rs773450903 Chr.10: 43109242-43109242 on Build GRCh38
rs773935854 Chr.10: 43109051-43109051 on Build GRCh38
rs763759702 Chr.10: 43109067-43109067 on Build GRCh38
rs770587835 Chr.10: 43109050-43109050 on Build GRCh38
rs781623106 Chr.10: 43109170-43109170 on Build GRCh38
rs766881522 Chr.10: 43109055-43109055 on Build GRCh38
rs113931414 Chr.10: 43109086-43109086 on Build GRCh38
rs886046987 Chr.10: 43109076-43109076 on Build GRCh38
rs201992974 Chr.10: 43109062-43109062 on Build GRCh38
rs771679592 Chr.10: 43109118-43109118 on Build GRCh38
rs995081128 Chr.10: 43109197-43109197 on Build GRCh38
rs758510657 Chr.10: 43109155-43109155 on Build GRCh38
rs771636042 Chr.10: 43109193-43109193 on Build GRCh38
rs781646869 Chr.10: 43109154-43109154 on Build GRCh38
rs140638866 Chr.10: 43109168-43109168 on Build GRCh38
rs772956976 Chr.10: 43109026-43109026 on Build GRCh38
rs779374978 Chr.10: 43109200-43109200 on Build GRCh38
rs546866208 Chr.10: 43109085-43109085 on Build GRCh38
rs760560422 Chr.10: 43109064-43109064 on Build GRCh38

View Full Product Details