Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 1649269-1649494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIRPG
Entrez Gene ID
55423
Gene Name
signal regulatory protein gamma
Gene Aliases
CD172g, SIRP-B2, SIRPB2, SIRPgamma, bA77C3.1
UniGene
Hs.590883
Gene Chromosome Location
Chr.20: 1629152-1686516 on Build GRCh38
Transcripts
NM_001039508.1 NM_018556.3 NM_080816.2 XM_005260749.3 XM_011529286.2 XM_011529287.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3544309 c.161C>T p.T54I Chr.20: 1649321-1649321 on Build GRCh38
3544308 c.193C>T p.L65L Chr.20: 1649289-1649289 on Build GRCh38
3544307 c.212G>A p.G71E Chr.20: 1649270-1649270 on Build GRCh38
723580 c.136G>A p.G46R Chr.20: 1649346-1649346 on Build GRCh38
4405833 c.112G>A p.E38K Chr.20: 1649370-1649370 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs62641738 Chr.20: 1649292-1649292 on Build GRCh38
rs1056739709 Chr.20: 1649310-1649310 on Build GRCh38
rs748834939 Chr.20: 1649377-1649377 on Build GRCh38
rs138452343 Chr.20: 1649301-1649301 on Build GRCh38
rs760807673 Chr.20: 1649336-1649336 on Build GRCh38
rs759660817 Chr.20: 1649347-1649347 on Build GRCh38
rs770579012 Chr.20: 1649354-1649354 on Build GRCh38
rs150700648 Chr.20: 1649389-1649389 on Build GRCh38
rs753474042 Chr.20: 1649422-1649422 on Build GRCh38
rs772308141 Chr.20: 1649330-1649330 on Build GRCh38
rs771321985 Chr.20: 1649393-1649393 on Build GRCh38
rs749842164 Chr.20: 1649326-1649326 on Build GRCh38
rs757162144 Chr.20: 1649356-1649356 on Build GRCh38
rs143978848 Chr.20: 1649321-1649321 on Build GRCh38
rs776659540 Chr.20: 1649411-1649411 on Build GRCh38
rs774806997 Chr.20: 1649308-1649308 on Build GRCh38
rs78831862 Chr.20: 1649340-1649340 on Build GRCh38
rs778396958 Chr.20: 1649450-1649450 on Build GRCh38
rs903588441 Chr.20: 1649385-1649385 on Build GRCh38
rs776991507 Chr.20: 1649394-1649394 on Build GRCh38
rs892083448 Chr.20: 1649440-1649440 on Build GRCh38
rs368139177 Chr.20: 1649406-1649406 on Build GRCh38
rs372025725 Chr.20: 1649374-1649374 on Build GRCh38
rs779451925 Chr.20: 1649388-1649388 on Build GRCh38
rs202125965 Chr.20: 1649398-1649398 on Build GRCh38
rs751532170 Chr.20: 1649352-1649352 on Build GRCh38
rs201954358 Chr.20: 1649392-1649392 on Build GRCh38
rs375060745 Chr.20: 1649380-1649380 on Build GRCh38
rs998452787 Chr.20: 1649441-1649441 on Build GRCh38
rs576810342 Chr.20: 1649383-1649383 on Build GRCh38
rs750234705 Chr.20: 1649403-1649403 on Build GRCh38
rs775245375 Chr.20: 1649348-1649348 on Build GRCh38
rs6074949 Chr.20: 1649463-1649463 on Build GRCh38
rs199911378 Chr.20: 1649302-1649302 on Build GRCh38
rs202224518 Chr.20: 1649375-1649375 on Build GRCh38
rs772559713 Chr.20: 1649390-1649390 on Build GRCh38
rs767440312 Chr.20: 1649401-1649401 on Build GRCh38
rs750193498 Chr.20: 1649371-1649371 on Build GRCh38
rs773564663 Chr.20: 1649333-1649333 on Build GRCh38
rs776682101 Chr.20: 1649396-1649398 on Build GRCh38
rs780854234 Chr.20: 1649364-1649364 on Build GRCh38
rs752758859 Chr.20: 1649399-1649399 on Build GRCh38
rs759749841 Chr.20: 1649396-1649396 on Build GRCh38
rs1030660946 Chr.20: 1649432-1649432 on Build GRCh38
rs201443740 Chr.20: 1649448-1649448 on Build GRCh38
rs960986801 Chr.20: 1649382-1649382 on Build GRCh38
rs754024064 Chr.20: 1649341-1649341 on Build GRCh38
rs139446471 Chr.20: 1649338-1649339 on Build GRCh38
rs200917950 Chr.20: 1649295-1649295 on Build GRCh38
rs1005532892 Chr.20: 1649361-1649361 on Build GRCh38
rs769421642 Chr.20: 1649293-1649293 on Build GRCh38
rs761688818 Chr.20: 1649400-1649400 on Build GRCh38

View Full Product Details