Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 17154574-17154847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRDMT1
Entrez Gene ID
1787
Gene Name
tRNA aspartic acid methyltransferase 1
Gene Aliases
DMNT2, DNMT2, MHSAIIP, PUMET, RNMT1
UniGene
Hs.351665
Gene Chromosome Location
Chr.10: 17136785-17202071 on Build GRCh38
Transcripts
NM_001321006.1 NM_001321007.1 NM_004412.6 XM_005252372.4 XM_005252374.4 XM_005252375.4 XM_005252378.3 XM_006717388.3 XM_011519343.2 XM_011519344.2 XM_011519345.2 XM_017015800.1 XM_017015801.1 XM_017015802.1 XM_017015803.1 XM_017015804.1 XM_017015805.1 XM_017015806.1 XM_017015807.1 XM_017015808.1 XM_017015809.1 XM_017015810.1 XM_017015811.1 XM_017015812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
916826 c.941T>C p.V314A Chr.10: 17154681-17154681 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs3736820 Chr.10: 17154671-17154671 on Build GRCh38
rs141892183 Chr.10: 17154777-17154777 on Build GRCh38
rs753337305 Chr.10: 17154734-17154734 on Build GRCh38
rs751420998 Chr.10: 17154687-17154687 on Build GRCh38
rs1016063451 Chr.10: 17154609-17154609 on Build GRCh38
rs566295441 Chr.10: 17154651-17154651 on Build GRCh38
rs758998465 Chr.10: 17154744-17154744 on Build GRCh38
rs779472336 Chr.10: 17154713-17154713 on Build GRCh38
rs368057422 Chr.10: 17154617-17154617 on Build GRCh38
rs1022819093 Chr.10: 17154736-17154736 on Build GRCh38
rs969987341 Chr.10: 17154806-17154806 on Build GRCh38
rs376914274 Chr.10: 17154766-17154766 on Build GRCh38
rs778117494 Chr.10: 17154765-17154765 on Build GRCh38
rs547139638 Chr.10: 17154635-17154635 on Build GRCh38
rs755634106 Chr.10: 17154708-17154708 on Build GRCh38
rs373596044 Chr.10: 17154731-17154731 on Build GRCh38
rs775858438 Chr.10: 17154678-17154679 on Build GRCh38
rs763925703 Chr.10: 17154683-17154683 on Build GRCh38
rs761484482 Chr.10: 17154695-17154695 on Build GRCh38
rs775577463 Chr.10: 17154677-17154677 on Build GRCh38
rs1030368992 Chr.10: 17154665-17154665 on Build GRCh38
rs762526907 Chr.10: 17154700-17154701 on Build GRCh38
rs182399517 Chr.10: 17154608-17154608 on Build GRCh38
rs374919278 Chr.10: 17154772-17154772 on Build GRCh38
rs963545979 Chr.10: 17154614-17154614 on Build GRCh38
rs771231409 Chr.10: 17154650-17154650 on Build GRCh38
rs759714036 Chr.10: 17154669-17154669 on Build GRCh38
rs747438698 Chr.10: 17154771-17154771 on Build GRCh38
rs766560520 Chr.10: 17154809-17154809 on Build GRCh38
rs112438197 Chr.10: 17154745-17154745 on Build GRCh38
rs750028293 Chr.10: 17154706-17154706 on Build GRCh38
rs572651585 Chr.10: 17154758-17154758 on Build GRCh38
rs781581334 Chr.10: 17154776-17154776 on Build GRCh38
rs763014908 Chr.10: 17154682-17154682 on Build GRCh38
rs528993266 Chr.10: 17154628-17154628 on Build GRCh38
rs149889117 Chr.10: 17154701-17154701 on Build GRCh38
rs770542622 Chr.10: 17154815-17154818 on Build GRCh38

View Full Product Details