Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42180209-42180474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLAT
Entrez Gene ID
5327
Gene Name
plasminogen activator, tissue type
Gene Aliases
T-PA, TPA
UniGene
Hs.491582
Gene Chromosome Location
Chr.8: 42174718-42207724 on Build GRCh38
Transcripts
NM_000930.4 NM_001319189.1 NM_033011.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1099982 c.1142G>A p.G381D Chr.8: 42180322-42180322 on Build GRCh38
6747850 c.1206T>C p.T402T Chr.8: 42180258-42180258 on Build GRCh38
1099981 c.1209C>T p.Y403Y Chr.8: 42180255-42180255 on Build GRCh38
6747847 c.1161T>C p.F387F Chr.8: 42180303-42180303 on Build GRCh38
4387326 c.1196A>G p.D399G Chr.8: 42180268-42180268 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746726957 Chr.8: 42180391-42180391 on Build GRCh38
rs116381625 Chr.8: 42180346-42180346 on Build GRCh38
rs769232665 Chr.8: 42180419-42180419 on Build GRCh38
rs762297581 Chr.8: 42180262-42180262 on Build GRCh38
rs199960677 Chr.8: 42180291-42180291 on Build GRCh38
rs752793328 Chr.8: 42180358-42180358 on Build GRCh38
rs747683509 Chr.8: 42180416-42180416 on Build GRCh38
rs200758880 Chr.8: 42180357-42180357 on Build GRCh38
rs374730133 Chr.8: 42180372-42180372 on Build GRCh38
rs570954622 Chr.8: 42180315-42180315 on Build GRCh38
rs777263586 Chr.8: 42180256-42180256 on Build GRCh38
rs753868871 Chr.8: 42180293-42180293 on Build GRCh38
rs375277780 Chr.8: 42180453-42180453 on Build GRCh38
rs749787941 Chr.8: 42180426-42180426 on Build GRCh38
rs8178782 Chr.8: 42180285-42180285 on Build GRCh38
rs765280155 Chr.8: 42180369-42180369 on Build GRCh38
rs759662055 Chr.8: 42180354-42180354 on Build GRCh38
rs774554600 Chr.8: 42180445-42180445 on Build GRCh38
rs765641172 Chr.8: 42180264-42180264 on Build GRCh38
rs528828208 Chr.8: 42180243-42180243 on Build GRCh38
rs548902992 Chr.8: 42180280-42180280 on Build GRCh38
rs779703103 Chr.8: 42180387-42180387 on Build GRCh38
rs8178781 Chr.8: 42180297-42180297 on Build GRCh38
rs780860854 Chr.8: 42180411-42180411 on Build GRCh38
rs370878686 Chr.8: 42180234-42180234 on Build GRCh38
rs756223584 Chr.8: 42180360-42180360 on Build GRCh38
rs144211716 Chr.8: 42180317-42180317 on Build GRCh38
rs149824937 Chr.8: 42180250-42180250 on Build GRCh38
rs759543775 Chr.8: 42180262-42180264 on Build GRCh38
rs755088165 Chr.8: 42180287-42180287 on Build GRCh38
rs767986898 Chr.8: 42180255-42180255 on Build GRCh38
rs775489173 Chr.8: 42180428-42180428 on Build GRCh38
rs757237150 Chr.8: 42180294-42180294 on Build GRCh38
rs775983939 Chr.8: 42180321-42180321 on Build GRCh38
rs372549798 Chr.8: 42180425-42180425 on Build GRCh38
rs148523981 Chr.8: 42180314-42180314 on Build GRCh38
rs968701455 Chr.8: 42180304-42180304 on Build GRCh38
rs1036121759 Chr.8: 42180405-42180405 on Build GRCh38
rs757393551 Chr.8: 42180249-42180249 on Build GRCh38
rs771818476 Chr.8: 42180244-42180244 on Build GRCh38
rs1000900649 Chr.8: 42180237-42180237 on Build GRCh38
rs373293390 Chr.8: 42180239-42180239 on Build GRCh38
rs758336862 Chr.8: 42180373-42180373 on Build GRCh38
rs8178780 Chr.8: 42180454-42180454 on Build GRCh38
rs745564392 Chr.8: 42180235-42180235 on Build GRCh38
rs932225923 Chr.8: 42180335-42180335 on Build GRCh38
rs773538670 Chr.8: 42180276-42180276 on Build GRCh38
rs763316163 Chr.8: 42180344-42180344 on Build GRCh38
rs2020922 Chr.8: 42180406-42180406 on Build GRCh38
rs765296951 Chr.8: 42180404-42180405 on Build GRCh38
rs770320667 Chr.8: 42180326-42180326 on Build GRCh38
rs146538335 Chr.8: 42180333-42180333 on Build GRCh38
rs771949840 Chr.8: 42180239-42180240 on Build GRCh38
rs201653314 Chr.8: 42180444-42180444 on Build GRCh38
rs547017710 Chr.8: 42180371-42180371 on Build GRCh38
rs748697637 Chr.8: 42180325-42180325 on Build GRCh38
rs2979892 Chr.8: 42180312-42180312 on Build GRCh38
rs368524202 Chr.8: 42180399-42180399 on Build GRCh38
rs751671151 Chr.8: 42180286-42180286 on Build GRCh38
rs773145601 Chr.8: 42180249-42180251 on Build GRCh38

View Full Product Details