Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 21146666-21146927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAP18
Entrez Gene ID
10284
Gene Name
Sin3A associated protein 18
Gene Aliases
2HOR0202, SAP18P
UniGene
Hs.524899
Gene Chromosome Location
Chr.13: 21140514-21149085 on Build GRCh38
Transcripts
NM_005870.4 XM_011534868.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6402586 c.191C>T p.A64V Chr.13: 21146813-21146813 on Build GRCh38
6771850 c.202G>T p.E68* Chr.13: 21146824-21146824 on Build GRCh38
1677518 c.304C>T p.R102* Chr.13: 21146926-21146926 on Build GRCh38
6838446 c.209C>G p.T70R Chr.13: 21146831-21146831 on Build GRCh38
1493124 c.305G>A p.R102Q Chr.13: 21146927-21146927 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1036746560 Chr.13: 21146730-21146730 on Build GRCh38
rs770685260 Chr.13: 21146885-21146885 on Build GRCh38
rs748376383 Chr.13: 21146876-21146876 on Build GRCh38
rs759393970 Chr.13: 21146893-21146893 on Build GRCh38
rs749330807 Chr.13: 21146848-21146848 on Build GRCh38
rs188317473 Chr.13: 21146695-21146695 on Build GRCh38
rs776479725 Chr.13: 21146888-21146888 on Build GRCh38
rs534303168 Chr.13: 21146865-21146865 on Build GRCh38
rs756624393 Chr.13: 21146792-21146792 on Build GRCh38
rs995314330 Chr.13: 21146737-21146737 on Build GRCh38
rs764669421 Chr.13: 21146829-21146829 on Build GRCh38
rs968775589 Chr.13: 21146847-21146847 on Build GRCh38
rs779316684 Chr.13: 21146868-21146868 on Build GRCh38
rs956068163 Chr.13: 21146811-21146811 on Build GRCh38
rs4620 Chr.13: 21146817-21146817 on Build GRCh38
rs150187315 Chr.13: 21146790-21146790 on Build GRCh38
rs530633079 Chr.13: 21146766-21146766 on Build GRCh38
rs763701092 Chr.13: 21146777-21146777 on Build GRCh38
rs7981250 Chr.13: 21146791-21146791 on Build GRCh38
rs898229355 Chr.13: 21146734-21146734 on Build GRCh38
rs912222899 Chr.13: 21146719-21146719 on Build GRCh38
rs11147974 Chr.13: 21146729-21146729 on Build GRCh38
rs1021730701 Chr.13: 21146846-21146846 on Build GRCh38
rs370343426 Chr.13: 21146783-21146783 on Build GRCh38
rs1051242844 Chr.13: 21146699-21146699 on Build GRCh38
rs751196296 Chr.13: 21146779-21146779 on Build GRCh38
rs781253125 Chr.13: 21146791-21146792 on Build GRCh38
rs954845353 Chr.13: 21146840-21146840 on Build GRCh38

View Full Product Details