Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 116698969-116699236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASTN2
Entrez Gene ID
23245
Gene Name
astrotactin 2
Gene Aliases
bA67K19.1
UniGene
Hs.601562
Gene Chromosome Location
Chr.9: 116425225-117415039 on Build GRCh38
Transcripts
NM_014010.4 NM_001099679.1 NM_012210.3 XM_005251813.3 XM_011518398.2 XM_017014486.1

Gene Symbol
TRIM32
Entrez Gene ID
22954
Gene Name
tripartite motif containing 32
Gene Aliases
BBS11, HT2A, LGMD2H, TATIP
UniGene
Hs.591910
Gene Chromosome Location
Chr.9: 116687302-116701300 on Build GRCh38
Transcripts
NM_014010.4 NM_001099679.1 NM_012210.3 XM_005251813.3 XM_011518398.2 XM_017014486.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3903533 c.1285C>A p.L429I Chr.9: 116699027-116699027 on Build GRCh38
3903532 c.1271C>T p.A424V Chr.9: 116699013-116699013 on Build GRCh38
3653560 c.1292C>T p.P431L Chr.9: 116699034-116699034 on Build GRCh38
4588403 c.1354C>T p.L452F Chr.9: 116699096-116699096 on Build GRCh38
5984605 c.1298C>A p.S433* Chr.9: 116699040-116699040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755075876 Chr.9: 116699197-116699197 on Build GRCh38
rs1661300 Chr.9: 116698996-116698996 on Build GRCh38
rs767176438 Chr.9: 116699018-116699018 on Build GRCh38
rs987882732 Chr.9: 116699063-116699063 on Build GRCh38
rs757634856 Chr.9: 116699027-116699027 on Build GRCh38
rs759016258 Chr.9: 116699144-116699144 on Build GRCh38
rs772922696 Chr.9: 116699126-116699126 on Build GRCh38
rs770803017 Chr.9: 116699140-116699140 on Build GRCh38
rs886043897 Chr.9: 116699129-116699129 on Build GRCh38
rs779090548 Chr.9: 116699029-116699029 on Build GRCh38
rs747954415 Chr.9: 116699116-116699116 on Build GRCh38
rs374136376 Chr.9: 116699158-116699158 on Build GRCh38
rs371002754 Chr.9: 116699174-116699174 on Build GRCh38
rs922859943 Chr.9: 116699203-116699203 on Build GRCh38
rs141965401 Chr.9: 116699200-116699200 on Build GRCh38
rs151116227 Chr.9: 116699142-116699142 on Build GRCh38
rs751757854 Chr.9: 116699189-116699189 on Build GRCh38
rs780303807 Chr.9: 116699074-116699074 on Build GRCh38
rs1025223714 Chr.9: 116699041-116699041 on Build GRCh38
rs760180890 Chr.9: 116699171-116699171 on Build GRCh38
rs752379777 Chr.9: 116699169-116699169 on Build GRCh38
rs886044913 Chr.9: 116699108-116699109 on Build GRCh38
rs765666809 Chr.9: 116699172-116699172 on Build GRCh38
rs150813870 Chr.9: 116699017-116699017 on Build GRCh38
rs142997889 Chr.9: 116699036-116699036 on Build GRCh38
rs781475334 Chr.9: 116699103-116699103 on Build GRCh38
rs762905941 Chr.9: 116699128-116699128 on Build GRCh38
rs111033570 Chr.9: 116699201-116699201 on Build GRCh38
rs769863991 Chr.9: 116699210-116699210 on Build GRCh38
rs747067557 Chr.9: 116699096-116699096 on Build GRCh38
rs777563978 Chr.9: 116699211-116699211 on Build GRCh38
rs746383181 Chr.9: 116699048-116699048 on Build GRCh38
rs748404804 Chr.9: 116699204-116699204 on Build GRCh38
rs758934164 Chr.9: 116699067-116699067 on Build GRCh38
rs765874460 Chr.9: 116698990-116698990 on Build GRCh38
rs754192979 Chr.9: 116699020-116699020 on Build GRCh38
rs35904234 Chr.9: 116699110-116699111 on Build GRCh38
rs780679793 Chr.9: 116699184-116699184 on Build GRCh38
rs759024082 Chr.9: 116699178-116699178 on Build GRCh38
rs200244154 Chr.9: 116699125-116699125 on Build GRCh38

View Full Product Details