Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169703132-169703367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SELL
Entrez Gene ID
6402
Gene Name
selectin L
Gene Aliases
CD62L, LAM1, LECAM1, LEU8, LNHR, LSEL, LYAM1, PLNHR, TQ1
UniGene
Hs.728756
Gene Chromosome Location
Chr.1: 169690665-169711702 on Build GRCh38
Transcripts
NM_000655.4 NR_029467.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3478079 c.980C>T p.P327L Chr.1: 169703266-169703266 on Build GRCh38
3478080 c.941C>T p.P314L Chr.1: 169703266-169703266 on Build GRCh38
3478082 c.920G>A p.G307E Chr.1: 169703287-169703287 on Build GRCh38
4868964 c.952T>G p.S318A Chr.1: 169703294-169703294 on Build GRCh38
263568 c.901A>G p.T301A Chr.1: 169703306-169703306 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774470940 Chr.1: 169703316-169703316 on Build GRCh38
rs746496297 Chr.1: 169703265-169703265 on Build GRCh38
rs4987314 Chr.1: 169703202-169703202 on Build GRCh38
rs377239098 Chr.1: 169703208-169703208 on Build GRCh38
rs555880863 Chr.1: 169703166-169703166 on Build GRCh38
rs750706314 Chr.1: 169703244-169703244 on Build GRCh38
rs754221297 Chr.1: 169703333-169703333 on Build GRCh38
rs762959718 Chr.1: 169703214-169703214 on Build GRCh38
rs756386504 Chr.1: 169703246-169703246 on Build GRCh38
rs920205193 Chr.1: 169703202-169703214 on Build GRCh38
rs931697163 Chr.1: 169703178-169703178 on Build GRCh38
rs561566828 Chr.1: 169703272-169703272 on Build GRCh38
rs766581126 Chr.1: 169703325-169703325 on Build GRCh38
rs369319963 Chr.1: 169703212-169703212 on Build GRCh38
rs745540146 Chr.1: 169703285-169703285 on Build GRCh38
rs1050204512 Chr.1: 169703179-169703179 on Build GRCh38
rs754334066 Chr.1: 169703265-169703268 on Build GRCh38
rs768624371 Chr.1: 169703314-169703314 on Build GRCh38
rs577651651 Chr.1: 169703199-169703199 on Build GRCh38
rs866691931 Chr.1: 169703293-169703293 on Build GRCh38
rs767729798 Chr.1: 169703320-169703320 on Build GRCh38
rs764013472 Chr.1: 169703218-169703218 on Build GRCh38
rs767817755 Chr.1: 169703237-169703237 on Build GRCh38
rs188911090 Chr.1: 169703322-169703322 on Build GRCh38
rs761945033 Chr.1: 169703318-169703318 on Build GRCh38
rs775357924 Chr.1: 169703288-169703288 on Build GRCh38
rs574888692 Chr.1: 169703286-169703286 on Build GRCh38
rs762722611 Chr.1: 169703295-169703295 on Build GRCh38
rs770464100 Chr.1: 169703267-169703267 on Build GRCh38
rs779458129 Chr.1: 169703264-169703264 on Build GRCh38
rs750618231 Chr.1: 169703321-169703321 on Build GRCh38
rs749694665 Chr.1: 169703260-169703260 on Build GRCh38
rs147585835 Chr.1: 169703252-169703252 on Build GRCh38
rs755508446 Chr.1: 169703261-169703261 on Build GRCh38
rs184513878 Chr.1: 169703220-169703220 on Build GRCh38
rs4987313 Chr.1: 169703219-169703219 on Build GRCh38

View Full Product Details