Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 60511039-60511300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIX6
Entrez Gene ID
4990
Gene Name
SIX homeobox 6
Gene Aliases
MCOPCT2, ODRMD, OPTX2, Six9
UniGene
Hs.194756
Gene Chromosome Location
Chr.14: 60509220-60511807 on Build GRCh38
Transcripts
NM_007374.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
470091 c.583C>G p.Q195E Chr.14: 60511094-60511094 on Build GRCh38
3815023 c.596A>C p.Q199P Chr.14: 60511107-60511107 on Build GRCh38
2246899 c.637C>T p.P213S Chr.14: 60511148-60511148 on Build GRCh38
1225806 c.598G>T p.G200C Chr.14: 60511109-60511109 on Build GRCh38
5382412 c.625G>A p.G209S Chr.14: 60511136-60511136 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368912930 Chr.14: 60511162-60511162 on Build GRCh38
rs752534682 Chr.14: 60511128-60511128 on Build GRCh38
rs766380615 Chr.14: 60511159-60511159 on Build GRCh38
rs752483519 Chr.14: 60511176-60511176 on Build GRCh38
rs114981075 Chr.14: 60511248-60511248 on Build GRCh38
rs745496406 Chr.14: 60511090-60511090 on Build GRCh38
rs746623345 Chr.14: 60511236-60511236 on Build GRCh38
rs761411577 Chr.14: 60511074-60511074 on Build GRCh38
rs770148963 Chr.14: 60511237-60511237 on Build GRCh38
rs897409621 Chr.14: 60511136-60511136 on Build GRCh38
rs916365037 Chr.14: 60511074-60511074 on Build GRCh38
rs765698334 Chr.14: 60511111-60511111 on Build GRCh38
rs375282881 Chr.14: 60511076-60511076 on Build GRCh38
rs758032582 Chr.14: 60511216-60511216 on Build GRCh38
rs886050563 Chr.14: 60511118-60511118 on Build GRCh38
rs45549246 Chr.14: 60511125-60511125 on Build GRCh38
rs750136469 Chr.14: 60511201-60511201 on Build GRCh38
rs530622461 Chr.14: 60511169-60511169 on Build GRCh38
rs201991012 Chr.14: 60511119-60511119 on Build GRCh38
rs766404727 Chr.14: 60511121-60511121 on Build GRCh38
rs778422614 Chr.14: 60511138-60511138 on Build GRCh38
rs762371105 Chr.14: 60511103-60511103 on Build GRCh38
rs540804738 Chr.14: 60511130-60511130 on Build GRCh38
rs201560655 Chr.14: 60511148-60511148 on Build GRCh38
rs991944828 Chr.14: 60511058-60511058 on Build GRCh38
rs200339406 Chr.14: 60511182-60511182 on Build GRCh38
rs372216093 Chr.14: 60511178-60511178 on Build GRCh38
rs767750775 Chr.14: 60511167-60511167 on Build GRCh38
rs140601427 Chr.14: 60511147-60511147 on Build GRCh38
rs1037308746 Chr.14: 60511135-60511135 on Build GRCh38
rs140916124 Chr.14: 60511132-60511132 on Build GRCh38
rs999129819 Chr.14: 60511170-60511170 on Build GRCh38
rs763918148 Chr.14: 60511242-60511242 on Build GRCh38
rs202029915 Chr.14: 60511146-60511146 on Build GRCh38
rs146372482 Chr.14: 60511133-60511133 on Build GRCh38
rs777038713 Chr.14: 60511093-60511093 on Build GRCh38
rs759639034 Chr.14: 60511265-60511265 on Build GRCh38
rs148118869 Chr.14: 60511127-60511127 on Build GRCh38
rs745524344 Chr.14: 60511142-60511142 on Build GRCh38
rs34787514 Chr.14: 60511165-60511165 on Build GRCh38
rs1011681192 Chr.14: 60511217-60511217 on Build GRCh38
rs773738794 Chr.14: 60511151-60511151 on Build GRCh38
rs745349858 Chr.14: 60511209-60511209 on Build GRCh38
rs771466473 Chr.14: 60511252-60511252 on Build GRCh38
rs757206532 Chr.14: 60511194-60511194 on Build GRCh38
rs763383468 Chr.14: 60511154-60511154 on Build GRCh38
rs778051865 Chr.14: 60511240-60511240 on Build GRCh38
rs916435051 Chr.14: 60511269-60511269 on Build GRCh38
rs533016019 Chr.14: 60511266-60511266 on Build GRCh38
rs764761660 Chr.14: 60511164-60511165 on Build GRCh38
rs770759922 Chr.14: 60511078-60511078 on Build GRCh38
rs756162271 Chr.14: 60511129-60511129 on Build GRCh38
rs759745079 Chr.14: 60511163-60511163 on Build GRCh38
rs773325780 Chr.14: 60511114-60511114 on Build GRCh38
rs774336797 Chr.14: 60511262-60511262 on Build GRCh38
rs775890009 Chr.14: 60511273-60511273 on Build GRCh38
rs958745864 Chr.14: 60511202-60511202 on Build GRCh38
rs778350770 Chr.14: 60511206-60511206 on Build GRCh38
rs779776142 Chr.14: 60511225-60511225 on Build GRCh38
rs768908961 Chr.14: 60511091-60511091 on Build GRCh38

View Full Product Details