Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57414530-57414763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC43A3
Entrez Gene ID
29015
Gene Name
solute carrier family 43 member 3
Gene Aliases
EEG1, FOAP-13, PRO1659, SEEEG-1
UniGene
Hs.99962
Gene Chromosome Location
Chr.11: 57406954-57427580 on Build GRCh38
Transcripts
NM_001278201.1 NM_001278206.1 NM_014096.3 NM_017611.2 NM_199329.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3449702 c.1007G>A p.G336D Chr.11: 57414668-57414668 on Build GRCh38
3415992 c.981C>T p.F327F Chr.11: 57414694-57414694 on Build GRCh38
5892153 c.1060+2T>G p.? Chr.11: 57414613-57414613 on Build GRCh38
4034057 c.1038C>G p.Y346* Chr.11: 57414637-57414637 on Build GRCh38
4801486 c.981C>A p.F327L Chr.11: 57414694-57414694 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747316550 Chr.11: 57414728-57414728 on Build GRCh38
rs1027657665 Chr.11: 57414655-57414655 on Build GRCh38
rs373710995 Chr.11: 57414591-57414591 on Build GRCh38
rs780811572 Chr.11: 57414587-57414587 on Build GRCh38
rs780529332 Chr.11: 57414725-57414725 on Build GRCh38
rs752588393 Chr.11: 57414693-57414693 on Build GRCh38
rs147287918 Chr.11: 57414654-57414654 on Build GRCh38
rs773590399 Chr.11: 57414632-57414632 on Build GRCh38
rs754865833 Chr.11: 57414566-57414566 on Build GRCh38
rs144879611 Chr.11: 57414599-57414599 on Build GRCh38
rs149064116 Chr.11: 57414605-57414605 on Build GRCh38
rs141024414 Chr.11: 57414694-57414694 on Build GRCh38
rs186807194 Chr.11: 57414645-57414645 on Build GRCh38
rs996890484 Chr.11: 57414667-57414667 on Build GRCh38
rs761955034 Chr.11: 57414629-57414629 on Build GRCh38
rs117212150 Chr.11: 57414602-57414602 on Build GRCh38
rs542073476 Chr.11: 57414698-57414698 on Build GRCh38
rs564824714 Chr.11: 57414572-57414579 on Build GRCh38
rs755551154 Chr.11: 57414735-57414735 on Build GRCh38
rs769955380 Chr.11: 57414631-57414631 on Build GRCh38
rs776823539 Chr.11: 57414623-57414623 on Build GRCh38
rs541432684 Chr.11: 57414653-57414653 on Build GRCh38
rs367910535 Chr.11: 57414606-57414606 on Build GRCh38
rs755801756 Chr.11: 57414595-57414595 on Build GRCh38
rs748887770 Chr.11: 57414601-57414601 on Build GRCh38
rs767324170 Chr.11: 57414666-57414666 on Build GRCh38
rs756859839 Chr.11: 57414703-57414703 on Build GRCh38
rs759565226 Chr.11: 57414661-57414661 on Build GRCh38

View Full Product Details