Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30155604-30155875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM10
Entrez Gene ID
10107
Gene Name
tripartite motif containing 10
Gene Aliases
HERF1, RFB30, RNF9
UniGene
Hs.709483
Gene Chromosome Location
Chr.6: 30151939-30163216 on Build GRCh38
Transcripts
NM_006778.3 NM_052828.2 XM_011514221.1 XM_011514222.2 XM_011514223.2 XM_011514225.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5870014 c.925C>T p.P309S Chr.6: 30155730-30155730 on Build GRCh38
5870016 c.925C>T p.P309S Chr.6: 30155730-30155730 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746406417 Chr.6: 30155763-30155763 on Build GRCh38
rs778193853 Chr.6: 30155744-30155744 on Build GRCh38
rs766354550 Chr.6: 30155687-30155687 on Build GRCh38
rs187250043 Chr.6: 30155684-30155684 on Build GRCh38
rs372890568 Chr.6: 30155685-30155685 on Build GRCh38
rs753843291 Chr.6: 30155714-30155714 on Build GRCh38
rs142556750 Chr.6: 30155821-30155821 on Build GRCh38
rs754494139 Chr.6: 30155719-30155719 on Build GRCh38
rs763124614 Chr.6: 30155677-30155677 on Build GRCh38
rs752369522 Chr.6: 30155737-30155737 on Build GRCh38
rs901065449 Chr.6: 30155712-30155712 on Build GRCh38
rs377207636 Chr.6: 30155731-30155731 on Build GRCh38
rs942533956 Chr.6: 30155645-30155645 on Build GRCh38
rs1046678845 Chr.6: 30155842-30155842 on Build GRCh38
rs758041021 Chr.6: 30155743-30155743 on Build GRCh38
rs757631272 Chr.6: 30155757-30155757 on Build GRCh38
rs373475134 Chr.6: 30155749-30155749 on Build GRCh38
rs369607797 Chr.6: 30155786-30155786 on Build GRCh38
rs750245014 Chr.6: 30155679-30155679 on Build GRCh38
rs767323816 Chr.6: 30155678-30155678 on Build GRCh38
rs905478817 Chr.6: 30155761-30155761 on Build GRCh38
rs377295787 Chr.6: 30155758-30155758 on Build GRCh38
rs781626852 Chr.6: 30155762-30155762 on Build GRCh38
rs1032557569 Chr.6: 30155800-30155800 on Build GRCh38
rs1046432363 Chr.6: 30155754-30155754 on Build GRCh38

View Full Product Details