Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 56003811-56004066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLRP5
Entrez Gene ID
126206
Gene Name
NLR family pyrin domain containing 5
Gene Aliases
CLR19.8, MATER, NALP5, PAN11, PYPAF8
UniGene
Hs.356872
Gene Chromosome Location
Chr.19: 55999726-56061810 on Build GRCh38
Transcripts
NM_153447.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
298740 c.406C>T p.R136* Chr.19: 56004059-56004059 on Build GRCh38
4000983 c.315C>T p.N105N Chr.19: 56003968-56003968 on Build GRCh38
4612579 c.245_259del15 p.L83_K87delLLKKK Chr.19: 56003898-56003912 on Build GRCh38
5914151 c.353G>A p.G118E Chr.19: 56004006-56004006 on Build GRCh38
4914582 c.202T>C p.C68R Chr.19: 56003855-56003855 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779572985 Chr.19: 56003981-56003981 on Build GRCh38
rs866571014 Chr.19: 56004027-56004027 on Build GRCh38
rs143801460 Chr.19: 56003862-56003862 on Build GRCh38
rs774526318 Chr.19: 56003925-56003925 on Build GRCh38
rs747175758 Chr.19: 56004004-56004004 on Build GRCh38
rs374211594 Chr.19: 56003975-56003975 on Build GRCh38
rs557226024 Chr.19: 56003866-56003866 on Build GRCh38
rs774551879 Chr.19: 56003873-56003876 on Build GRCh38
rs1032626691 Chr.19: 56003937-56003937 on Build GRCh38
rs773551212 Chr.19: 56003933-56003933 on Build GRCh38
rs749731517 Chr.19: 56003914-56003914 on Build GRCh38
rs749868596 Chr.19: 56003841-56003841 on Build GRCh38
rs756864732 Chr.19: 56003892-56003892 on Build GRCh38
rs973422972 Chr.19: 56004038-56004038 on Build GRCh38
rs765134419 Chr.19: 56003867-56003867 on Build GRCh38
rs761326403 Chr.19: 56003964-56003964 on Build GRCh38
rs769257961 Chr.19: 56003920-56003920 on Build GRCh38
rs373401163 Chr.19: 56003957-56003957 on Build GRCh38
rs754254004 Chr.19: 56003972-56003972 on Build GRCh38
rs545438956 Chr.19: 56004013-56004013 on Build GRCh38
rs199871361 Chr.19: 56003953-56003953 on Build GRCh38
rs752571450 Chr.19: 56003874-56003874 on Build GRCh38
rs774616335 Chr.19: 56003843-56003843 on Build GRCh38
rs772360032 Chr.19: 56003927-56003927 on Build GRCh38
rs768523862 Chr.19: 56004036-56004036 on Build GRCh38
rs756421184 Chr.19: 56003902-56003902 on Build GRCh38
rs749451274 Chr.19: 56003882-56003882 on Build GRCh38
rs771206697 Chr.19: 56004005-56004005 on Build GRCh38
rs1560691 Chr.19: 56003968-56003968 on Build GRCh38
rs370612026 Chr.19: 56004028-56004028 on Build GRCh38
rs35669530 Chr.19: 56003965-56003965 on Build GRCh38
rs746173695 Chr.19: 56003899-56003899 on Build GRCh38
rs761806403 Chr.19: 56004045-56004045 on Build GRCh38
rs536228838 Chr.19: 56003956-56003956 on Build GRCh38
rs780263166 Chr.19: 56003969-56003969 on Build GRCh38
rs199818039 Chr.19: 56003993-56003993 on Build GRCh38
rs751089414 Chr.19: 56003887-56003887 on Build GRCh38
rs775875299 Chr.19: 56003838-56003838 on Build GRCh38
rs767881593 Chr.19: 56003851-56003851 on Build GRCh38
rs955978842 Chr.19: 56003939-56003939 on Build GRCh38
rs778039113 Chr.19: 56004003-56004003 on Build GRCh38
rs761024250 Chr.19: 56003946-56003946 on Build GRCh38
rs758399773 Chr.19: 56003879-56003879 on Build GRCh38
rs770270582 Chr.19: 56003836-56003836 on Build GRCh38
rs780318657 Chr.19: 56004039-56004039 on Build GRCh38
rs780828462 Chr.19: 56003897-56003897 on Build GRCh38
rs543185262 Chr.19: 56003842-56003842 on Build GRCh38
rs775402782 Chr.19: 56003955-56003955 on Build GRCh38
rs748240899 Chr.19: 56003926-56003926 on Build GRCh38

View Full Product Details