Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 38403567-38403796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTC
Entrez Gene ID
5009
Gene Name
ornithine carbamoyltransferase
Gene Aliases
OCTD
UniGene
Hs.117050
Gene Chromosome Location
Chr.X: 38352483-38421450 on Build GRCh38
Transcripts
NM_000531.5 XM_017029556.1

SNP Detail

SNP ID Chromosome Location
rs72558423 Chr.X: 38403723-38403723 on Build GRCh38
rs72556293 Chr.X: 38403625-38403625 on Build GRCh38
rs72558404 Chr.X: 38403671-38403671 on Build GRCh38
rs72558408 Chr.X: 38403681-38403681 on Build GRCh38
rs72558415 Chr.X: 38403698-38403698 on Build GRCh38
rs72558418 Chr.X: 38403705-38403705 on Build GRCh38
rs68170503 Chr.X: 38403741-38403741 on Build GRCh38
rs67294955 Chr.X: 38403660-38403660 on Build GRCh38
rs72556302 Chr.X: 38403667-38403667 on Build GRCh38
rs1800327 Chr.X: 38403662-38403662 on Build GRCh38
rs72556289 Chr.X: 38403616-38403616 on Build GRCh38
rs67284661 Chr.X: 38403654-38403654 on Build GRCh38
rs72558419 Chr.X: 38403714-38403714 on Build GRCh38
rs72558424 Chr.X: 38403727-38403727 on Build GRCh38
rs72558416 Chr.X: 38403699-38403699 on Build GRCh38
rs569512666 Chr.X: 38403769-38403769 on Build GRCh38
rs72558414 Chr.X: 38403697-38403697 on Build GRCh38
rs72556290 Chr.X: 38403619-38403619 on Build GRCh38
rs72556298 Chr.X: 38403653-38403653 on Build GRCh38
rs72558426 Chr.X: 38403736-38403736 on Build GRCh38
rs72558405 Chr.X: 38403672-38403672 on Build GRCh38
rs72558417 Chr.X: 38403703-38403703 on Build GRCh38
rs374923521 Chr.X: 38403607-38403607 on Build GRCh38
rs759424912 Chr.X: 38403604-38403604 on Build GRCh38
rs72558410 Chr.X: 38403685-38403685 on Build GRCh38
rs772049322 Chr.X: 38403711-38403711 on Build GRCh38
rs72556296 Chr.X: 38403648-38403648 on Build GRCh38
rs72558421 Chr.X: 38403720-38403720 on Build GRCh38
rs66642398 Chr.X: 38403663-38403663 on Build GRCh38
rs72558407 Chr.X: 38403679-38403679 on Build GRCh38
rs72556301 Chr.X: 38403666-38403666 on Build GRCh38
rs72558420 Chr.X: 38403717-38403717 on Build GRCh38
rs72558422 Chr.X: 38403722-38403723 on Build GRCh38
rs72556292 Chr.X: 38403624-38403624 on Build GRCh38
rs72556294 Chr.X: 38403639-38403639 on Build GRCh38
rs72558403 Chr.X: 38403670-38403670 on Build GRCh38
rs767994989 Chr.X: 38403747-38403747 on Build GRCh38
rs188655698 Chr.X: 38403762-38403762 on Build GRCh38
rs72556291 Chr.X: 38403622-38403622 on Build GRCh38
rs72558413 Chr.X: 38403695-38403695 on Build GRCh38
rs796052012 Chr.X: 38403692-38403692 on Build GRCh38
rs72558409 Chr.X: 38403682-38403682 on Build GRCh38
rs200564773 Chr.X: 38403659-38403659 on Build GRCh38
rs779852274 Chr.X: 38403716-38403716 on Build GRCh38
rs72558406 Chr.X: 38403673-38403673 on Build GRCh38
rs72556295 Chr.X: 38403640-38403640 on Build GRCh38
rs756096405 Chr.X: 38403733-38403733 on Build GRCh38
rs777660144 Chr.X: 38403757-38403757 on Build GRCh38
rs72556299 Chr.X: 38403655-38403655 on Build GRCh38
rs72556300 Chr.X: 38403664-38403664 on Build GRCh38
rs281865552 Chr.X: 38403740-38403740 on Build GRCh38
rs67294956 Chr.X: 38403660-38403660 on Build GRCh38
rs72558412 Chr.X: 38403694-38403694 on Build GRCh38
rs72558427 Chr.X: 38403742-38403742 on Build GRCh38
rs745635680 Chr.X: 38403704-38403704 on Build GRCh38
rs72558425 Chr.X: 38403735-38403735 on Build GRCh38
rs756147250 Chr.X: 38403689-38403689 on Build GRCh38
rs796052016 Chr.X: 38403639-38403640 on Build GRCh38
rs72556297 Chr.X: 38403649-38403649 on Build GRCh38
rs72558411 Chr.X: 38403690-38403690 on Build GRCh38
rs372179665 Chr.X: 38403702-38403702 on Build GRCh38

View Full Product Details