Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15475836-15476073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGLYRP2
Entrez Gene ID
114770
Gene Name
peptidoglycan recognition protein 2
Gene Aliases
HMFT0141, PGLYRPL, PGRP-L, PGRPL, TAGL-like, tagL, tagL-alpha, tagl-beta
UniGene
Hs.282244
Gene Chromosome Location
Chr.19: 15468645-15479504 on Build GRCh38
Transcripts
NM_052890.3 XM_006722633.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4967039 c.776C>T p.T259M Chr.19: 15475894-15475894 on Build GRCh38
992268 c.694C>A p.L232I Chr.19: 15475976-15475976 on Build GRCh38
1129966 c.809T>A p.M270K Chr.19: 15475861-15475861 on Build GRCh38
317866 c.704C>A p.S235Y Chr.19: 15475966-15475966 on Build GRCh38
992265 c.700G>T p.G234C Chr.19: 15475970-15475970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757719223 Chr.19: 15476028-15476028 on Build GRCh38
rs767299609 Chr.19: 15475887-15475887 on Build GRCh38
rs150799464 Chr.19: 15475981-15475981 on Build GRCh38
rs765503342 Chr.19: 15475996-15475996 on Build GRCh38
rs370780981 Chr.19: 15475906-15475906 on Build GRCh38
rs751976579 Chr.19: 15476016-15476016 on Build GRCh38
rs746261223 Chr.19: 15475929-15475929 on Build GRCh38
rs761408370 Chr.19: 15475885-15475885 on Build GRCh38
rs35514478 Chr.19: 15476037-15476038 on Build GRCh38
rs758391088 Chr.19: 15476006-15476006 on Build GRCh38
rs771173904 Chr.19: 15475912-15475912 on Build GRCh38
rs373142402 Chr.19: 15475894-15475894 on Build GRCh38
rs760383491 Chr.19: 15475892-15475892 on Build GRCh38
rs140330499 Chr.19: 15475916-15475916 on Build GRCh38
rs374467861 Chr.19: 15476035-15476035 on Build GRCh38
rs768122396 Chr.19: 15475863-15475863 on Build GRCh38
rs141030763 Chr.19: 15475904-15475904 on Build GRCh38
rs750379441 Chr.19: 15475890-15475890 on Build GRCh38
rs747893560 Chr.19: 15476045-15476045 on Build GRCh38
rs755294160 Chr.19: 15475899-15475899 on Build GRCh38
rs376518031 Chr.19: 15475973-15475973 on Build GRCh38
rs766478335 Chr.19: 15475870-15475871 on Build GRCh38
rs147159919 Chr.19: 15475903-15475903 on Build GRCh38
rs892145 Chr.19: 15475861-15475861 on Build GRCh38
rs764281051 Chr.19: 15476009-15476009 on Build GRCh38
rs779380935 Chr.19: 15476042-15476042 on Build GRCh38
rs994730259 Chr.19: 15475987-15475987 on Build GRCh38
rs144572782 Chr.19: 15475969-15475969 on Build GRCh38
rs756474849 Chr.19: 15476036-15476036 on Build GRCh38
rs753025613 Chr.19: 15475997-15475997 on Build GRCh38
rs368081084 Chr.19: 15475953-15475953 on Build GRCh38
rs781574082 Chr.19: 15475920-15475920 on Build GRCh38
rs368743525 Chr.19: 15476048-15476048 on Build GRCh38
rs950979196 Chr.19: 15475877-15475877 on Build GRCh38
rs774004289 Chr.19: 15475956-15475956 on Build GRCh38
rs773808477 Chr.19: 15475882-15475882 on Build GRCh38
rs759430032 Chr.19: 15475983-15475983 on Build GRCh38
rs28404490 Chr.19: 15475900-15475900 on Build GRCh38
rs767287939 Chr.19: 15475893-15475893 on Build GRCh38
rs377488146 Chr.19: 15475962-15475962 on Build GRCh38
rs747605834 Chr.19: 15475907-15475907 on Build GRCh38
rs200781547 Chr.19: 15475972-15475972 on Build GRCh38
rs368981308 Chr.19: 15476034-15476034 on Build GRCh38

View Full Product Details