Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 13543965-13544214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKX3-2
Entrez Gene ID
579
Gene Name
NK3 homeobox 2
Gene Aliases
BAPX1, NKX3.2, NKX3B, SMMD
UniGene
Hs.590927
Gene Chromosome Location
Chr.4: 13540830-13544490 on Build GRCh38
Transcripts
NM_001189.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4953874 c.351C>G p.G117G Chr.4: 13544064-13544064 on Build GRCh38
4707096 c.337delG p.A113fs*11 Chr.4: 13544078-13544078 on Build GRCh38
4989534 c.323C>G p.A108G Chr.4: 13544092-13544092 on Build GRCh38
5422779 c.379C>A p.Q127K Chr.4: 13544036-13544036 on Build GRCh38
5449951 c.338C>G p.A113G Chr.4: 13544077-13544077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758415699 Chr.4: 13544160-13544160 on Build GRCh38
rs755427626 Chr.4: 13544164-13544164 on Build GRCh38
rs746486656 Chr.4: 13544112-13544112 on Build GRCh38
rs770494452 Chr.4: 13544121-13544121 on Build GRCh38
rs9684692 Chr.4: 13544168-13544168 on Build GRCh38
rs759321202 Chr.4: 13544090-13544090 on Build GRCh38
rs767003449 Chr.4: 13544001-13544001 on Build GRCh38
rs754091130 Chr.4: 13544161-13544161 on Build GRCh38
rs751869344 Chr.4: 13544138-13544138 on Build GRCh38
rs547806067 Chr.4: 13544183-13544183 on Build GRCh38
rs989621659 Chr.4: 13544115-13544115 on Build GRCh38
rs764167403 Chr.4: 13544137-13544137 on Build GRCh38
rs765431857 Chr.4: 13544049-13544049 on Build GRCh38
rs606231352 Chr.4: 13544077-13544078 on Build GRCh38
rs368263938 Chr.4: 13544039-13544039 on Build GRCh38
rs771590977 Chr.4: 13544079-13544079 on Build GRCh38
rs773171448 Chr.4: 13544032-13544032 on Build GRCh38
rs542815500 Chr.4: 13544100-13544100 on Build GRCh38
rs771586473 Chr.4: 13544109-13544109 on Build GRCh38
rs767746306 Chr.4: 13544152-13544152 on Build GRCh38
rs606231353 Chr.4: 13544078-13544079 on Build GRCh38
rs10032202 Chr.4: 13543986-13543986 on Build GRCh38
rs760235144 Chr.4: 13544083-13544083 on Build GRCh38
rs772550266 Chr.4: 13544081-13544081 on Build GRCh38
rs754779348 Chr.4: 13544042-13544042 on Build GRCh38
rs995678647 Chr.4: 13544006-13544006 on Build GRCh38
rs770400754 Chr.4: 13544172-13544172 on Build GRCh38
rs539837783 Chr.4: 13544065-13544065 on Build GRCh38
rs756642619 Chr.4: 13544154-13544154 on Build GRCh38
rs745410292 Chr.4: 13544177-13544177 on Build GRCh38
rs374854852 Chr.4: 13544191-13544191 on Build GRCh38
rs763034242 Chr.4: 13544095-13544095 on Build GRCh38
rs777761125 Chr.4: 13544054-13544054 on Build GRCh38
rs994351562 Chr.4: 13544030-13544030 on Build GRCh38
rs200167000 Chr.4: 13544180-13544180 on Build GRCh38
rs747796521 Chr.4: 13544078-13544078 on Build GRCh38
rs769742404 Chr.4: 13544130-13544130 on Build GRCh38
rs202191746 Chr.4: 13544092-13544092 on Build GRCh38
rs977897331 Chr.4: 13544097-13544097 on Build GRCh38
rs866703218 Chr.4: 13544067-13544067 on Build GRCh38
rs751529750 Chr.4: 13544098-13544098 on Build GRCh38
rs762814508 Chr.4: 13544131-13544131 on Build GRCh38
rs759515252 Chr.4: 13544123-13544123 on Build GRCh38
rs888537416 Chr.4: 13544048-13544048 on Build GRCh38
rs768566953 Chr.4: 13544189-13544189 on Build GRCh38
rs528913823 Chr.4: 13544099-13544099 on Build GRCh38
rs781766584 Chr.4: 13544060-13544060 on Build GRCh38
rs769650764 Chr.4: 13544179-13544179 on Build GRCh38
rs201772546 Chr.4: 13544142-13544142 on Build GRCh38
rs780614525 Chr.4: 13544102-13544102 on Build GRCh38
rs753562424 Chr.4: 13544040-13544040 on Build GRCh38
rs768359920 Chr.4: 13543987-13543987 on Build GRCh38
rs764405937 Chr.4: 13544096-13544096 on Build GRCh38
rs765052045 Chr.4: 13544091-13544091 on Build GRCh38
rs747132170 Chr.4: 13544058-13544058 on Build GRCh38
rs1005638560 Chr.4: 13544053-13544053 on Build GRCh38
rs112448315 Chr.4: 13544124-13544124 on Build GRCh38
rs145908097 Chr.4: 13544064-13544064 on Build GRCh38
rs756516183 Chr.4: 13544101-13544101 on Build GRCh38
rs552084314 Chr.4: 13544169-13544169 on Build GRCh38
rs571824367 Chr.4: 13544174-13544174 on Build GRCh38
rs752979207 Chr.4: 13544050-13544050 on Build GRCh38
rs758469136 Chr.4: 13544051-13544051 on Build GRCh38
rs774164249 Chr.4: 13543990-13543990 on Build GRCh38
rs563942204 Chr.4: 13544059-13544059 on Build GRCh38
rs749687098 Chr.4: 13544106-13544106 on Build GRCh38
rs761397954 Chr.4: 13543997-13543997 on Build GRCh38
rs776607528 Chr.4: 13544086-13544086 on Build GRCh38
rs901402069 Chr.4: 13543991-13543991 on Build GRCh38

View Full Product Details