Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 112285994-112286267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TECTB
Entrez Gene ID
6975
Gene Name
tectorin beta
Gene Aliases
-
UniGene
Hs.345485
Gene Chromosome Location
Chr.10: 112283656-112305035 on Build GRCh38
Transcripts
NM_058222.2 XM_017016604.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7092631 c.354C>T p.S118S Chr.10: 112286157-112286157 on Build GRCh38
5111966 c.330delA p.N112fs*18 Chr.10: 112286133-112286133 on Build GRCh38
218263 c.370C>A p.H124N Chr.10: 112286173-112286173 on Build GRCh38
3806451 c.387G>A p.V129V Chr.10: 112286190-112286190 on Build GRCh38
4733006 c.300C>A p.H100Q Chr.10: 112286103-112286103 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758206819 Chr.10: 112286234-112286234 on Build GRCh38
rs752390353 Chr.10: 112286068-112286068 on Build GRCh38
rs752558544 Chr.10: 112286131-112286131 on Build GRCh38
rs1055363289 Chr.10: 112286181-112286181 on Build GRCh38
rs752050151 Chr.10: 112286106-112286106 on Build GRCh38
rs770388534 Chr.10: 112286052-112286052 on Build GRCh38
rs746189488 Chr.10: 112286220-112286220 on Build GRCh38
rs376753403 Chr.10: 112286029-112286029 on Build GRCh38
rs756407563 Chr.10: 112286129-112286129 on Build GRCh38
rs775172936 Chr.10: 112286197-112286197 on Build GRCh38
rs778807306 Chr.10: 112286130-112286130 on Build GRCh38
rs373389035 Chr.10: 112286202-112286202 on Build GRCh38
rs746505949 Chr.10: 112286050-112286050 on Build GRCh38
rs1020185994 Chr.10: 112286062-112286062 on Build GRCh38
rs903090811 Chr.10: 112286069-112286069 on Build GRCh38
rs369178348 Chr.10: 112286162-112286162 on Build GRCh38
rs762243145 Chr.10: 112286205-112286205 on Build GRCh38
rs777734809 Chr.10: 112286138-112286138 on Build GRCh38
rs754152559 Chr.10: 112286228-112286228 on Build GRCh38
rs778860876 Chr.10: 112286040-112286040 on Build GRCh38
rs767562978 Chr.10: 112286208-112286208 on Build GRCh38
rs757333454 Chr.10: 112286026-112286026 on Build GRCh38
rs769461568 Chr.10: 112286192-112286192 on Build GRCh38
rs781449024 Chr.10: 112286027-112286027 on Build GRCh38
rs766682626 Chr.10: 112286227-112286227 on Build GRCh38
rs919342181 Chr.10: 112286098-112286098 on Build GRCh38
rs537248301 Chr.10: 112286204-112286204 on Build GRCh38
rs778869377 Chr.10: 112286233-112286233 on Build GRCh38
rs376277764 Chr.10: 112286144-112286144 on Build GRCh38
rs530455117 Chr.10: 112286103-112286103 on Build GRCh38
rs774561578 Chr.10: 112286145-112286145 on Build GRCh38
rs750558641 Chr.10: 112286213-112286213 on Build GRCh38
rs376075649 Chr.10: 112286239-112286239 on Build GRCh38
rs369962738 Chr.10: 112286107-112286107 on Build GRCh38
rs200939358 Chr.10: 112286245-112286245 on Build GRCh38
rs147615429 Chr.10: 112286077-112286077 on Build GRCh38
rs758078348 Chr.10: 112286224-112286224 on Build GRCh38
rs146778403 Chr.10: 112286142-112286142 on Build GRCh38
rs766997824 Chr.10: 112286056-112286056 on Build GRCh38
rs758316449 Chr.10: 112286132-112286132 on Build GRCh38
rs763401490 Chr.10: 112286097-112286097 on Build GRCh38
rs770079500 Chr.10: 112286088-112286088 on Build GRCh38
rs1011064125 Chr.10: 112286201-112286201 on Build GRCh38
rs908576161 Chr.10: 112286206-112286206 on Build GRCh38
rs750612131 Chr.10: 112286116-112286116 on Build GRCh38
rs746145914 Chr.10: 112286028-112286028 on Build GRCh38
rs118152880 Chr.10: 112286166-112286166 on Build GRCh38
rs777486967 Chr.10: 112286236-112286236 on Build GRCh38
rs772163784 Chr.10: 112286048-112286048 on Build GRCh38
rs199962432 Chr.10: 112286093-112286093 on Build GRCh38
rs754734402 Chr.10: 112286046-112286049 on Build GRCh38
rs748030223 Chr.10: 112286044-112286044 on Build GRCh38
rs201881485 Chr.10: 112286199-112286199 on Build GRCh38

View Full Product Details