Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 129855661-129855934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM45B
Entrez Gene ID
120224
Gene Name
transmembrane protein 45B
Gene Aliases
-
UniGene
Hs.504301
Gene Chromosome Location
Chr.11: 129815781-129860003 on Build GRCh38
Transcripts
NM_138788.3 XM_005271400.4 XM_011542586.2 XM_011542588.2 XM_017017188.1 XM_017017189.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5540535 c.467G>A p.G156E Chr.11: 129855789-129855789 on Build GRCh38
7133416 c.427G>C p.D143H Chr.11: 129855749-129855749 on Build GRCh38
5372032 c.470G>A p.G157E Chr.11: 129855792-129855792 on Build GRCh38
1676582 c.485C>T p.S162F Chr.11: 129855807-129855807 on Build GRCh38
6804087 c.435C>T p.H145H Chr.11: 129855757-129855757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024374759 Chr.11: 129855887-129855887 on Build GRCh38
rs771984526 Chr.11: 129855804-129855804 on Build GRCh38
rs773317053 Chr.11: 129855905-129855905 on Build GRCh38
rs980784116 Chr.11: 129855683-129855683 on Build GRCh38
rs769553545 Chr.11: 129855727-129855727 on Build GRCh38
rs377214268 Chr.11: 129855854-129855854 on Build GRCh38
rs377463069 Chr.11: 129855912-129855912 on Build GRCh38
rs959880355 Chr.11: 129855818-129855818 on Build GRCh38
rs148622359 Chr.11: 129855808-129855808 on Build GRCh38
rs143254724 Chr.11: 129855865-129855865 on Build GRCh38
rs755487204 Chr.11: 129855757-129855757 on Build GRCh38
rs767267855 Chr.11: 129855811-129855812 on Build GRCh38
rs756598307 Chr.11: 129855737-129855737 on Build GRCh38
rs752605654 Chr.11: 129855869-129855870 on Build GRCh38
rs764124298 Chr.11: 129855736-129855736 on Build GRCh38
rs368213460 Chr.11: 129855789-129855789 on Build GRCh38
rs780431156 Chr.11: 129855705-129855705 on Build GRCh38
rs375699692 Chr.11: 129855744-129855744 on Build GRCh38
rs750578484 Chr.11: 129855745-129855745 on Build GRCh38
rs142486105 Chr.11: 129855769-129855769 on Build GRCh38
rs146247963 Chr.11: 129855730-129855730 on Build GRCh38
rs549481514 Chr.11: 129855825-129855825 on Build GRCh38
rs934161191 Chr.11: 129855884-129855884 on Build GRCh38
rs759481364 Chr.11: 129855735-129855735 on Build GRCh38
rs755705345 Chr.11: 129855834-129855834 on Build GRCh38
rs139993952 Chr.11: 129855852-129855852 on Build GRCh38
rs567218321 Chr.11: 129855740-129855740 on Build GRCh38
rs780902733 Chr.11: 129855760-129855760 on Build GRCh38
rs771464054 Chr.11: 129855883-129855883 on Build GRCh38
rs762080530 Chr.11: 129855738-129855738 on Build GRCh38
rs748935225 Chr.11: 129855844-129855844 on Build GRCh38
rs781126896 Chr.11: 129855723-129855723 on Build GRCh38
rs142972985 Chr.11: 129855821-129855821 on Build GRCh38
rs771180226 Chr.11: 129855907-129855907 on Build GRCh38
rs750774015 Chr.11: 129855686-129855686 on Build GRCh38
rs151103658 Chr.11: 129855832-129855832 on Build GRCh38
rs779299054 Chr.11: 129855684-129855684 on Build GRCh38
rs896983090 Chr.11: 129855880-129855880 on Build GRCh38
rs1026063551 Chr.11: 129855810-129855810 on Build GRCh38
rs773452606 Chr.11: 129855812-129855812 on Build GRCh38
rs538053574 Chr.11: 129855913-129855913 on Build GRCh38
rs888891511 Chr.11: 129855765-129855765 on Build GRCh38
rs751934721 Chr.11: 129855824-129855824 on Build GRCh38
rs934934425 Chr.11: 129855733-129855733 on Build GRCh38
rs139170609 Chr.11: 129855766-129855766 on Build GRCh38
rs879234499 Chr.11: 129855893-129855893 on Build GRCh38
rs747499534 Chr.11: 129855707-129855707 on Build GRCh38
rs758705233 Chr.11: 129855746-129855746 on Build GRCh38
rs766724799 Chr.11: 129855748-129855748 on Build GRCh38
rs774168527 Chr.11: 129855799-129855799 on Build GRCh38
rs145935291 Chr.11: 129855801-129855801 on Build GRCh38
rs561834458 Chr.11: 129855761-129855761 on Build GRCh38
rs925952243 Chr.11: 129855851-129855851 on Build GRCh38
rs527243028 Chr.11: 129855787-129855787 on Build GRCh38
rs547067888 Chr.11: 129855788-129855788 on Build GRCh38
rs149287572 Chr.11: 129855729-129855729 on Build GRCh38
rs777380173 Chr.11: 129855841-129855841 on Build GRCh38
rs371815772 Chr.11: 129855728-129855728 on Build GRCh38
rs775422659 Chr.11: 129855875-129855875 on Build GRCh38
rs759012678 Chr.11: 129855689-129855689 on Build GRCh38
rs200613797 Chr.11: 129855709-129855709 on Build GRCh38
rs532840639 Chr.11: 129855817-129855817 on Build GRCh38
rs747852127 Chr.11: 129855720-129855720 on Build GRCh38
rs756912301 Chr.11: 129855847-129855847 on Build GRCh38
rs780816954 Chr.11: 129855712-129855712 on Build GRCh38
rs746902993 Chr.11: 129855882-129855882 on Build GRCh38
rs201855054 Chr.11: 129855870-129855870 on Build GRCh38
rs761888055 Chr.11: 129855805-129855805 on Build GRCh38
rs878905895 Chr.11: 129855892-129855893 on Build GRCh38
rs752045829 Chr.11: 129855754-129855754 on Build GRCh38

View Full Product Details