Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32945423-32945692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF19B
Entrez Gene ID
127544
Gene Name
ring finger protein 19B
Gene Aliases
IBRDC3, NKLAM
UniGene
Hs.591504
Gene Chromosome Location
Chr.1: 32932498-32965266 on Build GRCh38
Transcripts
NM_001127361.2 NM_001300826.1 NM_153341.3 XM_006710356.2 XM_006710357.3 XM_006710358.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5846419 c.670G>A p.V224I Chr.1: 32945535-32945535 on Build GRCh38
4031561 c.653C>T p.S218L Chr.1: 32945552-32945552 on Build GRCh38
5846418 c.1240G>A p.V414I Chr.1: 32945535-32945535 on Build GRCh38
6575934 c.645G>A p.V215V Chr.1: 32945560-32945560 on Build GRCh38
673784 c.691G>T p.G231C Chr.1: 32945514-32945514 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375304223 Chr.1: 32945613-32945613 on Build GRCh38
rs745335745 Chr.1: 32945638-32945687 on Build GRCh38
rs767772345 Chr.1: 32945518-32945518 on Build GRCh38
rs962213636 Chr.1: 32945645-32945645 on Build GRCh38
rs776423739 Chr.1: 32945622-32945622 on Build GRCh38
rs770385076 Chr.1: 32945496-32945497 on Build GRCh38
rs773970586 Chr.1: 32945636-32945636 on Build GRCh38
rs1029124119 Chr.1: 32945592-32945592 on Build GRCh38
rs759594850 Chr.1: 32945517-32945517 on Build GRCh38
rs766447585 Chr.1: 32945469-32945469 on Build GRCh38
rs149031975 Chr.1: 32945536-32945536 on Build GRCh38
rs746628622 Chr.1: 32945611-32945611 on Build GRCh38
rs957224992 Chr.1: 32945531-32945531 on Build GRCh38
rs779608521 Chr.1: 32945606-32945606 on Build GRCh38
rs1041356073 Chr.1: 32945496-32945496 on Build GRCh38
rs568234965 Chr.1: 32945590-32945590 on Build GRCh38
rs768352750 Chr.1: 32945617-32945617 on Build GRCh38
rs757907579 Chr.1: 32945577-32945577 on Build GRCh38
rs745419943 Chr.1: 32945585-32945585 on Build GRCh38
rs756466037 Chr.1: 32945550-32945550 on Build GRCh38
rs770083621 Chr.1: 32945499-32945499 on Build GRCh38
rs1034115182 Chr.1: 32945453-32945453 on Build GRCh38
rs761589664 Chr.1: 32945636-32945636 on Build GRCh38
rs200921038 Chr.1: 32945593-32945593 on Build GRCh38
rs751702823 Chr.1: 32945493-32945493 on Build GRCh38
rs778148778 Chr.1: 32945558-32945558 on Build GRCh38
rs992028354 Chr.1: 32945649-32945649 on Build GRCh38
rs761755779 Chr.1: 32945467-32945467 on Build GRCh38
rs1014740093 Chr.1: 32945615-32945615 on Build GRCh38
rs779552308 Chr.1: 32945583-32945583 on Build GRCh38
rs892652017 Chr.1: 32945449-32945449 on Build GRCh38
rs1006442562 Chr.1: 32945602-32945602 on Build GRCh38
rs975874092 Chr.1: 32945646-32945646 on Build GRCh38
rs200028105 Chr.1: 32945574-32945574 on Build GRCh38

View Full Product Details