Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 73030253-73030512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TERF1
Entrez Gene ID
7013
Gene Name
telomeric repeat binding factor 1
Gene Aliases
PIN2, TRBF1, TRF, TRF1, hTRF1-AS, t-TRF1
UniGene
Hs.442707
Gene Chromosome Location
Chr.8: 73008862-73047752 on Build GRCh38
Transcripts
NM_003218.3 NM_017489.2 XM_005251291.3 XM_005251292.3 XM_011517582.2 XM_017013792.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
354897 c.908C>T p.A303V Chr.8: 73030356-73030356 on Build GRCh38
5457404 c.933A>G p.T311T Chr.8: 73030381-73030381 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772982906 Chr.8: 73030358-73030358 on Build GRCh38
rs780617179 Chr.8: 73030443-73030443 on Build GRCh38
rs34742076 Chr.8: 73030477-73030477 on Build GRCh38
rs779191169 Chr.8: 73030285-73030285 on Build GRCh38
rs904609555 Chr.8: 73030335-73030335 on Build GRCh38
rs932717117 Chr.8: 73030288-73030288 on Build GRCh38
rs886935823 Chr.8: 73030400-73030400 on Build GRCh38
rs368465062 Chr.8: 73030435-73030435 on Build GRCh38
rs772528548 Chr.8: 73030296-73030296 on Build GRCh38
rs34231129 Chr.8: 73030286-73030286 on Build GRCh38
rs761142736 Chr.8: 73030344-73030347 on Build GRCh38
rs891688589 Chr.8: 73030386-73030386 on Build GRCh38
rs746726285 Chr.8: 73030431-73030431 on Build GRCh38
rs1001794731 Chr.8: 73030478-73030478 on Build GRCh38
rs562567411 Chr.8: 73030398-73030398 on Build GRCh38
rs750848681 Chr.8: 73030415-73030415 on Build GRCh38
rs1053376388 Chr.8: 73030382-73030382 on Build GRCh38
rs941328799 Chr.8: 73030403-73030403 on Build GRCh38
rs758851360 Chr.8: 73030426-73030426 on Build GRCh38
rs943317944 Chr.8: 73030315-73030315 on Build GRCh38
rs766103414 Chr.8: 73030373-73030373 on Build GRCh38
rs752919794 Chr.8: 73030380-73030380 on Build GRCh38
rs780503748 Chr.8: 73030428-73030428 on Build GRCh38
rs754353731 Chr.8: 73030421-73030423 on Build GRCh38
rs188011159 Chr.8: 73030357-73030357 on Build GRCh38
rs34940416 Chr.8: 73030481-73030481 on Build GRCh38
rs760878637 Chr.8: 73030390-73030390 on Build GRCh38
rs773427290 Chr.8: 73030322-73030324 on Build GRCh38
rs768286752 Chr.8: 73030345-73030345 on Build GRCh38
rs533827129 Chr.8: 73030356-73030356 on Build GRCh38
rs746605713 Chr.8: 73030330-73030330 on Build GRCh38
rs201072411 Chr.8: 73030410-73030410 on Build GRCh38
rs779877917 Chr.8: 73030300-73030300 on Build GRCh38
rs764212680 Chr.8: 73030391-73030391 on Build GRCh38
rs570512700 Chr.8: 73030396-73030396 on Build GRCh38
rs576926318 Chr.8: 73030366-73030366 on Build GRCh38
rs1049879260 Chr.8: 73030303-73030303 on Build GRCh38
rs541761052 Chr.8: 73030453-73030453 on Build GRCh38
rs976179029 Chr.8: 73030316-73030316 on Build GRCh38
rs766793774 Chr.8: 73030414-73030417 on Build GRCh38
rs969728793 Chr.8: 73030476-73030476 on Build GRCh38
rs201891730 Chr.8: 73030322-73030322 on Build GRCh38

View Full Product Details