Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 194687279-194687552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM43A
Entrez Gene ID
131583
Gene Name
family with sequence similarity 43 member A
Gene Aliases
-
UniGene
Hs.435080
Gene Chromosome Location
Chr.3: 194685893-194689037 on Build GRCh38
Transcripts
NM_153690.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1421805 c.696C>T p.H232H Chr.3: 194687522-194687522 on Build GRCh38
40071 c.611G>A p.R204Q Chr.3: 194687437-194687437 on Build GRCh38
6412605 c.718C>A p.P240T Chr.3: 194687544-194687544 on Build GRCh38
7274840 c.595T>C p.F199L Chr.3: 194687421-194687421 on Build GRCh38
7260593 c.583G>A p.A195T Chr.3: 194687409-194687409 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757519625 Chr.3: 194687411-194687411 on Build GRCh38
rs780580491 Chr.3: 194687506-194687506 on Build GRCh38
rs754023956 Chr.3: 194687318-194687318 on Build GRCh38
rs751347688 Chr.3: 194687466-194687466 on Build GRCh38
rs868580559 Chr.3: 194687319-194687319 on Build GRCh38
rs1025200912 Chr.3: 194687401-194687401 on Build GRCh38
rs759357016 Chr.3: 194687480-194687480 on Build GRCh38
rs750505092 Chr.3: 194687505-194687505 on Build GRCh38
rs754077007 Chr.3: 194687488-194687488 on Build GRCh38
rs1015105670 Chr.3: 194687375-194687375 on Build GRCh38
rs141624284 Chr.3: 194687330-194687330 on Build GRCh38
rs766332460 Chr.3: 194687456-194687456 on Build GRCh38
rs551024809 Chr.3: 194687349-194687349 on Build GRCh38
rs755295104 Chr.3: 194687520-194687520 on Build GRCh38
rs995595523 Chr.3: 194687336-194687336 on Build GRCh38
rs762853921 Chr.3: 194687440-194687440 on Build GRCh38
rs747738629 Chr.3: 194687528-194687528 on Build GRCh38
rs757365617 Chr.3: 194687325-194687325 on Build GRCh38
rs747395821 Chr.3: 194687368-194687368 on Build GRCh38
rs773212595 Chr.3: 194687435-194687435 on Build GRCh38
rs868592145 Chr.3: 194687403-194687403 on Build GRCh38
rs768347113 Chr.3: 194687381-194687381 on Build GRCh38
rs780824565 Chr.3: 194687523-194687523 on Build GRCh38
rs776216684 Chr.3: 194687402-194687402 on Build GRCh38
rs536841751 Chr.3: 194687380-194687380 on Build GRCh38
rs1026261298 Chr.3: 194687348-194687348 on Build GRCh38
rs887745829 Chr.3: 194687339-194687339 on Build GRCh38
rs757420477 Chr.3: 194687501-194687501 on Build GRCh38
rs569557732 Chr.3: 194687360-194687360 on Build GRCh38
rs898613204 Chr.3: 194687321-194687321 on Build GRCh38
rs765352022 Chr.3: 194687502-194687502 on Build GRCh38
rs761342624 Chr.3: 194687407-194687407 on Build GRCh38
rs769224916 Chr.3: 194687417-194687417 on Build GRCh38
rs1013861920 Chr.3: 194687382-194687382 on Build GRCh38
rs555267876 Chr.3: 194687510-194687510 on Build GRCh38
rs989900912 Chr.3: 194687527-194687527 on Build GRCh38
rs764438000 Chr.3: 194687482-194687482 on Build GRCh38
rs748069915 Chr.3: 194687492-194687492 on Build GRCh38

View Full Product Details