Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111534307-111534572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf88
Entrez Gene ID
399949
Gene Name
chromosome 11 open reading frame 88
Gene Aliases
-
UniGene
Hs.411600
Gene Chromosome Location
Chr.11: 111514785-111537031 on Build GRCh38
Transcripts
NM_001100388.1 NM_207430.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4497555 c.499C>T p.P167S Chr.11: 111534430-111534430 on Build GRCh38
7077174 c.481-1G>A p.? Chr.11: 111534411-111534411 on Build GRCh38
6614150 c.487C>A p.L163M Chr.11: 111534418-111534418 on Build GRCh38
2111553 c.501G>A p.P167P Chr.11: 111534432-111534432 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771057725 Chr.11: 111534444-111534444 on Build GRCh38
rs375225058 Chr.11: 111534478-111534478 on Build GRCh38
rs990144507 Chr.11: 111534517-111534517 on Build GRCh38
rs140132406 Chr.11: 111534341-111534341 on Build GRCh38
rs201561292 Chr.11: 111534445-111534445 on Build GRCh38
rs751533569 Chr.11: 111534368-111534368 on Build GRCh38
rs113149376 Chr.11: 111534378-111534378 on Build GRCh38
rs747796985 Chr.11: 111534481-111534481 on Build GRCh38
rs185152888 Chr.11: 111534528-111534528 on Build GRCh38
rs754724031 Chr.11: 111534374-111534374 on Build GRCh38
rs749965811 Chr.11: 111534362-111534362 on Build GRCh38
rs769502039 Chr.11: 111534486-111534486 on Build GRCh38
rs762526210 Chr.11: 111534364-111534364 on Build GRCh38
rs756388978 Chr.11: 111534432-111534432 on Build GRCh38
rs749262432 Chr.11: 111534440-111534440 on Build GRCh38
rs140802590 Chr.11: 111534510-111534510 on Build GRCh38
rs981823215 Chr.11: 111534477-111534477 on Build GRCh38
rs746418326 Chr.11: 111534476-111534476 on Build GRCh38
rs777828106 Chr.11: 111534434-111534434 on Build GRCh38
rs772928273 Chr.11: 111534515-111534515 on Build GRCh38
rs771073414 Chr.11: 111534544-111534545 on Build GRCh38
rs781010159 Chr.11: 111534409-111534409 on Build GRCh38
rs12270454 Chr.11: 111534395-111534395 on Build GRCh38
rs941059893 Chr.11: 111534512-111534512 on Build GRCh38
rs549928086 Chr.11: 111534543-111534543 on Build GRCh38
rs2298739 Chr.11: 111534365-111534365 on Build GRCh38
rs751342520 Chr.11: 111534396-111534400 on Build GRCh38
rs973775466 Chr.11: 111534522-111534522 on Build GRCh38
rs61753535 Chr.11: 111534431-111534431 on Build GRCh38

View Full Product Details