Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32182402-32182627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGER
Entrez Gene ID
177
Gene Name
advanced glycosylation end product-specific receptor
Gene Aliases
RAGE, SCARJ1
UniGene
Hs.534342
Gene Chromosome Location
Chr.6: 32180968-32185247 on Build GRCh38
Transcripts
NM_001136.4 NM_001206929.1 NM_001206932.1 NM_001206934.1 NM_001206936.1 NM_001206940.1 NM_001206954.1 NM_001206966.1 NM_172197.2 NR_038190.1 XM_017010328.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6205687 c.883C>T p.Q295* Chr.6: 32182600-32182600 on Build GRCh38
5441396 c.819G>A p.L273L Chr.6: 32182619-32182619 on Build GRCh38
328183 c.811T>A p.W271R Chr.6: 32182579-32182579 on Build GRCh38
6205686 c.748C>T p.Q250* Chr.6: 32182600-32182600 on Build GRCh38
6205685 c.838C>T p.Q280* Chr.6: 32182600-32182600 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772866400 Chr.6: 32182492-32182492 on Build GRCh38
rs746666846 Chr.6: 32182570-32182570 on Build GRCh38
rs770666915 Chr.6: 32182495-32182495 on Build GRCh38
rs184003 Chr.6: 32182519-32182519 on Build GRCh38
rs755557287 Chr.6: 32182430-32182430 on Build GRCh38
rs771700388 Chr.6: 32182491-32182491 on Build GRCh38
rs761693422 Chr.6: 32182567-32182567 on Build GRCh38
rs777368483 Chr.6: 32182568-32182568 on Build GRCh38
rs747726238 Chr.6: 32182440-32182440 on Build GRCh38
rs779456062 Chr.6: 32182550-32182550 on Build GRCh38
rs778364595 Chr.6: 32182437-32182437 on Build GRCh38
rs752145121 Chr.6: 32182552-32182552 on Build GRCh38
rs768455447 Chr.6: 32182442-32182442 on Build GRCh38
rs1008075972 Chr.6: 32182546-32182546 on Build GRCh38
rs936884506 Chr.6: 32182443-32182443 on Build GRCh38
rs150335895 Chr.6: 32182495-32182496 on Build GRCh38
rs774557097 Chr.6: 32182515-32182515 on Build GRCh38
rs750677811 Chr.6: 32182576-32182577 on Build GRCh38
rs767664023 Chr.6: 32182549-32182550 on Build GRCh38
rs769835092 Chr.6: 32182499-32182499 on Build GRCh38
rs766628456 Chr.6: 32182544-32182544 on Build GRCh38
rs755437046 Chr.6: 32182545-32182545 on Build GRCh38
rs13209187 Chr.6: 32182477-32182477 on Build GRCh38
rs144335694 Chr.6: 32182526-32182526 on Build GRCh38
rs1052682431 Chr.6: 32182468-32182468 on Build GRCh38
rs761050938 Chr.6: 32182543-32182543 on Build GRCh38
rs780821490 Chr.6: 32182600-32182600 on Build GRCh38
rs58892390 Chr.6: 32182522-32182522 on Build GRCh38
rs140930365 Chr.6: 32182579-32182579 on Build GRCh38
rs13212247 Chr.6: 32182535-32182535 on Build GRCh38
rs745731408 Chr.6: 32182605-32182605 on Build GRCh38
rs186754929 Chr.6: 32182494-32182494 on Build GRCh38
rs13209183 Chr.6: 32182469-32182469 on Build GRCh38
rs943143587 Chr.6: 32182461-32182461 on Build GRCh38

View Full Product Details