Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47475315-47475578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF1
Entrez Gene ID
10658
Gene Name
CUGBP, Elav-like family member 1
Gene Aliases
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
UniGene
Hs.595333
Gene Chromosome Location
Chr.11: 47465937-47565569 on Build GRCh38
Transcripts
NM_001025596.2 NM_001172639.1 NM_001172640.1 NM_006560.3 NM_198700.2 XM_011519848.2 XM_011519850.2 XM_011519852.2 XM_011519854.1 XM_011519855.1 XM_011519856.1 XM_011519857.1 XM_011519858.1 XM_011519859.1 XM_017017101.1 XM_017017102.1 XM_017017103.1 XM_017017104.1 XM_017017105.1 XM_017017106.1 XM_017017107.1 XM_017017108.1 XM_017017109.1 XM_017017110.1 XM_017017111.1 XM_017017112.1 XM_017017113.1 XM_017017114.1 XM_017017115.1 XM_017017116.1 XM_017017117.1 XM_017017118.1 XM_017017119.1 XM_017017120.1 XM_017017121.1 XM_017017122.1 XM_017017123.1 XM_017017124.1 XM_017017125.1 XM_017017126.1 XM_017017127.1 XM_017017128.1 XM_017017129.1 XM_017017130.1 XM_017017131.1 XM_017017132.1 XM_017017133.1 XM_017017134.1 XM_017017135.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7312018 c.1041C>A p.G347G Chr.11: 47475484-47475484 on Build GRCh38
3447836 c.1156C>T p.Q386* Chr.11: 47475369-47475369 on Build GRCh38
3397720 c.1118C>T p.A373V Chr.11: 47475407-47475407 on Build GRCh38
6410348 c.1072G>T p.A358S Chr.11: 47475453-47475453 on Build GRCh38
6430707 c.1037G>T p.S346I Chr.11: 47475488-47475488 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039990907 Chr.11: 47475463-47475463 on Build GRCh38
rs1045245622 Chr.11: 47475363-47475363 on Build GRCh38
rs748800033 Chr.11: 47475540-47475540 on Build GRCh38
rs755756226 Chr.11: 47475489-47475490 on Build GRCh38
rs201396363 Chr.11: 47475549-47475549 on Build GRCh38
rs138642328 Chr.11: 47475453-47475453 on Build GRCh38
rs752515334 Chr.11: 47475421-47475421 on Build GRCh38
rs933939864 Chr.11: 47475506-47475506 on Build GRCh38
rs755488939 Chr.11: 47475496-47475496 on Build GRCh38
rs371919477 Chr.11: 47475498-47475498 on Build GRCh38
rs772774823 Chr.11: 47475376-47475376 on Build GRCh38
rs780643943 Chr.11: 47475469-47475469 on Build GRCh38
rs549182146 Chr.11: 47475550-47475550 on Build GRCh38
rs1052453430 Chr.11: 47475424-47475424 on Build GRCh38
rs776538692 Chr.11: 47475499-47475499 on Build GRCh38
rs376361989 Chr.11: 47475533-47475533 on Build GRCh38
rs943362358 Chr.11: 47475368-47475368 on Build GRCh38
rs774366238 Chr.11: 47475548-47475548 on Build GRCh38
rs936787594 Chr.11: 47475418-47475418 on Build GRCh38
rs370804375 Chr.11: 47475377-47475377 on Build GRCh38
rs776368969 Chr.11: 47475357-47475357 on Build GRCh38
rs767221177 Chr.11: 47475368-47475368 on Build GRCh38
rs567789492 Chr.11: 47475542-47475542 on Build GRCh38
rs565562546 Chr.11: 47475407-47475407 on Build GRCh38
rs550992692 Chr.11: 47475488-47475488 on Build GRCh38
rs552070800 Chr.11: 47475373-47475373 on Build GRCh38
rs1041544061 Chr.11: 47475512-47475512 on Build GRCh38
rs368359967 Chr.11: 47475439-47475439 on Build GRCh38
rs1008243646 Chr.11: 47475500-47475500 on Build GRCh38
rs767164612 Chr.11: 47475411-47475411 on Build GRCh38
rs375230946 Chr.11: 47475396-47475396 on Build GRCh38
rs757623542 Chr.11: 47475429-47475429 on Build GRCh38
rs530956678 Chr.11: 47475468-47475468 on Build GRCh38
rs865992638 Chr.11: 47475476-47475476 on Build GRCh38
rs373086764 Chr.11: 47475541-47475541 on Build GRCh38
rs925447400 Chr.11: 47475406-47475406 on Build GRCh38
rs749959777 Chr.11: 47475487-47475488 on Build GRCh38
rs761503748 Chr.11: 47475358-47475358 on Build GRCh38
rs774087153 Chr.11: 47475403-47475403 on Build GRCh38
rs772792797 Chr.11: 47475535-47475535 on Build GRCh38
rs866597190 Chr.11: 47475434-47475434 on Build GRCh38
rs201135792 Chr.11: 47475484-47475484 on Build GRCh38

View Full Product Details