Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 94400091-94400352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL1A2
Entrez Gene ID
1278
Gene Name
collagen type I alpha 2 chain
Gene Aliases
OI4
UniGene
Hs.489142
Gene Chromosome Location
Chr.7: 94394561-94431232 on Build GRCh38
Transcripts
NM_000089.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3883463 c.196G>T p.G66C Chr.7: 94400259-94400259 on Build GRCh38
1496710 c.133-2A>T p.? Chr.7: 94400194-94400194 on Build GRCh38
1093245 c.157G>T p.D53Y Chr.7: 94400220-94400220 on Build GRCh38
4529529 c.161G>A p.G54D Chr.7: 94400224-94400224 on Build GRCh38
6632756 c.212C>T p.P71L Chr.7: 94400275-94400275 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374356493 Chr.7: 94400276-94400276 on Build GRCh38
rs756659600 Chr.7: 94400243-94400243 on Build GRCh38
rs868627404 Chr.7: 94400195-94400195 on Build GRCh38
rs760621026 Chr.7: 94400128-94400128 on Build GRCh38
rs779932887 Chr.7: 94400246-94400247 on Build GRCh38
rs576980117 Chr.7: 94400118-94400118 on Build GRCh38
rs141279380 Chr.7: 94400169-94400169 on Build GRCh38
rs754484189 Chr.7: 94400309-94400312 on Build GRCh38
rs563774670 Chr.7: 94400127-94400127 on Build GRCh38
rs745496110 Chr.7: 94400267-94400267 on Build GRCh38
rs751435484 Chr.7: 94400302-94400302 on Build GRCh38
rs546389896 Chr.7: 94400155-94400155 on Build GRCh38
rs370095931 Chr.7: 94400179-94400179 on Build GRCh38
rs200306037 Chr.7: 94400173-94400173 on Build GRCh38
rs755226579 Chr.7: 94400156-94400156 on Build GRCh38
rs775240284 Chr.7: 94400274-94400274 on Build GRCh38
rs776365460 Chr.7: 94400283-94400283 on Build GRCh38
rs748798643 Chr.7: 94400177-94400177 on Build GRCh38
rs753406780 Chr.7: 94400264-94400265 on Build GRCh38
rs760571966 Chr.7: 94400208-94400208 on Build GRCh38
rs765294984 Chr.7: 94400134-94400134 on Build GRCh38
rs756667586 Chr.7: 94400224-94400224 on Build GRCh38
rs879344129 Chr.7: 94400211-94400211 on Build GRCh38
rs1800221 Chr.7: 94400238-94400238 on Build GRCh38
rs770491939 Chr.7: 94400171-94400171 on Build GRCh38
rs752565906 Chr.7: 94400116-94400116 on Build GRCh38
rs148639088 Chr.7: 94400201-94400201 on Build GRCh38
rs780090471 Chr.7: 94400143-94400143 on Build GRCh38
rs370976741 Chr.7: 94400141-94400141 on Build GRCh38
rs771749616 Chr.7: 94400271-94400271 on Build GRCh38
rs113180440 Chr.7: 94400194-94400194 on Build GRCh38
rs992330364 Chr.7: 94400181-94400181 on Build GRCh38
rs771800420 Chr.7: 94400202-94400202 on Build GRCh38
rs935909926 Chr.7: 94400135-94400135 on Build GRCh38
rs267601641 Chr.7: 94400241-94400241 on Build GRCh38
rs772333670 Chr.7: 94400130-94400130 on Build GRCh38
rs750541300 Chr.7: 94400136-94400136 on Build GRCh38
rs774555831 Chr.7: 94400303-94400303 on Build GRCh38
rs781697789 Chr.7: 94400167-94400167 on Build GRCh38
rs773839903 Chr.7: 94400191-94400191 on Build GRCh38
rs759802957 Chr.7: 94400310-94400310 on Build GRCh38
rs201137000 Chr.7: 94400232-94400232 on Build GRCh38
rs201699348 Chr.7: 94400222-94400222 on Build GRCh38
rs768467735 Chr.7: 94400282-94400282 on Build GRCh38
rs369695645 Chr.7: 94400213-94400213 on Build GRCh38
rs767987949 Chr.7: 94400219-94400219 on Build GRCh38
rs764064979 Chr.7: 94400209-94400209 on Build GRCh38
rs561632203 Chr.7: 94400218-94400218 on Build GRCh38
rs761823309 Chr.7: 94400285-94400285 on Build GRCh38
rs778502385 Chr.7: 94400261-94400261 on Build GRCh38
rs377637698 Chr.7: 94400212-94400212 on Build GRCh38

View Full Product Details