Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43340176-43340439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAL
Entrez Gene ID
161823
Gene Name
adenosine deaminase like
Gene Aliases
-
UniGene
Hs.533913
Gene Chromosome Location
Chr.15: 43330351-43354555 on Build GRCh38
Transcripts
NM_001012969.3 NM_001159280.2 NM_001324364.1 NM_001324365.1 NM_001324366.1 NM_001324367.1 NM_001324368.1 XM_011521291.2 XM_017021964.1 XM_017021965.1 XM_017021966.1 XM_017021967.1 XM_017021968.1 XM_017021969.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1162073 c.323C>T p.P108L Chr.15: 43340332-43340332 on Build GRCh38
6838784 c.341C>G p.T114S Chr.15: 43340350-43340350 on Build GRCh38
1587860 c.276A>C p.K92N Chr.15: 43340285-43340285 on Build GRCh38
6593337 c.292G>A p.G98S Chr.15: 43340301-43340301 on Build GRCh38
1587859 c.277G>T p.E93* Chr.15: 43340286-43340286 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs887077306 Chr.15: 43340206-43340206 on Build GRCh38
rs758524039 Chr.15: 43340219-43340219 on Build GRCh38
rs986993705 Chr.15: 43340321-43340321 on Build GRCh38
rs745727213 Chr.15: 43340278-43340278 on Build GRCh38
rs773946552 Chr.15: 43340299-43340299 on Build GRCh38
rs1046941217 Chr.15: 43340394-43340394 on Build GRCh38
rs771913927 Chr.15: 43340370-43340370 on Build GRCh38
rs201947910 Chr.15: 43340300-43340300 on Build GRCh38
rs765890584 Chr.15: 43340303-43340303 on Build GRCh38
rs780378127 Chr.15: 43340234-43340234 on Build GRCh38
rs762376532 Chr.15: 43340301-43340301 on Build GRCh38
rs763437323 Chr.15: 43340316-43340316 on Build GRCh38
rs180869501 Chr.15: 43340333-43340333 on Build GRCh38
rs2278857 Chr.15: 43340351-43340351 on Build GRCh38
rs779523086 Chr.15: 43340373-43340373 on Build GRCh38
rs778650386 Chr.15: 43340272-43340272 on Build GRCh38
rs372506209 Chr.15: 43340324-43340324 on Build GRCh38
rs185470752 Chr.15: 43340340-43340340 on Build GRCh38
rs567893046 Chr.15: 43340383-43340383 on Build GRCh38
rs770416182 Chr.15: 43340402-43340402 on Build GRCh38
rs768462225 Chr.15: 43340379-43340379 on Build GRCh38
rs749129363 Chr.15: 43340249-43340261 on Build GRCh38
rs751583544 Chr.15: 43340247-43340247 on Build GRCh38
rs778689854 Chr.15: 43340280-43340281 on Build GRCh38
rs553026787 Chr.15: 43340211-43340211 on Build GRCh38
rs572800903 Chr.15: 43340213-43340213 on Build GRCh38
rs777647413 Chr.15: 43340220-43340220 on Build GRCh38
rs545197260 Chr.15: 43340233-43340233 on Build GRCh38
rs752608772 Chr.15: 43340393-43340393 on Build GRCh38
rs369347554 Chr.15: 43340280-43340280 on Build GRCh38
rs201589904 Chr.15: 43340271-43340271 on Build GRCh38
rs777633836 Chr.15: 43340253-43340253 on Build GRCh38
rs780648393 Chr.15: 43340267-43340267 on Build GRCh38
rs372787096 Chr.15: 43340292-43340292 on Build GRCh38
rs755058385 Chr.15: 43340250-43340250 on Build GRCh38
rs374797273 Chr.15: 43340296-43340296 on Build GRCh38
rs759366978 Chr.15: 43340304-43340304 on Build GRCh38
rs956388893 Chr.15: 43340319-43340319 on Build GRCh38
rs745647075 Chr.15: 43340367-43340367 on Build GRCh38
rs751824116 Chr.15: 43340325-43340325 on Build GRCh38
rs931226459 Chr.15: 43340390-43340390 on Build GRCh38
rs868067928 Chr.15: 43340352-43340352 on Build GRCh38
rs757208601 Chr.15: 43340278-43340278 on Build GRCh38
rs989204889 Chr.15: 43340251-43340251 on Build GRCh38
rs755005330 Chr.15: 43340327-43340327 on Build GRCh38
rs369006563 Chr.15: 43340308-43340308 on Build GRCh38
rs370966532 Chr.15: 43340378-43340378 on Build GRCh38
rs968961535 Chr.15: 43340279-43340279 on Build GRCh38

View Full Product Details