Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 31709200-31709473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMN1
Entrez Gene ID
196394
Gene Name
antagonist of mitotic exit network 1 homolog
Gene Aliases
-
UniGene
Hs.585084
Gene Chromosome Location
Chr.12: 31671137-31729178 on Build GRCh38
Transcripts
NM_001113402.1 NM_001278411.1 NM_001278412.1 NR_103522.1 NR_103523.1 XM_011520604.2 XM_017018963.1 XM_017018964.1 XM_017018965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3460174 c.39-1G>T p.? Chr.12: 31709426-31709426 on Build GRCh38
375188 c.159A>C p.S53S Chr.12: 31709305-31709305 on Build GRCh38
3811878 c.75T>A p.Y25* Chr.12: 31709389-31709389 on Build GRCh38
5562243 c.102C>T p.P34P Chr.12: 31709362-31709362 on Build GRCh38
2205660 c.64A>C p.I22L Chr.12: 31709400-31709400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs935301729 Chr.12: 31709402-31709402 on Build GRCh38
rs749400512 Chr.12: 31709319-31709319 on Build GRCh38
rs757797225 Chr.12: 31709244-31709244 on Build GRCh38
rs576754326 Chr.12: 31709445-31709445 on Build GRCh38
rs771911909 Chr.12: 31709328-31709328 on Build GRCh38
rs771195352 Chr.12: 31709418-31709418 on Build GRCh38
rs745993535 Chr.12: 31709265-31709265 on Build GRCh38
rs772953536 Chr.12: 31709330-31709330 on Build GRCh38
rs768192500 Chr.12: 31709444-31709444 on Build GRCh38
rs953800205 Chr.12: 31709293-31709293 on Build GRCh38
rs776540263 Chr.12: 31709346-31709346 on Build GRCh38
rs762467514 Chr.12: 31709396-31709396 on Build GRCh38
rs556657531 Chr.12: 31709370-31709370 on Build GRCh38
rs913745434 Chr.12: 31709347-31709347 on Build GRCh38
rs781754450 Chr.12: 31709264-31709264 on Build GRCh38
rs774755294 Chr.12: 31709453-31709454 on Build GRCh38
rs764548854 Chr.12: 31709414-31709414 on Build GRCh38
rs371807376 Chr.12: 31709276-31709276 on Build GRCh38
rs756222294 Chr.12: 31709453-31709453 on Build GRCh38
rs773053288 Chr.12: 31709256-31709277 on Build GRCh38
rs374840560 Chr.12: 31709332-31709332 on Build GRCh38
rs770907515 Chr.12: 31709344-31709344 on Build GRCh38
rs762243265 Chr.12: 31709441-31709441 on Build GRCh38
rs775224507 Chr.12: 31709392-31709392 on Build GRCh38
rs925304425 Chr.12: 31709397-31709397 on Build GRCh38
rs988117476 Chr.12: 31709408-31709408 on Build GRCh38
rs762716616 Chr.12: 31709382-31709383 on Build GRCh38
rs979674807 Chr.12: 31709387-31709387 on Build GRCh38
rs766688534 Chr.12: 31709449-31709449 on Build GRCh38
rs764588493 Chr.12: 31709379-31709379 on Build GRCh38
rs756134843 Chr.12: 31709275-31709275 on Build GRCh38

View Full Product Details