Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 28878972-28879245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2A1
Entrez Gene ID
487
Gene Name
ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 1
Gene Aliases
ATP2A, SERCA1
UniGene
Hs.657344
Gene Chromosome Location
Chr.16: 28878488-28904509 on Build GRCh38
Transcripts
NM_004320.4 NM_173201.3 NR_046287.1 NR_046288.1 NR_046289.1 NR_046290.1

Gene Symbol
ATP2A1-AS1
Entrez Gene ID
100289092
Gene Name
ATP2A1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.666631
Gene Chromosome Location
Chr.16: 28878957-28879921 on Build GRCh38
Transcripts
NM_004320.4 NM_173201.3 NR_046287.1 NR_046288.1 NR_046289.1 NR_046290.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4652708 c.129T>C p.A43A Chr.16: 28879109-28879109 on Build GRCh38
7356952 c.130G>A p.E44K Chr.16: 28879110-28879110 on Build GRCh38
3110318 c.130G>A p.E44K Chr.16: 28879110-28879110 on Build GRCh38
4919420 c.119-2A>G p.? Chr.16: 28879097-28879097 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753059788 Chr.16: 28879153-28879153 on Build GRCh38
rs150403167 Chr.16: 28879108-28879108 on Build GRCh38
rs572322724 Chr.16: 28879170-28879170 on Build GRCh38
rs201535825 Chr.16: 28879097-28879097 on Build GRCh38
rs751153897 Chr.16: 28879068-28879068 on Build GRCh38
rs374059415 Chr.16: 28879117-28879117 on Build GRCh38
rs562933275 Chr.16: 28879185-28879185 on Build GRCh38
rs199741284 Chr.16: 28879150-28879150 on Build GRCh38
rs773792059 Chr.16: 28879057-28879057 on Build GRCh38
rs1031594037 Chr.16: 28879180-28879180 on Build GRCh38
rs766793145 Chr.16: 28879027-28879028 on Build GRCh38
rs565341699 Chr.16: 28879041-28879041 on Build GRCh38
rs767078815 Chr.16: 28879139-28879140 on Build GRCh38
rs201116614 Chr.16: 28879056-28879056 on Build GRCh38
rs755600693 Chr.16: 28879155-28879155 on Build GRCh38
rs751641213 Chr.16: 28879144-28879144 on Build GRCh38
rs775186108 Chr.16: 28879096-28879096 on Build GRCh38
rs748814656 Chr.16: 28879046-28879046 on Build GRCh38
rs182379583 Chr.16: 28879166-28879166 on Build GRCh38
rs778421757 Chr.16: 28879165-28879165 on Build GRCh38
rs992194665 Chr.16: 28879190-28879190 on Build GRCh38
rs770518938 Chr.16: 28879162-28879162 on Build GRCh38
rs543046369 Chr.16: 28879174-28879174 on Build GRCh38
rs769423552 Chr.16: 28879201-28879201 on Build GRCh38
rs1021904906 Chr.16: 28879084-28879084 on Build GRCh38
rs367969170 Chr.16: 28879139-28879139 on Build GRCh38
rs80127885 Chr.16: 28879222-28879222 on Build GRCh38
rs113714958 Chr.16: 28879178-28879178 on Build GRCh38
rs1028035561 Chr.16: 28879103-28879103 on Build GRCh38
rs576372299 Chr.16: 28879199-28879199 on Build GRCh38
rs554670861 Chr.16: 28879160-28879160 on Build GRCh38
rs759488565 Chr.16: 28879061-28879061 on Build GRCh38
rs781433311 Chr.16: 28879028-28879028 on Build GRCh38
rs748507232 Chr.16: 28879040-28879040 on Build GRCh38
rs769373031 Chr.16: 28879045-28879045 on Build GRCh38
rs999207735 Chr.16: 28879009-28879009 on Build GRCh38
rs767398402 Chr.16: 28879094-28879094 on Build GRCh38
rs745534425 Chr.16: 28879006-28879006 on Build GRCh38
rs750655997 Chr.16: 28879109-28879109 on Build GRCh38
rs752210009 Chr.16: 28879154-28879155 on Build GRCh38
rs756494370 Chr.16: 28879159-28879159 on Build GRCh38
rs760761542 Chr.16: 28879079-28879081 on Build GRCh38
rs371321547 Chr.16: 28879145-28879145 on Build GRCh38
rs919417882 Chr.16: 28879184-28879184 on Build GRCh38
rs529691613 Chr.16: 28879076-28879076 on Build GRCh38
rs770431288 Chr.16: 28879054-28879054 on Build GRCh38

View Full Product Details