Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54871497-54871738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHCR24
Entrez Gene ID
1718
Gene Name
24-dehydrocholesterol reductase
Gene Aliases
DCE, Nbla03646, SELADIN1, seladin-1
UniGene
Hs.498727
Gene Chromosome Location
Chr.1: 54849627-54887248 on Build GRCh38
Transcripts
NM_014762.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5847016 c.640G>A p.V214I Chr.1: 54871586-54871586 on Build GRCh38
3741321 c.721C>T p.R241C Chr.1: 54871505-54871505 on Build GRCh38
4755620 c.692T>C p.I231T Chr.1: 54871534-54871534 on Build GRCh38
136136 c.616G>A p.E206K Chr.1: 54871610-54871610 on Build GRCh38
6876513 c.614C>T p.S205F Chr.1: 54871612-54871612 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759168678 Chr.1: 54871561-54871561 on Build GRCh38
rs756361953 Chr.1: 54871630-54871630 on Build GRCh38
rs755078643 Chr.1: 54871644-54871644 on Build GRCh38
rs145971053 Chr.1: 54871515-54871515 on Build GRCh38
rs772464402 Chr.1: 54871548-54871548 on Build GRCh38
rs114852984 Chr.1: 54871678-54871678 on Build GRCh38
rs779358484 Chr.1: 54871647-54871647 on Build GRCh38
rs2303535 Chr.1: 54871707-54871707 on Build GRCh38
rs764798976 Chr.1: 54871566-54871566 on Build GRCh38
rs748700105 Chr.1: 54871648-54871648 on Build GRCh38
rs1019107299 Chr.1: 54871530-54871530 on Build GRCh38
rs180903448 Chr.1: 54871599-54871599 on Build GRCh38
rs376885844 Chr.1: 54871538-54871538 on Build GRCh38
rs201280799 Chr.1: 54871547-54871547 on Build GRCh38
rs143779194 Chr.1: 54871587-54871587 on Build GRCh38
rs370233954 Chr.1: 54871643-54871643 on Build GRCh38
rs750647923 Chr.1: 54871628-54871628 on Build GRCh38
rs147213053 Chr.1: 54871611-54871611 on Build GRCh38
rs545948766 Chr.1: 54871680-54871680 on Build GRCh38
rs375614612 Chr.1: 54871662-54871662 on Build GRCh38
rs902454280 Chr.1: 54871680-54871680 on Build GRCh38
rs2303534 Chr.1: 54871642-54871642 on Build GRCh38
rs528459787 Chr.1: 54871706-54871706 on Build GRCh38
rs759137429 Chr.1: 54871541-54871541 on Build GRCh38
rs150688144 Chr.1: 54871610-54871610 on Build GRCh38
rs139813073 Chr.1: 54871537-54871537 on Build GRCh38
rs1010206690 Chr.1: 54871586-54871586 on Build GRCh38
rs756200011 Chr.1: 54871522-54871522 on Build GRCh38
rs772411201 Chr.1: 54871653-54871653 on Build GRCh38
rs776322733 Chr.1: 54871557-54871557 on Build GRCh38
rs762400676 Chr.1: 54871598-54871598 on Build GRCh38
rs762566563 Chr.1: 54871591-54871591 on Build GRCh38
rs919273988 Chr.1: 54871699-54871699 on Build GRCh38
rs780333161 Chr.1: 54871637-54871637 on Build GRCh38

View Full Product Details