Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 110805200-110805471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOXL
Entrez Gene ID
55289
Gene Name
acyl-CoA oxidase-like
Gene Aliases
-
UniGene
Hs.253320
Gene Chromosome Location
Chr.2: 110732539-111118571 on Build GRCh38
Transcripts
NM_001142807.1 XM_011511404.2 XM_011511405.2 XM_011511406.2 XM_011511407.2 XM_011511409.1 XM_011511411.2 XM_011511413.1 XM_011511414.2 XM_011511415.2 XM_011511416.2 XM_011511419.2 XM_011511420.2 XM_011511421.2 XM_011511422.1 XM_011511423.1 XM_011511427.2 XM_011511428.2 XM_011511429.2 XM_011511432.2 XM_017004431.1 XM_017004432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6840577 c.624T>A p.F208L Chr.2: 110805266-110805266 on Build GRCh38
3971510 c.737C>T p.A246V Chr.2: 110805379-110805379 on Build GRCh38
6592411 c.657G>A p.S219S Chr.2: 110805299-110805299 on Build GRCh38
6840579 c.624T>A p.F208L Chr.2: 110805266-110805266 on Build GRCh38
6592402 c.623T>C p.F208S Chr.2: 110805265-110805265 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760567246 Chr.2: 110805256-110805256 on Build GRCh38
rs752076886 Chr.2: 110805280-110805280 on Build GRCh38
rs770582252 Chr.2: 110805224-110805224 on Build GRCh38
rs139248421 Chr.2: 110805299-110805299 on Build GRCh38
rs750745063 Chr.2: 110805274-110805274 on Build GRCh38
rs768494006 Chr.2: 110805315-110805315 on Build GRCh38
rs779843583 Chr.2: 110805396-110805396 on Build GRCh38
rs77331476 Chr.2: 110805298-110805298 on Build GRCh38
rs773459124 Chr.2: 110805415-110805415 on Build GRCh38
rs778611641 Chr.2: 110805225-110805225 on Build GRCh38
rs145955915 Chr.2: 110805324-110805324 on Build GRCh38
rs200684044 Chr.2: 110805398-110805398 on Build GRCh38
rs201243594 Chr.2: 110805339-110805339 on Build GRCh38
rs766727093 Chr.2: 110805342-110805342 on Build GRCh38
rs775472248 Chr.2: 110805246-110805246 on Build GRCh38
rs764764010 Chr.2: 110805361-110805361 on Build GRCh38
rs753349384 Chr.2: 110805290-110805290 on Build GRCh38
rs368626810 Chr.2: 110805276-110805276 on Build GRCh38
rs547761662 Chr.2: 110805379-110805379 on Build GRCh38
rs927751032 Chr.2: 110805375-110805375 on Build GRCh38
rs773547035 Chr.2: 110805328-110805328 on Build GRCh38
rs769963732 Chr.2: 110805408-110805408 on Build GRCh38
rs950515204 Chr.2: 110805358-110805358 on Build GRCh38
rs766703849 Chr.2: 110805278-110805278 on Build GRCh38
rs754872667 Chr.2: 110805400-110805400 on Build GRCh38
rs149961196 Chr.2: 110805355-110805355 on Build GRCh38
rs1017308709 Chr.2: 110805407-110805407 on Build GRCh38
rs778523879 Chr.2: 110805295-110805295 on Build GRCh38
rs201845957 Chr.2: 110805404-110805404 on Build GRCh38
rs777277902 Chr.2: 110805289-110805289 on Build GRCh38
rs749382866 Chr.2: 110805405-110805405 on Build GRCh38
rs771817264 Chr.2: 110805244-110805244 on Build GRCh38
rs746913084 Chr.2: 110805303-110805303 on Build GRCh38
rs531804513 Chr.2: 110805316-110805316 on Build GRCh38
rs748238722 Chr.2: 110805321-110805321 on Build GRCh38
rs771061182 Chr.2: 110805427-110805427 on Build GRCh38
rs776875130 Chr.2: 110805272-110805272 on Build GRCh38
rs755472496 Chr.2: 110805286-110805286 on Build GRCh38
rs201481495 Chr.2: 110805406-110805406 on Build GRCh38
rs745658750 Chr.2: 110805226-110805226 on Build GRCh38
rs768738862 Chr.2: 110805264-110805264 on Build GRCh38
rs756825931 Chr.2: 110805294-110805294 on Build GRCh38
rs73956467 Chr.2: 110805445-110805445 on Build GRCh38
rs756733784 Chr.2: 110805359-110805359 on Build GRCh38
rs753260925 Chr.2: 110805356-110805356 on Build GRCh38
rs748902034 Chr.2: 110805223-110805223 on Build GRCh38
rs774603432 Chr.2: 110805343-110805343 on Build GRCh38
rs527716900 Chr.2: 110805366-110805366 on Build GRCh38
rs777189289 Chr.2: 110805221-110805221 on Build GRCh38
rs760020348 Chr.2: 110805354-110805354 on Build GRCh38
rs1021658036 Chr.2: 110805261-110805261 on Build GRCh38
rs745564973 Chr.2: 110805297-110805297 on Build GRCh38
rs925965590 Chr.2: 110805247-110805247 on Build GRCh38
rs530334434 Chr.2: 110805424-110805424 on Build GRCh38
rs1011601189 Chr.2: 110805222-110805222 on Build GRCh38
rs371550981 Chr.2: 110805443-110805443 on Build GRCh38

View Full Product Details