Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12135575-12135800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF20
Entrez Gene ID
7568
Gene Name
zinc finger protein 20
Gene Aliases
KOX13
UniGene
Hs.512823
Gene Chromosome Location
Chr.19: 12131350-12140407 on Build GRCh38
Transcripts
NM_001203250.1 NM_021143.3 NR_037802.1

Gene Symbol
ZNF625-ZNF20
Entrez Gene ID
100529855
Gene Name
ZNF625-ZNF20 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.512823
Gene Chromosome Location
Chr.19: 12131350-12156731 on Build GRCh38
Transcripts
NM_001203250.1 NM_021143.3 NR_037802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6264915 c.139+6A>G p.? Chr.19: 12135763-12135763 on Build GRCh38
3821971 c.261C>G p.F87L Chr.19: 12135788-12135788 on Build GRCh38
1390530 c.275C>A p.S92Y Chr.19: 12135774-12135774 on Build GRCh38
3821973 c.249G>A p.M83I Chr.19: 12135800-12135800 on Build GRCh38
3821972 c.120C>G p.F40L Chr.19: 12135788-12135788 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs940260777 Chr.19: 12135717-12135717 on Build GRCh38
rs200963867 Chr.19: 12135724-12135724 on Build GRCh38
rs758632550 Chr.19: 12135729-12135729 on Build GRCh38
rs1048550273 Chr.19: 12135697-12135697 on Build GRCh38
rs777650155 Chr.19: 12135715-12135715 on Build GRCh38
rs1009143656 Chr.19: 12135739-12135739 on Build GRCh38
rs926040676 Chr.19: 12135614-12135614 on Build GRCh38
rs754444888 Chr.19: 12135722-12135722 on Build GRCh38
rs748154061 Chr.19: 12135772-12135772 on Build GRCh38
rs764610532 Chr.19: 12135604-12135604 on Build GRCh38
rs538981794 Chr.19: 12135750-12135750 on Build GRCh38
rs578178189 Chr.19: 12135727-12135727 on Build GRCh38
rs554485146 Chr.19: 12135765-12135765 on Build GRCh38
rs755921499 Chr.19: 12135731-12135732 on Build GRCh38
rs372005323 Chr.19: 12135746-12135746 on Build GRCh38
rs575832150 Chr.19: 12135620-12135620 on Build GRCh38
rs780031016 Chr.19: 12135756-12135757 on Build GRCh38
rs148045501 Chr.19: 12135655-12135655 on Build GRCh38
rs932791240 Chr.19: 12135658-12135658 on Build GRCh38
rs538397923 Chr.19: 12135683-12135683 on Build GRCh38
rs755064804 Chr.19: 12135756-12135756 on Build GRCh38
rs750656868 Chr.19: 12135728-12135728 on Build GRCh38

View Full Product Details