Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184188092-184188321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCF3
Entrez Gene ID
55324
Gene Name
ATP binding cassette subfamily F member 3
Gene Aliases
EST201864
UniGene
Hs.361323
Gene Chromosome Location
Chr.3: 184186075-184194012 on Build GRCh38
Transcripts
NM_018358.2 XM_011512959.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4115676 c.686G>A p.R229Q Chr.3: 184188257-184188257 on Build GRCh38
5445407 c.661A>G p.K221E Chr.3: 184188232-184188232 on Build GRCh38
4115675 c.680G>C p.S227T Chr.3: 184188251-184188251 on Build GRCh38
6545086 c.715G>T p.V239F Chr.3: 184188286-184188286 on Build GRCh38
7168377 c.739G>A p.D247N Chr.3: 184188310-184188310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781220084 Chr.3: 184188180-184188180 on Build GRCh38
rs778305777 Chr.3: 184188136-184188136 on Build GRCh38
rs566108440 Chr.3: 184188203-184188203 on Build GRCh38
rs368028401 Chr.3: 184188225-184188225 on Build GRCh38
rs371929198 Chr.3: 184188286-184188286 on Build GRCh38
rs776826785 Chr.3: 184188274-184188274 on Build GRCh38
rs760962119 Chr.3: 184188193-184188193 on Build GRCh38
rs746799914 Chr.3: 184188160-184188160 on Build GRCh38
rs530224305 Chr.3: 184188285-184188285 on Build GRCh38
rs1044314437 Chr.3: 184188271-184188271 on Build GRCh38
rs759226975 Chr.3: 184188147-184188147 on Build GRCh38
rs757952970 Chr.3: 184188158-184188158 on Build GRCh38
rs748016722 Chr.3: 184188246-184188246 on Build GRCh38
rs772268623 Chr.3: 184188185-184188185 on Build GRCh38
rs747252180 Chr.3: 184188257-184188257 on Build GRCh38
rs756732064 Chr.3: 184188133-184188133 on Build GRCh38
rs771694452 Chr.3: 184188204-184188204 on Build GRCh38
rs775755658 Chr.3: 184188189-184188189 on Build GRCh38
rs764453976 Chr.3: 184188201-184188201 on Build GRCh38
rs199665518 Chr.3: 184188184-184188184 on Build GRCh38
rs1040316094 Chr.3: 184188278-184188278 on Build GRCh38
rs867122369 Chr.3: 184188169-184188169 on Build GRCh38
rs754827507 Chr.3: 184188174-184188174 on Build GRCh38
rs777071943 Chr.3: 184188202-184188202 on Build GRCh38
rs779914897 Chr.3: 184188159-184188159 on Build GRCh38
rs766012470 Chr.3: 184188298-184188298 on Build GRCh38
rs374036545 Chr.3: 184188207-184188207 on Build GRCh38
rs981749323 Chr.3: 184188117-184188117 on Build GRCh38
rs772142140 Chr.3: 184188252-184188252 on Build GRCh38
rs781166608 Chr.3: 184188242-184188242 on Build GRCh38
rs755983326 Chr.3: 184188247-184188247 on Build GRCh38
rs768965312 Chr.3: 184188262-184188262 on Build GRCh38
rs972283870 Chr.3: 184188183-184188183 on Build GRCh38
rs565561529 Chr.3: 184188134-184188134 on Build GRCh38
rs755553236 Chr.3: 184188135-184188135 on Build GRCh38
rs754345529 Chr.3: 184188131-184188131 on Build GRCh38
rs775630869 Chr.3: 184188256-184188256 on Build GRCh38
rs748060492 Chr.3: 184188181-184188181 on Build GRCh38
rs924023051 Chr.3: 184188275-184188275 on Build GRCh38
rs532627156 Chr.3: 184188182-184188182 on Build GRCh38
rs201744647 Chr.3: 184188175-184188175 on Build GRCh38
rs751272921 Chr.3: 184188234-184188234 on Build GRCh38
rs757996688 Chr.3: 184188210-184188210 on Build GRCh38
rs766065416 Chr.3: 184188224-184188224 on Build GRCh38
rs777824805 Chr.3: 184188249-184188249 on Build GRCh38
rs761095198 Chr.3: 184188120-184188120 on Build GRCh38
rs754774008 Chr.3: 184188237-184188237 on Build GRCh38
rs916857446 Chr.3: 184188190-184188190 on Build GRCh38
rs762316248 Chr.3: 184188294-184188294 on Build GRCh38

View Full Product Details