Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 148761629-148761862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPC2
Entrez Gene ID
26122
Gene Name
enhancer of polycomb homolog 2
Gene Aliases
EPC-LIKE
UniGene
Hs.23270
Gene Chromosome Location
Chr.2: 148644780-148788491 on Build GRCh38
Transcripts
NM_015630.3 XM_011510941.2 XM_011510943.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1305688 c.728G>C p.R243T Chr.2: 148761843-148761843 on Build GRCh38
5343778 c.700A>T p.M234L Chr.2: 148761815-148761815 on Build GRCh38
6654814 c.732C>T p.A244A Chr.2: 148761847-148761847 on Build GRCh38
716466 c.679G>C p.D227H Chr.2: 148761794-148761794 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760285441 Chr.2: 148761805-148761805 on Build GRCh38
rs557068296 Chr.2: 148761820-148761820 on Build GRCh38
rs757711511 Chr.2: 148761826-148761826 on Build GRCh38
rs1038538457 Chr.2: 148761811-148761811 on Build GRCh38
rs766960465 Chr.2: 148761733-148761733 on Build GRCh38
rs576361867 Chr.2: 148761664-148761664 on Build GRCh38
rs566315586 Chr.2: 148761728-148761728 on Build GRCh38
rs760067887 Chr.2: 148761795-148761795 on Build GRCh38
rs941001025 Chr.2: 148761760-148761760 on Build GRCh38
rs763837339 Chr.2: 148761808-148761808 on Build GRCh38
rs759479351 Chr.2: 148761732-148761732 on Build GRCh38
rs149635989 Chr.2: 148761660-148761660 on Build GRCh38
rs16829242 Chr.2: 148761729-148761729 on Build GRCh38
rs934219086 Chr.2: 148761706-148761706 on Build GRCh38
rs185996865 Chr.2: 148761715-148761715 on Build GRCh38
rs561637463 Chr.2: 148761772-148761772 on Build GRCh38
rs904916042 Chr.2: 148761813-148761813 on Build GRCh38
rs374375613 Chr.2: 148761744-148761745 on Build GRCh38
rs772432316 Chr.2: 148761831-148761831 on Build GRCh38
rs752230178 Chr.2: 148761801-148761801 on Build GRCh38
rs771352384 Chr.2: 148761705-148761705 on Build GRCh38

View Full Product Details