Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54148055-54148282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT3
Entrez Gene ID
4849
Gene Name
CCR4-NOT transcription complex subunit 3
Gene Aliases
LENG2, NOT3, NOT3H
UniGene
Hs.343571
Gene Chromosome Location
Chr.19: 54137689-54155708 on Build GRCh38
Transcripts
NM_014516.3 XM_005278279.1 XM_005278280.2 XM_005278281.1 XM_005278282.2 XM_011526992.1 XM_011526993.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5856663 c.974C>T p.S325F Chr.19: 54148227-54148227 on Build GRCh38
1612771 c.393G>T p.Q131H Chr.19: 54148189-54148189 on Build GRCh38
5730368 c.443C>T p.P148L Chr.19: 54148239-54148239 on Build GRCh38
1612769 c.380T>C p.V127A Chr.19: 54148176-54148176 on Build GRCh38
1564235 c.942T>G p.P314P Chr.19: 54148195-54148195 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776345967 Chr.19: 54148256-54148256 on Build GRCh38
rs368877712 Chr.19: 54148142-54148142 on Build GRCh38
rs777481101 Chr.19: 54148124-54148127 on Build GRCh38
rs943465333 Chr.19: 54148100-54148100 on Build GRCh38
rs758530739 Chr.19: 54148198-54148198 on Build GRCh38
rs149108037 Chr.19: 54148163-54148163 on Build GRCh38
rs766226888 Chr.19: 54148182-54148182 on Build GRCh38
rs138658963 Chr.19: 54148193-54148193 on Build GRCh38
rs372481652 Chr.19: 54148236-54148236 on Build GRCh38
rs369505503 Chr.19: 54148122-54148122 on Build GRCh38
rs752679696 Chr.19: 54148194-54148194 on Build GRCh38
rs971897825 Chr.19: 54148234-54148234 on Build GRCh38
rs772901111 Chr.19: 54148162-54148162 on Build GRCh38
rs587750007 Chr.19: 54148213-54148213 on Build GRCh38
rs376542458 Chr.19: 54148115-54148115 on Build GRCh38
rs146219248 Chr.19: 54148192-54148192 on Build GRCh38
rs587607054 Chr.19: 54148080-54148080 on Build GRCh38
rs1015932395 Chr.19: 54148227-54148227 on Build GRCh38
rs587744329 Chr.19: 54148190-54148190 on Build GRCh38
rs868121459 Chr.19: 54148181-54148181 on Build GRCh38
rs373233998 Chr.19: 54148143-54148143 on Build GRCh38
rs776083392 Chr.19: 54148225-54148225 on Build GRCh38
rs190206269 Chr.19: 54148212-54148212 on Build GRCh38
rs747278028 Chr.19: 54148208-54148208 on Build GRCh38
rs587645855 Chr.19: 54148229-54148229 on Build GRCh38
rs904696747 Chr.19: 54148133-54148133 on Build GRCh38
rs747333221 Chr.19: 54148113-54148113 on Build GRCh38
rs760377575 Chr.19: 54148237-54148237 on Build GRCh38
rs778122851 Chr.19: 54148207-54148207 on Build GRCh38
rs766035831 Chr.19: 54148255-54148255 on Build GRCh38
rs954677471 Chr.19: 54148241-54148241 on Build GRCh38
rs963556829 Chr.19: 54148233-54148233 on Build GRCh38
rs370792103 Chr.19: 54148228-54148228 on Build GRCh38
rs767027855 Chr.19: 54148144-54148144 on Build GRCh38
rs770344355 Chr.19: 54148137-54148137 on Build GRCh38
rs746448157 Chr.19: 54148125-54148125 on Build GRCh38
rs910541075 Chr.19: 54148088-54148088 on Build GRCh38
rs1043157433 Chr.19: 54148116-54148116 on Build GRCh38
rs1026547087 Chr.19: 54148235-54148235 on Build GRCh38

View Full Product Details