Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 143202025-143202286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP15
Entrez Gene ID
55843
Gene Name
Rho GTPase activating protein 15
Gene Aliases
BM046
UniGene
Hs.171011
Gene Chromosome Location
Chr.2: 143070973-143768352 on Build GRCh38
Transcripts
NM_018460.3 XM_011511479.2 XM_011511481.2 XM_011511482.2 XM_011511484.1 XM_017004499.1 XM_017004500.1 XM_017004501.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5777593 c.226G>C p.E76Q Chr.2: 143202194-143202194 on Build GRCh38
6409085 c.182G>T p.R61M Chr.2: 143202150-143202150 on Build GRCh38
5697066 c.177C>A p.H59Q Chr.2: 143202145-143202145 on Build GRCh38
6543105 c.231A>T p.Q77H Chr.2: 143202199-143202199 on Build GRCh38
5692000 c.191C>A p.S64* Chr.2: 143202159-143202159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188609203 Chr.2: 143202079-143202079 on Build GRCh38
rs142961157 Chr.2: 143202178-143202178 on Build GRCh38
rs868214615 Chr.2: 143202157-143202157 on Build GRCh38
rs751938042 Chr.2: 143202195-143202195 on Build GRCh38
rs753302914 Chr.2: 143202086-143202086 on Build GRCh38
rs979135492 Chr.2: 143202127-143202127 on Build GRCh38
rs750060707 Chr.2: 143202094-143202094 on Build GRCh38
rs761284438 Chr.2: 143202187-143202187 on Build GRCh38
rs377536889 Chr.2: 143202241-143202241 on Build GRCh38
rs369534497 Chr.2: 143202130-143202130 on Build GRCh38
rs930698761 Chr.2: 143202134-143202134 on Build GRCh38
rs758074426 Chr.2: 143202194-143202194 on Build GRCh38
rs149299444 Chr.2: 143202153-143202153 on Build GRCh38
rs555531347 Chr.2: 143202121-143202121 on Build GRCh38
rs781684394 Chr.2: 143202116-143202116 on Build GRCh38
rs974918272 Chr.2: 143202108-143202108 on Build GRCh38
rs755465667 Chr.2: 143202107-143202107 on Build GRCh38
rs747338116 Chr.2: 143202106-143202106 on Build GRCh38
rs749499312 Chr.2: 143202145-143202145 on Build GRCh38
rs781617647 Chr.2: 143202219-143202219 on Build GRCh38
rs778499726 Chr.2: 143202091-143202091 on Build GRCh38
rs748676895 Chr.2: 143202225-143202225 on Build GRCh38
rs148758472 Chr.2: 143202058-143202058 on Build GRCh38
rs756721700 Chr.2: 143202090-143202090 on Build GRCh38
rs777970233 Chr.2: 143202245-143202245 on Build GRCh38
rs750112728 Chr.2: 143202190-143202190 on Build GRCh38
rs966953085 Chr.2: 143202065-143202065 on Build GRCh38
rs267598909 Chr.2: 143202138-143202138 on Build GRCh38
rs567006761 Chr.2: 143202078-143202078 on Build GRCh38
rs758807954 Chr.2: 143202101-143202101 on Build GRCh38
rs541207996 Chr.2: 143202160-143202160 on Build GRCh38
rs780362238 Chr.2: 143202105-143202105 on Build GRCh38
rs112415929 Chr.2: 143202073-143202073 on Build GRCh38
rs79440190 Chr.2: 143202216-143202216 on Build GRCh38
rs150534050 Chr.2: 143202111-143202111 on Build GRCh38
rs968781049 Chr.2: 143202124-143202124 on Build GRCh38
rs779149726 Chr.2: 143202163-143202163 on Build GRCh38
rs764044960 Chr.2: 143202084-143202084 on Build GRCh38
rs77631323 Chr.2: 143202165-143202165 on Build GRCh38

View Full Product Details