Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 227654154-227654411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF678
Entrez Gene ID
339500
Gene Name
zinc finger protein 678
Gene Aliases
-
UniGene
Hs.30323
Gene Chromosome Location
Chr.1: 227563519-227677443 on Build GRCh38
Transcripts
NM_178549.3 NR_033184.1 NR_102302.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4396908 c.266C>T p.S89F Chr.1: 227654351-227654351 on Build GRCh38
4517281 c.144_145CC>TT p.Q49* Chr.1: 227654394-227654395 on Build GRCh38
904887 c.143G>A p.C48Y Chr.1: 227654393-227654393 on Build GRCh38
6124829 c.156A>T p.T52T Chr.1: 227654406-227654406 on Build GRCh38
339730 c.126G>T p.Q42H Chr.1: 227654376-227654376 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550703386 Chr.1: 227654253-227654253 on Build GRCh38
rs933664147 Chr.1: 227654219-227654219 on Build GRCh38
rs577667494 Chr.1: 227654204-227654204 on Build GRCh38
rs529335857 Chr.1: 227654233-227654233 on Build GRCh38
rs893130636 Chr.1: 227654364-227654364 on Build GRCh38
rs948917791 Chr.1: 227654349-227654349 on Build GRCh38
rs771878571 Chr.1: 227654252-227654252 on Build GRCh38
rs369768128 Chr.1: 227654331-227654331 on Build GRCh38
rs770104033 Chr.1: 227654353-227654353 on Build GRCh38
rs779884178 Chr.1: 227654322-227654322 on Build GRCh38
rs747539296 Chr.1: 227654338-227654338 on Build GRCh38
rs775710861 Chr.1: 227654355-227654355 on Build GRCh38
rs533862737 Chr.1: 227654361-227654361 on Build GRCh38
rs771454019 Chr.1: 227654351-227654351 on Build GRCh38
rs551579013 Chr.1: 227654283-227654283 on Build GRCh38
rs776906880 Chr.1: 227654352-227654352 on Build GRCh38
rs955006570 Chr.1: 227654227-227654227 on Build GRCh38
rs566705673 Chr.1: 227654333-227654333 on Build GRCh38
rs778422768 Chr.1: 227654336-227654336 on Build GRCh38
rs774478709 Chr.1: 227654369-227654369 on Build GRCh38
rs773320654 Chr.1: 227654248-227654248 on Build GRCh38
rs755873043 Chr.1: 227654309-227654309 on Build GRCh38
rs562121771 Chr.1: 227654344-227654344 on Build GRCh38
rs761707208 Chr.1: 227654382-227654382 on Build GRCh38
rs375893393 Chr.1: 227654297-227654297 on Build GRCh38
rs934919893 Chr.1: 227654356-227654356 on Build GRCh38
rs754463109 Chr.1: 227654335-227654335 on Build GRCh38
rs190428363 Chr.1: 227654261-227654261 on Build GRCh38
rs763102777 Chr.1: 227654358-227654358 on Build GRCh38
rs537732152 Chr.1: 227654254-227654259 on Build GRCh38
rs72748099 Chr.1: 227654280-227654280 on Build GRCh38
rs767275379 Chr.1: 227654307-227654307 on Build GRCh38
rs982446354 Chr.1: 227654228-227654228 on Build GRCh38
rs111255164 Chr.1: 227654191-227654191 on Build GRCh38

View Full Product Details