Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 60890874-60891137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC8
Entrez Gene ID
1161
Gene Name
ERCC excision repair 8, CSA ubiquitin ligase complex subunit
Gene Aliases
CKN1, CSA, UVSS2
UniGene
Hs.435237
Gene Chromosome Location
Chr.5: 60873832-60945078 on Build GRCh38
Transcripts
NM_000082.3 NM_001007233.2 NM_001290285.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3345484 c.978G>C p.Q326H Chr.5: 60890952-60890952 on Build GRCh38
4820564 c.937G>A p.G313S Chr.5: 60890993-60890993 on Build GRCh38
6372001 c.901T>C p.C301R Chr.5: 60891029-60891029 on Build GRCh38
5403671 c.887A>G p.K296R Chr.5: 60891043-60891043 on Build GRCh38
5005207 c.1004A>G p.K335R Chr.5: 60890926-60890926 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753663016 Chr.5: 60890913-60890913 on Build GRCh38
rs377001963 Chr.5: 60890992-60890992 on Build GRCh38
rs780224304 Chr.5: 60891105-60891105 on Build GRCh38
rs770215298 Chr.5: 60891093-60891093 on Build GRCh38
rs1031250600 Chr.5: 60890921-60890921 on Build GRCh38
rs121434323 Chr.5: 60890964-60890964 on Build GRCh38
rs56186848 Chr.5: 60891009-60891009 on Build GRCh38
rs373203544 Chr.5: 60890990-60890990 on Build GRCh38
rs868640647 Chr.5: 60891045-60891045 on Build GRCh38
rs906678233 Chr.5: 60890909-60890909 on Build GRCh38
rs989442602 Chr.5: 60890939-60890939 on Build GRCh38
rs199805243 Chr.5: 60890997-60890997 on Build GRCh38
rs747530954 Chr.5: 60890945-60890945 on Build GRCh38
rs786205573 Chr.5: 60890914-60890914 on Build GRCh38
rs746217955 Chr.5: 60890962-60890962 on Build GRCh38
rs781699664 Chr.5: 60891073-60891073 on Build GRCh38
rs766565870 Chr.5: 60890978-60890978 on Build GRCh38
rs760761028 Chr.5: 60890988-60890988 on Build GRCh38
rs374386779 Chr.5: 60891031-60891031 on Build GRCh38
rs1051320962 Chr.5: 60891064-60891064 on Build GRCh38
rs751639405 Chr.5: 60891047-60891047 on Build GRCh38
rs767147543 Chr.5: 60890972-60890972 on Build GRCh38
rs183505264 Chr.5: 60891081-60891081 on Build GRCh38
rs933739444 Chr.5: 60891058-60891058 on Build GRCh38
rs369317106 Chr.5: 60890908-60890908 on Build GRCh38
rs748218049 Chr.5: 60890933-60890933 on Build GRCh38
rs377473708 Chr.5: 60891063-60891063 on Build GRCh38
rs150632283 Chr.5: 60890971-60890971 on Build GRCh38
rs766822890 Chr.5: 60890940-60890940 on Build GRCh38
rs201642761 Chr.5: 60891019-60891019 on Build GRCh38
rs925487680 Chr.5: 60890980-60890980 on Build GRCh38
rs76592390 Chr.5: 60891008-60891008 on Build GRCh38
rs764902694 Chr.5: 60890993-60890993 on Build GRCh38
rs758506477 Chr.5: 60890937-60890937 on Build GRCh38
rs1029101745 Chr.5: 60890957-60890957 on Build GRCh38
rs755213944 Chr.5: 60890915-60890915 on Build GRCh38
rs777838268 Chr.5: 60891022-60891022 on Build GRCh38
rs141845482 Chr.5: 60890918-60890918 on Build GRCh38
rs771365290 Chr.5: 60890960-60890960 on Build GRCh38
rs200733672 Chr.5: 60891002-60891002 on Build GRCh38
rs772779979 Chr.5: 60890973-60890973 on Build GRCh38
rs958033034 Chr.5: 60890929-60890929 on Build GRCh38
rs138173863 Chr.5: 60890907-60890907 on Build GRCh38

View Full Product Details