Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49889090-49889323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAIM2
Entrez Gene ID
23017
Gene Name
Fas apoptotic inhibitory molecule 2
Gene Aliases
LFG, LFG2, NGP35, NMP35, TMBIM2
UniGene
Hs.567424
Gene Chromosome Location
Chr.12: 49866896-49904275 on Build GRCh38
Transcripts
NM_012306.3 XM_005268730.3 XM_017019040.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1739900 c.655G>C p.D219H Chr.12: 49889199-49889199 on Build GRCh38
673564 c.686T>A p.V229E Chr.12: 49889168-49889168 on Build GRCh38
6072802 c.693C>T p.L231L Chr.12: 49889161-49889161 on Build GRCh38
4682313 c.654C>A p.F218L Chr.12: 49889200-49889200 on Build GRCh38
6553259 c.725C>A p.A242D Chr.12: 49889129-49889129 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141699858 Chr.12: 49889241-49889241 on Build GRCh38
rs1023685116 Chr.12: 49889270-49889270 on Build GRCh38
rs755012762 Chr.12: 49889146-49889146 on Build GRCh38
rs757416721 Chr.12: 49889179-49889179 on Build GRCh38
rs960736409 Chr.12: 49889210-49889210 on Build GRCh38
rs760203252 Chr.12: 49889131-49889131 on Build GRCh38
rs371670853 Chr.12: 49889236-49889236 on Build GRCh38
rs769887096 Chr.12: 49889192-49889192 on Build GRCh38
rs200244697 Chr.12: 49889194-49889194 on Build GRCh38
rs762236456 Chr.12: 49889220-49889220 on Build GRCh38
rs996596105 Chr.12: 49889239-49889239 on Build GRCh38
rs374845263 Chr.12: 49889198-49889198 on Build GRCh38
rs374323035 Chr.12: 49889213-49889213 on Build GRCh38
rs780505235 Chr.12: 49889201-49889201 on Build GRCh38
rs781381072 Chr.12: 49889180-49889180 on Build GRCh38
rs766216160 Chr.12: 49889135-49889135 on Build GRCh38
rs200316208 Chr.12: 49889191-49889191 on Build GRCh38
rs753548300 Chr.12: 49889143-49889143 on Build GRCh38
rs202170303 Chr.12: 49889178-49889178 on Build GRCh38
rs1016158598 Chr.12: 49889182-49889182 on Build GRCh38
rs771598269 Chr.12: 49889235-49889235 on Build GRCh38
rs749610879 Chr.12: 49889212-49889212 on Build GRCh38
rs35839552 Chr.12: 49889184-49889185 on Build GRCh38
rs141214854 Chr.12: 49889186-49889186 on Build GRCh38
rs1025582315 Chr.12: 49889200-49889200 on Build GRCh38
rs774851990 Chr.12: 49889215-49889215 on Build GRCh38
rs778983503 Chr.12: 49889169-49889169 on Build GRCh38

View Full Product Details