Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 28332284-28332509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AZI2
Entrez Gene ID
64343
Gene Name
5-azacytidine induced 2
Gene Aliases
AZ2, NAP1, TILP
UniGene
Hs.706676
Gene Chromosome Location
Chr.3: 28322353-28349129 on Build GRCh38
Transcripts
NM_001134432.1 NM_001134433.1 NM_001271650.1 NM_022461.4 XM_005265392.2 XM_011534009.2 XM_011534010.2 XM_011534011.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
730374 c.634C>G p.L212V Chr.3: 28332382-28332382 on Build GRCh38
4859426 c.634C>G p.L212V Chr.3: 28332382-28332382 on Build GRCh38
6329940 c.647+7C>T p.? Chr.3: 28332362-28332362 on Build GRCh38
6329941 c.647+7C>T p.? Chr.3: 28332362-28332362 on Build GRCh38
5460574 c.620G>C p.R207T Chr.3: 28332396-28332396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765251029 Chr.3: 28332343-28332343 on Build GRCh38
rs749811326 Chr.3: 28332422-28332422 on Build GRCh38
rs774575223 Chr.3: 28332427-28332427 on Build GRCh38
rs184488429 Chr.3: 28332360-28332360 on Build GRCh38
rs774554713 Chr.3: 28332324-28332324 on Build GRCh38
rs773597346 Chr.3: 28332413-28332413 on Build GRCh38
rs972974102 Chr.3: 28332374-28332374 on Build GRCh38
rs746519068 Chr.3: 28332377-28332377 on Build GRCh38
rs772334782 Chr.3: 28332361-28332361 on Build GRCh38
rs199908700 Chr.3: 28332474-28332474 on Build GRCh38
rs759902312 Chr.3: 28332335-28332335 on Build GRCh38
rs758331900 Chr.3: 28332359-28332359 on Build GRCh38
rs753695478 Chr.3: 28332337-28332337 on Build GRCh38
rs768276037 Chr.3: 28332397-28332397 on Build GRCh38
rs147974643 Chr.3: 28332425-28332425 on Build GRCh38
rs369977984 Chr.3: 28332472-28332472 on Build GRCh38
rs759627878 Chr.3: 28332434-28332434 on Build GRCh38
rs377211948 Chr.3: 28332394-28332394 on Build GRCh38
rs746983701 Chr.3: 28332326-28332326 on Build GRCh38
rs750367749 Chr.3: 28332352-28332352 on Build GRCh38
rs760918766 Chr.3: 28332467-28332467 on Build GRCh38
rs775814419 Chr.3: 28332462-28332462 on Build GRCh38
rs770018277 Chr.3: 28332410-28332410 on Build GRCh38
rs748570829 Chr.3: 28332396-28332396 on Build GRCh38
rs371749088 Chr.3: 28332338-28332338 on Build GRCh38
rs767871424 Chr.3: 28332441-28332441 on Build GRCh38
rs934233936 Chr.3: 28332417-28332417 on Build GRCh38
rs754883022 Chr.3: 28332448-28332449 on Build GRCh38
rs751470447 Chr.3: 28332447-28332449 on Build GRCh38
rs912546618 Chr.3: 28332332-28332332 on Build GRCh38

View Full Product Details