Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 99362188-99362457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNNM1
Entrez Gene ID
26507
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 1
Gene Aliases
ACDP1, CLP-1
UniGene
Hs.274579
Gene Chromosome Location
Chr.10: 99329099-99394330 on Build GRCh38
Transcripts
NM_020348.2 XM_005269692.3 XM_011539629.1 XM_011539630.1 XM_011539631.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
368492 c.896G>A p.R299H Chr.10: 99362359-99362359 on Build GRCh38
6631291 c.1927G>A p.V643M Chr.10: 99362295-99362295 on Build GRCh38
6128264 c.931C>T p.Q311* Chr.10: 99362394-99362394 on Build GRCh38
6128263 c.2026C>T p.Q676* Chr.10: 99362394-99362394 on Build GRCh38
4673126 c.872C>G p.A291G Chr.10: 99362335-99362335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779563342 Chr.10: 99362370-99362370 on Build GRCh38
rs753261982 Chr.10: 99362382-99362382 on Build GRCh38
rs754027026 Chr.10: 99362218-99362218 on Build GRCh38
rs138799879 Chr.10: 99362259-99362259 on Build GRCh38
rs372135038 Chr.10: 99362428-99362428 on Build GRCh38
rs760696046 Chr.10: 99362321-99362321 on Build GRCh38
rs772053824 Chr.10: 99362295-99362295 on Build GRCh38
rs758801855 Chr.10: 99362258-99362258 on Build GRCh38
rs928298452 Chr.10: 99362254-99362254 on Build GRCh38
rs747819215 Chr.10: 99362400-99362400 on Build GRCh38
rs765588515 Chr.10: 99362234-99362234 on Build GRCh38
rs906860804 Chr.10: 99362361-99362361 on Build GRCh38
rs776671800 Chr.10: 99362427-99362427 on Build GRCh38
rs765346829 Chr.10: 99362349-99362349 on Build GRCh38
rs754586698 Chr.10: 99362387-99362387 on Build GRCh38
rs375324631 Chr.10: 99362220-99362220 on Build GRCh38
rs142279958 Chr.10: 99362294-99362294 on Build GRCh38
rs771929003 Chr.10: 99362403-99362403 on Build GRCh38
rs867673472 Chr.10: 99362235-99362235 on Build GRCh38
rs144833574 Chr.10: 99362307-99362307 on Build GRCh38
rs746896028 Chr.10: 99362415-99362415 on Build GRCh38
rs747571350 Chr.10: 99362274-99362274 on Build GRCh38
rs367774976 Chr.10: 99362292-99362292 on Build GRCh38
rs537907747 Chr.10: 99362273-99362273 on Build GRCh38
rs117724968 Chr.10: 99362327-99362327 on Build GRCh38
rs766582539 Chr.10: 99362214-99362214 on Build GRCh38
rs371216334 Chr.10: 99362293-99362293 on Build GRCh38
rs369315734 Chr.10: 99362358-99362358 on Build GRCh38
rs777743526 Chr.10: 99362405-99362405 on Build GRCh38
rs199947342 Chr.10: 99362323-99362323 on Build GRCh38
rs778420366 Chr.10: 99362393-99362393 on Build GRCh38
rs759710493 Chr.10: 99362345-99362345 on Build GRCh38
rs201507104 Chr.10: 99362275-99362275 on Build GRCh38
rs1024244354 Chr.10: 99362433-99362433 on Build GRCh38
rs760474935 Chr.10: 99362245-99362251 on Build GRCh38
rs902528898 Chr.10: 99362364-99362364 on Build GRCh38
rs534085966 Chr.10: 99362420-99362420 on Build GRCh38
rs764454140 Chr.10: 99362359-99362359 on Build GRCh38
rs757790447 Chr.10: 99362365-99362365 on Build GRCh38
rs776597958 Chr.10: 99362339-99362339 on Build GRCh38
rs752982308 Chr.10: 99362257-99362257 on Build GRCh38
rs779431010 Chr.10: 99362279-99362279 on Build GRCh38
rs751993329 Chr.10: 99362362-99362362 on Build GRCh38
rs189765421 Chr.10: 99362209-99362209 on Build GRCh38
rs753029505 Chr.10: 99362351-99362351 on Build GRCh38

View Full Product Details