Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37373596-37373865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELFN2
Entrez Gene ID
114794
Gene Name
extracellular leucine rich repeat and fibronectin type III domain containing 2
Gene Aliases
LRRC62, PPP1R29
UniGene
Hs.344170
Gene Chromosome Location
Chr.22: 37340644-37427470 on Build GRCh38
Transcripts
NM_052906.4 NR_110512.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
333180 c.1778C>T p.P593L Chr.22: 37373757-37373757 on Build GRCh38
3668604 c.1717G>C p.E573Q Chr.22: 37373818-37373818 on Build GRCh38
4680754 c.1676C>T p.S559L Chr.22: 37373859-37373859 on Build GRCh38
1666715 c.1895C>T p.S632L Chr.22: 37373640-37373640 on Build GRCh38
4642641 c.1706G>A p.S569N Chr.22: 37373829-37373829 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766332484 Chr.22: 37373639-37373639 on Build GRCh38
rs148946952 Chr.22: 37373622-37373622 on Build GRCh38
rs760583940 Chr.22: 37373632-37373632 on Build GRCh38
rs747076955 Chr.22: 37373684-37373684 on Build GRCh38
rs754275848 Chr.22: 37373726-37373726 on Build GRCh38
rs144139706 Chr.22: 37373672-37373672 on Build GRCh38
rs755348427 Chr.22: 37373727-37373727 on Build GRCh38
rs554867479 Chr.22: 37373755-37373755 on Build GRCh38
rs199787830 Chr.22: 37373659-37373659 on Build GRCh38
rs577460497 Chr.22: 37373838-37373838 on Build GRCh38
rs544695745 Chr.22: 37373839-37373839 on Build GRCh38
rs758584905 Chr.22: 37373740-37373740 on Build GRCh38
rs550532926 Chr.22: 37373698-37373698 on Build GRCh38
rs777867053 Chr.22: 37373821-37373821 on Build GRCh38
rs747077460 Chr.22: 37373743-37373743 on Build GRCh38
rs113261780 Chr.22: 37373812-37373812 on Build GRCh38
rs573004348 Chr.22: 37373756-37373756 on Build GRCh38
rs749920264 Chr.22: 37373805-37373805 on Build GRCh38
rs146145172 Chr.22: 37373690-37373690 on Build GRCh38
rs757943128 Chr.22: 37373657-37373657 on Build GRCh38
rs778030367 Chr.22: 37373742-37373742 on Build GRCh38
rs765698464 Chr.22: 37373816-37373816 on Build GRCh38
rs755295450 Chr.22: 37373670-37373670 on Build GRCh38
rs138855230 Chr.22: 37373819-37373819 on Build GRCh38
rs962768922 Chr.22: 37373671-37373671 on Build GRCh38
rs1012891029 Chr.22: 37373681-37373681 on Build GRCh38
rs775190367 Chr.22: 37373764-37373764 on Build GRCh38
rs779305364 Chr.22: 37373731-37373731 on Build GRCh38
rs546148270 Chr.22: 37373646-37373646 on Build GRCh38
rs760100008 Chr.22: 37373807-37373807 on Build GRCh38
rs756621288 Chr.22: 37373663-37373663 on Build GRCh38
rs765016780 Chr.22: 37373647-37373647 on Build GRCh38
rs759220443 Chr.22: 37373769-37373771 on Build GRCh38
rs753099065 Chr.22: 37373739-37373739 on Build GRCh38
rs202143334 Chr.22: 37373776-37373776 on Build GRCh38
rs538345336 Chr.22: 37373675-37373675 on Build GRCh38
rs771037552 Chr.22: 37373689-37373689 on Build GRCh38
rs772322776 Chr.22: 37373677-37373677 on Build GRCh38
rs1050969469 Chr.22: 37373660-37373660 on Build GRCh38
rs61735214 Chr.22: 37373650-37373650 on Build GRCh38
rs756742507 Chr.22: 37373715-37373715 on Build GRCh38
rs752333780 Chr.22: 37373656-37373656 on Build GRCh38
rs753049260 Chr.22: 37373818-37373818 on Build GRCh38
rs763772323 Chr.22: 37373699-37373699 on Build GRCh38
rs762787861 Chr.22: 37373770-37373770 on Build GRCh38
rs1045689613 Chr.22: 37373626-37373626 on Build GRCh38
rs768216185 Chr.22: 37373773-37373773 on Build GRCh38
rs780517698 Chr.22: 37373666-37373666 on Build GRCh38
rs557704677 Chr.22: 37373692-37373692 on Build GRCh38
rs763572596 Chr.22: 37373658-37373658 on Build GRCh38
rs746669142 Chr.22: 37373623-37373623 on Build GRCh38
rs558982154 Chr.22: 37373777-37373777 on Build GRCh38
rs759413380 Chr.22: 37373693-37373693 on Build GRCh38
rs199881580 Chr.22: 37373746-37373746 on Build GRCh38
rs766901884 Chr.22: 37373718-37373718 on Build GRCh38
rs765069782 Chr.22: 37373697-37373697 on Build GRCh38
rs975026802 Chr.22: 37373762-37373762 on Build GRCh38
rs1026515777 Chr.22: 37373728-37373728 on Build GRCh38
rs201029178 Chr.22: 37373840-37373840 on Build GRCh38
rs142797056 Chr.22: 37373645-37373645 on Build GRCh38
rs749675532 Chr.22: 37373668-37373668 on Build GRCh38
rs569021339 Chr.22: 37373744-37373744 on Build GRCh38
rs142310849 Chr.22: 37373678-37373678 on Build GRCh38
rs781295181 Chr.22: 37373753-37373753 on Build GRCh38
rs367945618 Chr.22: 37373665-37373665 on Build GRCh38

View Full Product Details