Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 80442472-80442715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM46D
Entrez Gene ID
169966
Gene Name
family with sequence similarity 46 member D
Gene Aliases
CT1.26, CT112
UniGene
Hs.367959
Gene Chromosome Location
Chr.X: 80335504-80445311 on Build GRCh38
Transcripts
NM_001170574.1 NM_152630.4

SNP Detail

SNP ID Chromosome Location
rs747484824 Chr.X: 80442544-80442544 on Build GRCh38
rs79116942 Chr.X: 80442596-80442596 on Build GRCh38
rs772030179 Chr.X: 80442515-80442515 on Build GRCh38
rs755654386 Chr.X: 80442662-80442662 on Build GRCh38
rs749977497 Chr.X: 80442650-80442650 on Build GRCh38
rs759757458 Chr.X: 80442591-80442591 on Build GRCh38
rs1031757178 Chr.X: 80442603-80442603 on Build GRCh38
rs771522107 Chr.X: 80442551-80442551 on Build GRCh38
rs773473520 Chr.X: 80442537-80442537 on Build GRCh38
rs766257081 Chr.X: 80442663-80442663 on Build GRCh38
rs761588244 Chr.X: 80442644-80442644 on Build GRCh38
rs17855533 Chr.X: 80442567-80442567 on Build GRCh38
rs140244380 Chr.X: 80442672-80442672 on Build GRCh38
rs777296127 Chr.X: 80442562-80442562 on Build GRCh38
rs955765761 Chr.X: 80442657-80442657 on Build GRCh38
rs773874789 Chr.X: 80442609-80442609 on Build GRCh38
rs766993924 Chr.X: 80442645-80442645 on Build GRCh38
rs370538052 Chr.X: 80442605-80442605 on Build GRCh38
rs151135944 Chr.X: 80442647-80442647 on Build GRCh38

View Full Product Details