Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98823213-98823478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT12
Entrez Gene ID
79695
Gene Name
polypeptide N-acetylgalactosaminyltransferase 12
Gene Aliases
CRCS1, GalNAc-T12
UniGene
Hs.47099
Gene Chromosome Location
Chr.9: 98807699-98850081 on Build GRCh38
Transcripts
NM_024642.4 XM_006717287.1 XM_011519018.2 XM_011519020.1 XM_017015133.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4686265 c.489C>T p.S163S Chr.9: 98823373-98823373 on Build GRCh38
7165348 c.454C>G p.L152V Chr.9: 98823338-98823338 on Build GRCh38
1458874 c.457C>T p.L153F Chr.9: 98823341-98823341 on Build GRCh38
150648 c.372-7T>A p.? Chr.9: 98823249-98823249 on Build GRCh38
4494988 c.443C>T p.A148V Chr.9: 98823327-98823327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867136237 Chr.9: 98823414-98823414 on Build GRCh38
rs761534603 Chr.9: 98823400-98823400 on Build GRCh38
rs1032747720 Chr.9: 98823430-98823430 on Build GRCh38
rs140383365 Chr.9: 98823356-98823356 on Build GRCh38
rs758204776 Chr.9: 98823295-98823295 on Build GRCh38
rs775071389 Chr.9: 98823263-98823263 on Build GRCh38
rs747224942 Chr.9: 98823388-98823390 on Build GRCh38
rs768255136 Chr.9: 98823388-98823388 on Build GRCh38
rs754099502 Chr.9: 98823444-98823444 on Build GRCh38
rs751656135 Chr.9: 98823326-98823326 on Build GRCh38
rs781218198 Chr.9: 98823234-98823234 on Build GRCh38
rs780049629 Chr.9: 98823412-98823412 on Build GRCh38
rs988917585 Chr.9: 98823455-98823455 on Build GRCh38
rs748355501 Chr.9: 98823435-98823435 on Build GRCh38
rs775040546 Chr.9: 98823375-98823375 on Build GRCh38
rs762919200 Chr.9: 98823419-98823419 on Build GRCh38
rs955730120 Chr.9: 98823451-98823451 on Build GRCh38
rs779388117 Chr.9: 98823449-98823449 on Build GRCh38
rs200879096 Chr.9: 98823385-98823385 on Build GRCh38
rs1014512076 Chr.9: 98823361-98823361 on Build GRCh38
rs749861743 Chr.9: 98823248-98823248 on Build GRCh38
rs746381338 Chr.9: 98823369-98823369 on Build GRCh38
rs146834885 Chr.9: 98823344-98823344 on Build GRCh38
rs138795909 Chr.9: 98823405-98823405 on Build GRCh38
rs906520882 Chr.9: 98823420-98823420 on Build GRCh38
rs374545406 Chr.9: 98823434-98823434 on Build GRCh38
rs757762438 Chr.9: 98823445-98823445 on Build GRCh38
rs143801019 Chr.9: 98823289-98823289 on Build GRCh38
rs779620212 Chr.9: 98823358-98823358 on Build GRCh38
rs370705797 Chr.9: 98823288-98823288 on Build GRCh38
rs943412876 Chr.9: 98823252-98823252 on Build GRCh38
rs778075539 Chr.9: 98823438-98823438 on Build GRCh38
rs755075072 Chr.9: 98823330-98823330 on Build GRCh38
rs374167975 Chr.9: 98823243-98823243 on Build GRCh38
rs771521076 Chr.9: 98823372-98823372 on Build GRCh38
rs201974583 Chr.9: 98823357-98823357 on Build GRCh38
rs2295923 Chr.9: 98823249-98823249 on Build GRCh38
rs774029336 Chr.9: 98823251-98823251 on Build GRCh38
rs757603243 Chr.9: 98823443-98823445 on Build GRCh38
rs756339026 Chr.9: 98823345-98823345 on Build GRCh38
rs781330491 Chr.9: 98823335-98823335 on Build GRCh38
rs199886730 Chr.9: 98823436-98823436 on Build GRCh38
rs1053923044 Chr.9: 98823332-98823332 on Build GRCh38
rs757214097 Chr.9: 98823309-98823309 on Build GRCh38
rs749630939 Chr.9: 98823354-98823354 on Build GRCh38
rs377392016 Chr.9: 98823443-98823443 on Build GRCh38
rs1023064486 Chr.9: 98823365-98823365 on Build GRCh38
rs185007553 Chr.9: 98823242-98823242 on Build GRCh38
rs758912709 Chr.9: 98823257-98823257 on Build GRCh38
rs114902755 Chr.9: 98823259-98823259 on Build GRCh38
rs776411813 Chr.9: 98823398-98823398 on Build GRCh38
rs765088669 Chr.9: 98823311-98823311 on Build GRCh38
rs767545154 Chr.9: 98823427-98823427 on Build GRCh38
rs143127190 Chr.9: 98823276-98823276 on Build GRCh38
rs746409084 Chr.9: 98823454-98823454 on Build GRCh38

View Full Product Details