Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 132784871-132785120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKX6-2
Entrez Gene ID
84504
Gene Name
NK6 homeobox 2
Gene Aliases
GTX, NKX6.2, NKX6B
UniGene
Hs.134013
Gene Chromosome Location
Chr.10: 132783179-132786033 on Build GRCh38
Transcripts
NM_177400.2 XM_017016789.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4556973 c.709G>A p.E237K Chr.10: 132785041-132785041 on Build GRCh38
427270 c.832T>C p.*278R Chr.10: 132784918-132784918 on Build GRCh38
6551479 c.814G>A p.G272S Chr.10: 132784936-132784936 on Build GRCh38
4615411 c.823delG p.D275fs*>3 Chr.10: 132784927-132784927 on Build GRCh38
4651046 c.821G>A p.G274E Chr.10: 132784929-132784929 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762595063 Chr.10: 132785055-132785055 on Build GRCh38
rs764618887 Chr.10: 132785005-132785005 on Build GRCh38
rs564611111 Chr.10: 132785067-132785067 on Build GRCh38
rs778100062 Chr.10: 132785080-132785080 on Build GRCh38
rs761241891 Chr.10: 132784926-132784926 on Build GRCh38
rs768861183 Chr.10: 132784896-132784896 on Build GRCh38
rs774974344 Chr.10: 132785049-132785049 on Build GRCh38
rs755991557 Chr.10: 132785065-132785065 on Build GRCh38
rs757158370 Chr.10: 132785026-132785026 on Build GRCh38
rs753311012 Chr.10: 132784986-132784986 on Build GRCh38
rs777233341 Chr.10: 132785042-132785042 on Build GRCh38
rs749333667 Chr.10: 132784967-132784967 on Build GRCh38
rs774871835 Chr.10: 132784905-132784905 on Build GRCh38
rs375835728 Chr.10: 132785016-132785016 on Build GRCh38
rs766905480 Chr.10: 132784897-132784897 on Build GRCh38
rs138717832 Chr.10: 132785029-132785029 on Build GRCh38
rs1054351142 Chr.10: 132784900-132784900 on Build GRCh38
rs758031501 Chr.10: 132785013-132785013 on Build GRCh38
rs964850833 Chr.10: 132784976-132784976 on Build GRCh38
rs757040969 Chr.10: 132785062-132785062 on Build GRCh38
rs1026380199 Chr.10: 132784947-132784947 on Build GRCh38
rs778577841 Chr.10: 132784943-132784943 on Build GRCh38
rs888062238 Chr.10: 132784953-132784953 on Build GRCh38
rs769846991 Chr.10: 132785011-132785012 on Build GRCh38
rs1006895085 Chr.10: 132784954-132784954 on Build GRCh38
rs754599912 Chr.10: 132784940-132784940 on Build GRCh38
rs772420555 Chr.10: 132785034-132785034 on Build GRCh38
rs146723849 Chr.10: 132785102-132785102 on Build GRCh38
rs376879063 Chr.10: 132785061-132785061 on Build GRCh38
rs371958067 Chr.10: 132785035-132785035 on Build GRCh38
rs758226145 Chr.10: 132784944-132784944 on Build GRCh38
rs1000820788 Chr.10: 132785099-132785099 on Build GRCh38
rs985042447 Chr.10: 132784997-132784997 on Build GRCh38
rs767189347 Chr.10: 132785063-132785063 on Build GRCh38
rs770894115 Chr.10: 132784949-132784949 on Build GRCh38
rs201923471 Chr.10: 132784939-132784939 on Build GRCh38
rs766849675 Chr.10: 132784982-132784982 on Build GRCh38
rs781138890 Chr.10: 132785028-132785028 on Build GRCh38
rs143052208 Chr.10: 132785023-132785023 on Build GRCh38
rs376083581 Chr.10: 132785002-132785002 on Build GRCh38
rs748511291 Chr.10: 132785039-132785040 on Build GRCh38
rs752392318 Chr.10: 132785008-132785008 on Build GRCh38
rs776523212 Chr.10: 132784926-132784927 on Build GRCh38
rs762381662 Chr.10: 132785099-132785101 on Build GRCh38
rs371988067 Chr.10: 132784951-132784951 on Build GRCh38
rs768786455 Chr.10: 132784968-132784968 on Build GRCh38
rs751009544 Chr.10: 132784924-132784924 on Build GRCh38
rs761153531 Chr.10: 132784981-132784981 on Build GRCh38
rs747348604 Chr.10: 132785041-132785041 on Build GRCh38
rs762096371 Chr.10: 132784970-132784970 on Build GRCh38
rs368915941 Chr.10: 132784946-132784946 on Build GRCh38
rs759933944 Chr.10: 132785045-132785045 on Build GRCh38
rs779114105 Chr.10: 132785031-132785031 on Build GRCh38
rs368950766 Chr.10: 132785101-132785101 on Build GRCh38
rs368306294 Chr.10: 132785032-132785032 on Build GRCh38
rs374752014 Chr.10: 132784941-132784941 on Build GRCh38
rs775120314 Chr.10: 132785026-132785026 on Build GRCh38
rs748173584 Chr.10: 132785068-132785068 on Build GRCh38
rs763681042 Chr.10: 132785060-132785060 on Build GRCh38
rs764780626 Chr.10: 132785048-132785048 on Build GRCh38
rs768171810 Chr.10: 132784910-132784910 on Build GRCh38
rs745836424 Chr.10: 132785004-132785006 on Build GRCh38
rs772614153 Chr.10: 132784972-132784972 on Build GRCh38
rs765863998 Chr.10: 132784938-132784938 on Build GRCh38
rs747167400 Chr.10: 132785084-132785084 on Build GRCh38
rs142771323 Chr.10: 132785093-132785093 on Build GRCh38
rs773700509 Chr.10: 132784979-132784979 on Build GRCh38
rs758714609 Chr.10: 132785070-132785070 on Build GRCh38

View Full Product Details