Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129635315-129635566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB6
Entrez Gene ID
140459
Gene Name
ankyrin repeat and SOCS box containing 6
Gene Aliases
-
UniGene
Hs.125037
Gene Chromosome Location
Chr.9: 129634604-129642169 on Build GRCh38
Transcripts
NM_001202403.1 NM_017873.3 NM_177999.2 NM_001286796.1 NM_001286797.1 NM_001286798.1 NM_001286799.1 NM_001286800.1 NM_001286801.1 NM_001286802.1 NM_001286803.1 NM_014064.3 NR_104596.1 XM_005251939.3 XM_017014642.1

Gene Symbol
NTMT1
Entrez Gene ID
28989
Gene Name
N-terminal Xaa-Pro-Lys N-methyltransferase 1
Gene Aliases
AD-003, C9orf32, HOMT1A, METTL11A, NRMT, NTM1A
UniGene
Hs.744027
Gene Chromosome Location
Chr.9: 129608884-129636742 on Build GRCh38
Transcripts
NM_001202403.1 NM_017873.3 NM_177999.2 NM_001286796.1 NM_001286797.1 NM_001286798.1 NM_001286799.1 NM_001286800.1 NM_001286801.1 NM_001286802.1 NM_001286803.1 NM_014064.3 NR_104596.1 XM_005251939.3 XM_017014642.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
455458 c.597C>T p.L199L Chr.9: 129635389-129635389 on Build GRCh38
6730668 c.572G>A p.R191H Chr.9: 129635364-129635364 on Build GRCh38
3375122 c.633C>G p.P211P Chr.9: 129635425-129635425 on Build GRCh38
6730667 c.659T>G p.F220C Chr.9: 129635451-129635451 on Build GRCh38
1105860 c.626A>T p.N209I Chr.9: 129635418-129635418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186769206 Chr.9: 129635440-129635440 on Build GRCh38
rs377409812 Chr.9: 129635413-129635413 on Build GRCh38
rs939123509 Chr.9: 129635529-129635529 on Build GRCh38
rs769882744 Chr.9: 129635345-129635345 on Build GRCh38
rs1039720016 Chr.9: 129635538-129635538 on Build GRCh38
rs755236847 Chr.9: 129635503-129635503 on Build GRCh38
rs751434608 Chr.9: 129635414-129635414 on Build GRCh38
rs747296201 Chr.9: 129635453-129635453 on Build GRCh38
rs901194971 Chr.9: 129635543-129635543 on Build GRCh38
rs753986887 Chr.9: 129635356-129635356 on Build GRCh38
rs757171119 Chr.9: 129635419-129635419 on Build GRCh38
rs964569112 Chr.9: 129635378-129635378 on Build GRCh38
rs367834442 Chr.9: 129635346-129635346 on Build GRCh38
rs559597162 Chr.9: 129635379-129635379 on Build GRCh38
rs201709994 Chr.9: 129635438-129635438 on Build GRCh38
rs374533917 Chr.9: 129635339-129635339 on Build GRCh38
rs766525095 Chr.9: 129635347-129635347 on Build GRCh38
rs148491892 Chr.9: 129635334-129635334 on Build GRCh38
rs145300752 Chr.9: 129635338-129635338 on Build GRCh38
rs372578515 Chr.9: 129635500-129635500 on Build GRCh38
rs371201800 Chr.9: 129635357-129635357 on Build GRCh38
rs779185577 Chr.9: 129635506-129635506 on Build GRCh38
rs776328397 Chr.9: 129635402-129635406 on Build GRCh38
rs745394326 Chr.9: 129635422-129635422 on Build GRCh38
rs749337017 Chr.9: 129635351-129635351 on Build GRCh38
rs762271965 Chr.9: 129635361-129635361 on Build GRCh38
rs780090427 Chr.9: 129635437-129635437 on Build GRCh38
rs997440678 Chr.9: 129635548-129635548 on Build GRCh38
rs746156795 Chr.9: 129635341-129635341 on Build GRCh38
rs769251299 Chr.9: 129635489-129635489 on Build GRCh38
rs748210162 Chr.9: 129635507-129635507 on Build GRCh38
rs779632244 Chr.9: 129635426-129635426 on Build GRCh38
rs769308190 Chr.9: 129635355-129635355 on Build GRCh38
rs55943174 Chr.9: 129635467-129635467 on Build GRCh38
rs748725251 Chr.9: 129635445-129635445 on Build GRCh38
rs769513261 Chr.9: 129635465-129635465 on Build GRCh38
rs935517544 Chr.9: 129635499-129635499 on Build GRCh38
rs772438141 Chr.9: 129635446-129635446 on Build GRCh38
rs151072869 Chr.9: 129635425-129635425 on Build GRCh38
rs141570249 Chr.9: 129635402-129635402 on Build GRCh38
rs369469508 Chr.9: 129635468-129635468 on Build GRCh38
rs775408144 Chr.9: 129635505-129635506 on Build GRCh38
rs766628583 Chr.9: 129635490-129635490 on Build GRCh38
rs541925695 Chr.9: 129635482-129635482 on Build GRCh38
rs140979412 Chr.9: 129635439-129635439 on Build GRCh38
rs139480010 Chr.9: 129635364-129635364 on Build GRCh38
rs773666852 Chr.9: 129635448-129635448 on Build GRCh38
rs758976003 Chr.9: 129635508-129635508 on Build GRCh38
rs761847792 Chr.9: 129635469-129635469 on Build GRCh38
rs772553176 Chr.9: 129635363-129635363 on Build GRCh38
rs752235207 Chr.9: 129635383-129635383 on Build GRCh38
rs78602607 Chr.9: 129635537-129635537 on Build GRCh38
rs767314125 Chr.9: 129635470-129635470 on Build GRCh38
rs759283340 Chr.9: 129635375-129635375 on Build GRCh38
rs755807015 Chr.9: 129635483-129635483 on Build GRCh38
rs755511942 Chr.9: 129635366-129635366 on Build GRCh38

View Full Product Details