Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84657743-84657992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL36
Entrez Gene ID
79786
Gene Name
kelch like family member 36
Gene Aliases
C16orf44
UniGene
Hs.222731
Gene Chromosome Location
Chr.16: 84648504-84667686 on Build GRCh38
Transcripts
NM_001303451.1 NM_024731.3 XM_005256149.2 XM_017023674.1 XM_017023675.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1212659 c.1000C>T p.R334W Chr.16: 84657807-84657807 on Build GRCh38
6145265 c.1004G>T p.R335L Chr.16: 84657811-84657811 on Build GRCh38
4696549 c.1119C>T p.R373R Chr.16: 84657926-84657926 on Build GRCh38
3378012 c.987G>A p.T329T Chr.16: 84657794-84657794 on Build GRCh38
5448922 c.1003C>T p.R335W Chr.16: 84657810-84657810 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771659583 Chr.16: 84657839-84657839 on Build GRCh38
rs374631476 Chr.16: 84657815-84657815 on Build GRCh38
rs770274200 Chr.16: 84657852-84657852 on Build GRCh38
rs763314913 Chr.16: 84657855-84657855 on Build GRCh38
rs747740776 Chr.16: 84657786-84657786 on Build GRCh38
rs200720185 Chr.16: 84657829-84657829 on Build GRCh38
rs774622456 Chr.16: 84657956-84657956 on Build GRCh38
rs943865876 Chr.16: 84657873-84657873 on Build GRCh38
rs897067827 Chr.16: 84657924-84657924 on Build GRCh38
rs1006183080 Chr.16: 84657910-84657910 on Build GRCh38
rs771289520 Chr.16: 84657793-84657793 on Build GRCh38
rs758083086 Chr.16: 84657887-84657887 on Build GRCh38
rs767452225 Chr.16: 84657807-84657807 on Build GRCh38
rs748809986 Chr.16: 84657773-84657773 on Build GRCh38
rs145942400 Chr.16: 84657764-84657764 on Build GRCh38
rs769236608 Chr.16: 84657934-84657934 on Build GRCh38
rs762149427 Chr.16: 84657861-84657861 on Build GRCh38
rs932318591 Chr.16: 84657851-84657851 on Build GRCh38
rs755921577 Chr.16: 84657871-84657871 on Build GRCh38
rs552730604 Chr.16: 84657838-84657838 on Build GRCh38
rs370173265 Chr.16: 84657864-84657864 on Build GRCh38
rs778327445 Chr.16: 84657836-84657836 on Build GRCh38
rs765750342 Chr.16: 84657795-84657795 on Build GRCh38
rs757610434 Chr.16: 84657913-84657913 on Build GRCh38
rs933785145 Chr.16: 84657823-84657823 on Build GRCh38
rs751849523 Chr.16: 84657801-84657801 on Build GRCh38
rs778413315 Chr.16: 84657880-84657880 on Build GRCh38
rs766028929 Chr.16: 84657872-84657872 on Build GRCh38
rs777396169 Chr.16: 84657892-84657892 on Build GRCh38
rs749836425 Chr.16: 84657925-84657925 on Build GRCh38
rs139725379 Chr.16: 84657857-84657857 on Build GRCh38
rs145227614 Chr.16: 84657876-84657876 on Build GRCh38
rs371430023 Chr.16: 84657796-84657796 on Build GRCh38
rs746278354 Chr.16: 84657893-84657893 on Build GRCh38
rs768258861 Chr.16: 84657776-84657776 on Build GRCh38
rs903811176 Chr.16: 84657846-84657846 on Build GRCh38
rs750194993 Chr.16: 84657825-84657825 on Build GRCh38
rs376811926 Chr.16: 84657779-84657779 on Build GRCh38
rs761955063 Chr.16: 84657961-84657961 on Build GRCh38
rs150040885 Chr.16: 84657860-84657860 on Build GRCh38
rs201726115 Chr.16: 84657765-84657765 on Build GRCh38
rs3196203 Chr.16: 84657848-84657848 on Build GRCh38
rs117873154 Chr.16: 84657828-84657828 on Build GRCh38
rs555409936 Chr.16: 84657901-84657901 on Build GRCh38
rs72797514 Chr.16: 84657827-84657827 on Build GRCh38
rs199852251 Chr.16: 84657881-84657881 on Build GRCh38
rs765034976 Chr.16: 84657968-84657969 on Build GRCh38
rs780220325 Chr.16: 84657918-84657918 on Build GRCh38
rs750226879 Chr.16: 84657869-84657869 on Build GRCh38
rs776733336 Chr.16: 84657854-84657854 on Build GRCh38
rs777027054 Chr.16: 84657847-84657847 on Build GRCh38
rs865836636 Chr.16: 84657808-84657808 on Build GRCh38
rs140158625 Chr.16: 84657797-84657797 on Build GRCh38
rs367668625 Chr.16: 84657830-84657830 on Build GRCh38
rs757492004 Chr.16: 84657803-84657803 on Build GRCh38
rs776937288 Chr.16: 84657794-84657794 on Build GRCh38

View Full Product Details