Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 85038080-85038353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRIQ1
Entrez Gene ID
84125
Gene Name
leucine rich repeats and IQ motif containing 1
Gene Aliases
-
UniGene
Hs.402200
Gene Chromosome Location
Chr.12: 85036321-85273004 on Build GRCh38
Transcripts
NM_001079910.1 XM_011538817.2 XM_011538818.2 XM_011538819.2 XM_011538823.2 XM_011538824.2 XM_011538825.2 XM_017020019.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4781598 c.95A>T p.D32V Chr.12: 85038271-85038271 on Build GRCh38
162257 c.106G>C p.E36Q Chr.12: 85038282-85038282 on Build GRCh38
4781597 c.95A>T p.D32V Chr.12: 85038271-85038271 on Build GRCh38
1705994 c.65C>T p.S22F Chr.12: 85038241-85038241 on Build GRCh38
1628886 c.132A>G p.T44T Chr.12: 85038308-85038308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772001444 Chr.12: 85038238-85038238 on Build GRCh38
rs899528349 Chr.12: 85038236-85038236 on Build GRCh38
rs766219757 Chr.12: 85038211-85038211 on Build GRCh38
rs959676214 Chr.12: 85038299-85038299 on Build GRCh38
rs943801503 Chr.12: 85038195-85038195 on Build GRCh38
rs752117291 Chr.12: 85038239-85038239 on Build GRCh38
rs200816060 Chr.12: 85038217-85038217 on Build GRCh38
rs142648811 Chr.12: 85038180-85038180 on Build GRCh38
rs761010014 Chr.12: 85038182-85038182 on Build GRCh38
rs764195939 Chr.12: 85038188-85038188 on Build GRCh38
rs759531826 Chr.12: 85038221-85038221 on Build GRCh38
rs749679666 Chr.12: 85038167-85038167 on Build GRCh38
rs1044605147 Chr.12: 85038314-85038314 on Build GRCh38
rs780226987 Chr.12: 85038272-85038272 on Build GRCh38
rs970896907 Chr.12: 85038282-85038282 on Build GRCh38
rs774849673 Chr.12: 85038173-85038173 on Build GRCh38
rs987657702 Chr.12: 85038139-85038139 on Build GRCh38
rs767102858 Chr.12: 85038237-85038237 on Build GRCh38
rs754551249 Chr.12: 85038145-85038145 on Build GRCh38
rs768732797 Chr.12: 85038274-85038274 on Build GRCh38
rs762063431 Chr.12: 85038298-85038298 on Build GRCh38
rs758449891 Chr.12: 85038127-85038127 on Build GRCh38
rs961981311 Chr.12: 85038146-85038146 on Build GRCh38
rs780561594 Chr.12: 85038149-85038149 on Build GRCh38
rs780631570 Chr.12: 85038281-85038282 on Build GRCh38
rs919838099 Chr.12: 85038110-85038110 on Build GRCh38
rs955108835 Chr.12: 85038125-85038125 on Build GRCh38
rs747891458 Chr.12: 85038155-85038155 on Build GRCh38
rs779053258 Chr.12: 85038183-85038185 on Build GRCh38
rs376466720 Chr.12: 85038172-85038172 on Build GRCh38
rs778240540 Chr.12: 85038165-85038165 on Build GRCh38
rs370626218 Chr.12: 85038197-85038197 on Build GRCh38
rs770904994 Chr.12: 85038302-85038302 on Build GRCh38
rs552263969 Chr.12: 85038140-85038140 on Build GRCh38
rs1054006864 Chr.12: 85038249-85038249 on Build GRCh38
rs532940608 Chr.12: 85038114-85038114 on Build GRCh38
rs7312075 Chr.12: 85038262-85038262 on Build GRCh38
rs753427296 Chr.12: 85038247-85038247 on Build GRCh38
rs772263603 Chr.12: 85038179-85038179 on Build GRCh38
rs894065089 Chr.12: 85038261-85038261 on Build GRCh38
rs746304363 Chr.12: 85038174-85038174 on Build GRCh38
rs745961853 Chr.12: 85038232-85038255 on Build GRCh38
rs747089819 Chr.12: 85038273-85038273 on Build GRCh38
rs766450698 Chr.12: 85038129-85038129 on Build GRCh38
rs777007740 Chr.12: 85038294-85038294 on Build GRCh38
rs755698692 Chr.12: 85038157-85038157 on Build GRCh38
rs777309605 Chr.12: 85038243-85038243 on Build GRCh38
rs772440195 Chr.12: 85038269-85038269 on Build GRCh38
rs746435033 Chr.12: 85038263-85038263 on Build GRCh38
rs755610150 Chr.12: 85038242-85038242 on Build GRCh38
rs776873315 Chr.12: 85038189-85038189 on Build GRCh38
rs774262709 Chr.12: 85038324-85038324 on Build GRCh38

View Full Product Details