Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 14165320-14165549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIPI
Entrez Gene ID
149998
Gene Name
lipase I
Gene Aliases
CT17, LPDL, PLA1C, PRED5, mPA-PLA1 beta
UniGene
Hs.139907
Gene Chromosome Location
Chr.21: 14108813-14210953 on Build GRCh38
Transcripts
NM_001302998.1 NM_001302999.1 NM_001303000.1 NM_001303001.1 NM_198996.3 XM_006723965.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5914363 c.797-6C>T p.? Chr.21: 14165396-14165396 on Build GRCh38
7425164 c.828A>T p.R276S Chr.21: 14165359-14165359 on Build GRCh38
6539504 c.866G>C p.C289S Chr.21: 14165321-14165321 on Build GRCh38
5858429 c.827G>A p.R276K Chr.21: 14165360-14165360 on Build GRCh38
1029434 c.803A>G p.Q268R Chr.21: 14165384-14165384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756197762 Chr.21: 14165364-14165364 on Build GRCh38
rs903964168 Chr.21: 14165427-14165427 on Build GRCh38
rs183861681 Chr.21: 14165420-14165420 on Build GRCh38
rs371755788 Chr.21: 14165350-14165350 on Build GRCh38
rs912346740 Chr.21: 14165480-14165480 on Build GRCh38
rs777078641 Chr.21: 14165437-14165437 on Build GRCh38
rs998719962 Chr.21: 14165372-14165372 on Build GRCh38
rs368302427 Chr.21: 14165438-14165438 on Build GRCh38
rs75730972 Chr.21: 14165406-14165406 on Build GRCh38
rs757696384 Chr.21: 14165381-14165381 on Build GRCh38
rs779266298 Chr.21: 14165382-14165382 on Build GRCh38
rs773197231 Chr.21: 14165413-14165413 on Build GRCh38
rs746546020 Chr.21: 14165392-14165392 on Build GRCh38
rs772222280 Chr.21: 14165425-14165425 on Build GRCh38
rs757935220 Chr.21: 14165410-14165411 on Build GRCh38
rs780806532 Chr.21: 14165400-14165400 on Build GRCh38
rs889699322 Chr.21: 14165517-14165517 on Build GRCh38
rs759591291 Chr.21: 14165399-14165400 on Build GRCh38
rs752203308 Chr.21: 14165400-14165401 on Build GRCh38
rs765265767 Chr.21: 14165401-14165401 on Build GRCh38
rs774673432 Chr.21: 14165412-14165412 on Build GRCh38
rs907030769 Chr.21: 14165352-14165352 on Build GRCh38
rs986511634 Chr.21: 14165457-14165457 on Build GRCh38
rs764031915 Chr.21: 14165368-14165368 on Build GRCh38
rs750775979 Chr.21: 14165383-14165383 on Build GRCh38
rs546295944 Chr.21: 14165461-14165461 on Build GRCh38
rs867097666 Chr.21: 14165390-14165390 on Build GRCh38
rs149286024 Chr.21: 14165379-14165379 on Build GRCh38
rs779729305 Chr.21: 14165386-14165386 on Build GRCh38
rs559394379 Chr.21: 14165473-14165473 on Build GRCh38
rs957310833 Chr.21: 14165409-14165409 on Build GRCh38
rs769795834 Chr.21: 14165410-14165410 on Build GRCh38
rs572908058 Chr.21: 14165485-14165485 on Build GRCh38
rs768230006 Chr.21: 14165393-14165393 on Build GRCh38

View Full Product Details